Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117433351-117433857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCAML1
Entrez Gene ID
57453
Gene Name
DS cell adhesion molecule like 1
Gene Aliases
DSCAM2
UniGene
Hs.659513
Gene Chromosome Location
Chr.11: 117427772-117797261 on Build GRCh38
Transcripts
NM_020693.3 XM_011542917.1 XM_011542918.1 XM_011542919.2 XM_011542920.2 XM_011542921.2 XM_011542924.2 XM_011542925.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
380847 c.5074G>C p.E1692Q Chr.11: 117433454-117433454 on Build GRCh38
5507640 c.5057-4G>T p.? Chr.11: 117433475-117433475 on Build GRCh38
5626415 c.5079G>T p.M1693I Chr.11: 117433449-117433449 on Build GRCh38
5371732 c.5058T>G p.D1686E Chr.11: 117433470-117433470 on Build GRCh38
3443801 c.5064G>A p.K1688K Chr.11: 117433464-117433464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013489422 Chr.11: 117433748-117433748 on Build GRCh38
rs532265217 Chr.11: 117433761-117433761 on Build GRCh38
rs1046484430 Chr.11: 117433764-117433764 on Build GRCh38
rs963513570 Chr.11: 117433808-117433808 on Build GRCh38
rs1012254746 Chr.11: 117433422-117433422 on Build GRCh38
rs148374689 Chr.11: 117433645-117433645 on Build GRCh38
rs896573337 Chr.11: 117433811-117433811 on Build GRCh38
rs757423317 Chr.11: 117433431-117433432 on Build GRCh38
rs748491153 Chr.11: 117433423-117433423 on Build GRCh38
rs910589085 Chr.11: 117433573-117433573 on Build GRCh38
rs972279702 Chr.11: 117433432-117433432 on Build GRCh38
rs764139859 Chr.11: 117433819-117433819 on Build GRCh38
rs375428721 Chr.11: 117433430-117433430 on Build GRCh38
rs779480217 Chr.11: 117433395-117433395 on Build GRCh38
rs1016804484 Chr.11: 117433380-117433380 on Build GRCh38
rs772397754 Chr.11: 117433511-117433511 on Build GRCh38
rs35399839 Chr.11: 117433458-117433458 on Build GRCh38
rs999372393 Chr.11: 117433793-117433793 on Build GRCh38
rs754268526 Chr.11: 117433503-117433503 on Build GRCh38
rs574553184 Chr.11: 117433675-117433675 on Build GRCh38
rs534480765 Chr.11: 117433807-117433807 on Build GRCh38
rs185498972 Chr.11: 117433577-117433577 on Build GRCh38
rs1035119932 Chr.11: 117433396-117433396 on Build GRCh38
rs751075167 Chr.11: 117433399-117433399 on Build GRCh38
rs936522346 Chr.11: 117433651-117433651 on Build GRCh38
rs543224986 Chr.11: 117433677-117433677 on Build GRCh38
rs775894349 Chr.11: 117433516-117433516 on Build GRCh38
rs754586952 Chr.11: 117433409-117433409 on Build GRCh38
rs79617322 Chr.11: 117433776-117433776 on Build GRCh38
rs7927139 Chr.11: 117433711-117433711 on Build GRCh38
rs759707747 Chr.11: 117433510-117433510 on Build GRCh38
rs892366418 Chr.11: 117433725-117433725 on Build GRCh38
rs554380268 Chr.11: 117433646-117433646 on Build GRCh38
rs780846692 Chr.11: 117433417-117433417 on Build GRCh38
rs539036589 Chr.11: 117433526-117433526 on Build GRCh38
rs1044017343 Chr.11: 117433768-117433768 on Build GRCh38
rs771343543 Chr.11: 117433482-117433482 on Build GRCh38
rs558478955 Chr.11: 117433556-117433556 on Build GRCh38
rs960752421 Chr.11: 117433412-117433412 on Build GRCh38
rs1002326892 Chr.11: 117433785-117433785 on Build GRCh38
rs1012740515 Chr.11: 117433828-117433828 on Build GRCh38
rs759783554 Chr.11: 117433628-117433628 on Build GRCh38
rs1043151582 Chr.11: 117433644-117433644 on Build GRCh38
rs986903889 Chr.11: 117433612-117433612 on Build GRCh38
rs974721083 Chr.11: 117433551-117433551 on Build GRCh38
rs747434149 Chr.11: 117433406-117433406 on Build GRCh38
rs963546244 Chr.11: 117433387-117433387 on Build GRCh38
rs2242133 Chr.11: 117433480-117433480 on Build GRCh38
rs987866206 Chr.11: 117433522-117433522 on Build GRCh38
rs754153890 Chr.11: 117433407-117433408 on Build GRCh38
rs921588617 Chr.11: 117433709-117433709 on Build GRCh38
rs918788910 Chr.11: 117433572-117433572 on Build GRCh38
rs538490881 Chr.11: 117433643-117433643 on Build GRCh38
rs559387180 Chr.11: 117433800-117433800 on Build GRCh38

View Full Product Details