Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119190619-119191050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC153
Entrez Gene ID
283152
Gene Name
coiled-coil domain containing 153
Gene Aliases
-
UniGene
Hs.114777
Gene Chromosome Location
Chr.11: 119190254-119206232 on Build GRCh38
Transcripts
NM_001145018.1 XM_006718818.3 XM_011542763.2 XM_011542764.2 XM_011542770.2 XM_011542771.2 XM_011542772.2 XM_011542774.2 XM_017017577.1 XM_017017578.1 XM_017017579.1 XM_017017580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6621742 c.459G>A p.A153A Chr.11: 119190731-119190731 on Build GRCh38
3444151 c.156C>T p.D52D Chr.11: 119190776-119190776 on Build GRCh38
5500029 c.151C>A p.R51R Chr.11: 119190781-119190781 on Build GRCh38
5500028 c.409C>A p.R137R Chr.11: 119190781-119190781 on Build GRCh38
3444149 c.208G>A p.E70K Chr.11: 119190724-119190724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372673954 Chr.11: 119190851-119190851 on Build GRCh38
rs577239774 Chr.11: 119191007-119191007 on Build GRCh38
rs551117674 Chr.11: 119190676-119190676 on Build GRCh38
rs774646680 Chr.11: 119190781-119190781 on Build GRCh38
rs763519154 Chr.11: 119190887-119190887 on Build GRCh38
rs772954493 Chr.11: 119190862-119190862 on Build GRCh38
rs879528139 Chr.11: 119190699-119190699 on Build GRCh38
rs746676207 Chr.11: 119190748-119190748 on Build GRCh38
rs770115149 Chr.11: 119190694-119190695 on Build GRCh38
rs145189171 Chr.11: 119190780-119190780 on Build GRCh38
rs758003793 Chr.11: 119190691-119190691 on Build GRCh38
rs766751811 Chr.11: 119190696-119190696 on Build GRCh38
rs779474924 Chr.11: 119190845-119190845 on Build GRCh38
rs1056297463 Chr.11: 119190747-119190747 on Build GRCh38
rs142300259 Chr.11: 119190970-119190970 on Build GRCh38
rs751209983 Chr.11: 119190823-119190823 on Build GRCh38
rs763382937 Chr.11: 119190648-119190648 on Build GRCh38
rs760300390 Chr.11: 119190788-119190788 on Build GRCh38
rs754683401 Chr.11: 119190824-119190824 on Build GRCh38
rs749829167 Chr.11: 119190838-119190838 on Build GRCh38
rs896754324 Chr.11: 119190909-119190909 on Build GRCh38
rs1045032924 Chr.11: 119190937-119190937 on Build GRCh38
rs898530138 Chr.11: 119190956-119190956 on Build GRCh38
rs770253730 Chr.11: 119190771-119190771 on Build GRCh38
rs201913499 Chr.11: 119190810-119190810 on Build GRCh38
rs1019520613 Chr.11: 119191021-119191021 on Build GRCh38
rs544567745 Chr.11: 119191020-119191020 on Build GRCh38
rs779631788 Chr.11: 119190733-119190733 on Build GRCh38
rs760622411 Chr.11: 119190858-119190858 on Build GRCh38
rs1025891252 Chr.11: 119190683-119190683 on Build GRCh38
rs766168472 Chr.11: 119190811-119190811 on Build GRCh38
rs748676783 Chr.11: 119190770-119190770 on Build GRCh38
rs143016004 Chr.11: 119190731-119190731 on Build GRCh38
rs985703144 Chr.11: 119190969-119190969 on Build GRCh38
rs773506973 Chr.11: 119190661-119190661 on Build GRCh38
rs1056702838 Chr.11: 119191022-119191022 on Build GRCh38
rs530437204 Chr.11: 119190917-119190917 on Build GRCh38
rs951258607 Chr.11: 119191012-119191012 on Build GRCh38
rs775813345 Chr.11: 119190854-119190854 on Build GRCh38
rs759900677 Chr.11: 119190783-119190783 on Build GRCh38
rs753807225 Chr.11: 119190705-119190705 on Build GRCh38
rs999383610 Chr.11: 119190737-119190737 on Build GRCh38
rs771415747 Chr.11: 119190843-119190843 on Build GRCh38
rs138528075 Chr.11: 119190710-119190710 on Build GRCh38
rs778096962 Chr.11: 119190836-119190836 on Build GRCh38
rs1012477787 Chr.11: 119190927-119190927 on Build GRCh38
rs752193104 Chr.11: 119190828-119190828 on Build GRCh38
rs200006921 Chr.11: 119190801-119190801 on Build GRCh38
rs759157685 Chr.11: 119190892-119190892 on Build GRCh38
rs765044196 Chr.11: 119190790-119190790 on Build GRCh38
rs780635321 Chr.11: 119190757-119190757 on Build GRCh38
rs138881906 Chr.11: 119190787-119190787 on Build GRCh38
rs911184233 Chr.11: 119190671-119190671 on Build GRCh38
rs768793794 Chr.11: 119190861-119190861 on Build GRCh38
rs746062897 Chr.11: 119190847-119190847 on Build GRCh38
rs780731762 Chr.11: 119190826-119190826 on Build GRCh38
rs1014647836 Chr.11: 119190830-119190830 on Build GRCh38
rs751404323 Chr.11: 119190881-119190881 on Build GRCh38
rs144583320 Chr.11: 119190709-119190709 on Build GRCh38
rs1050196868 Chr.11: 119190657-119190657 on Build GRCh38
rs569056721 Chr.11: 119190660-119190660 on Build GRCh38
rs963681279 Chr.11: 119190744-119190744 on Build GRCh38
rs534516706 Chr.11: 119190964-119190964 on Build GRCh38
rs767707599 Chr.11: 119190701-119190701 on Build GRCh38
rs773599416 Chr.11: 119190772-119190772 on Build GRCh38
rs754420871 Chr.11: 119190756-119190756 on Build GRCh38
rs774749032 Chr.11: 119190698-119190698 on Build GRCh38

View Full Product Details