Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32576182-32576678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD4
Entrez Gene ID
121512
Gene Name
FYVE, RhoGEF and PH domain containing 4
Gene Aliases
CMT4H, FRABP, ZFYVE6
UniGene
Hs.117835
Gene Chromosome Location
Chr.12: 32399523-32646050 on Build GRCh38
Transcripts
NM_001304480.1 NM_001304481.1 NM_001304483.1 NM_001304484.1 NM_139241.3 XM_005253304.4 XM_005253307.3 XM_005253308.4 XM_005253309.1 XM_011520554.1 XM_011520555.1 XM_011520556.1 XM_011520557.1 XM_017018803.1 XM_017018804.1 XM_017018805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4429369 c.64G>T p.D22Y Chr.12: 32576421-32576421 on Build GRCh38
1299323 c.29C>G p.S10C Chr.12: 32576386-32576386 on Build GRCh38
6531469 c.64G>C p.D22H Chr.12: 32576421-32576421 on Build GRCh38
1207028 c.14A>G p.K5R Chr.12: 32576371-32576371 on Build GRCh38
357208 c.32G>T p.C11F Chr.12: 32576389-32576389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748985432 Chr.12: 32576221-32576222 on Build GRCh38
rs951445911 Chr.12: 32576279-32576279 on Build GRCh38
rs374604191 Chr.12: 32576353-32576353 on Build GRCh38
rs199744649 Chr.12: 32576322-32576322 on Build GRCh38
rs540449675 Chr.12: 32576434-32576434 on Build GRCh38
rs755688214 Chr.12: 32576242-32576242 on Build GRCh38
rs200681247 Chr.12: 32576456-32576456 on Build GRCh38
rs896975962 Chr.12: 32576621-32576621 on Build GRCh38
rs1008743458 Chr.12: 32576311-32576311 on Build GRCh38
rs1015811800 Chr.12: 32576258-32576258 on Build GRCh38
rs758603361 Chr.12: 32576433-32576433 on Build GRCh38
rs769409352 Chr.12: 32576226-32576226 on Build GRCh38
rs1054695516 Chr.12: 32576526-32576526 on Build GRCh38
rs755068251 Chr.12: 32576415-32576415 on Build GRCh38
rs749179569 Chr.12: 32576494-32576494 on Build GRCh38
rs768347955 Chr.12: 32576496-32576496 on Build GRCh38
rs745872665 Chr.12: 32576346-32576346 on Build GRCh38
rs552886155 Chr.12: 32576564-32576564 on Build GRCh38
rs879100859 Chr.12: 32576534-32576534 on Build GRCh38
rs770713248 Chr.12: 32576339-32576339 on Build GRCh38
rs751006919 Chr.12: 32576302-32576302 on Build GRCh38
rs779132042 Chr.12: 32576319-32576319 on Build GRCh38
rs998149065 Chr.12: 32576639-32576639 on Build GRCh38
rs762290444 Chr.12: 32576290-32576290 on Build GRCh38
rs748732880 Chr.12: 32576387-32576387 on Build GRCh38
rs913587128 Chr.12: 32576429-32576429 on Build GRCh38
rs997411831 Chr.12: 32576656-32576656 on Build GRCh38
rs543106084 Chr.12: 32576645-32576645 on Build GRCh38
rs745428859 Chr.12: 32576486-32576487 on Build GRCh38
rs770282468 Chr.12: 32576374-32576379 on Build GRCh38
rs774846804 Chr.12: 32576239-32576239 on Build GRCh38
rs751215632 Chr.12: 32576287-32576287 on Build GRCh38
rs766829554 Chr.12: 32576386-32576386 on Build GRCh38
rs756566376 Chr.12: 32576303-32576303 on Build GRCh38
rs753832597 Chr.12: 32576312-32576312 on Build GRCh38
rs562775953 Chr.12: 32576655-32576655 on Build GRCh38
rs369365466 Chr.12: 32576379-32576379 on Build GRCh38
rs761554382 Chr.12: 32576368-32576368 on Build GRCh38
rs769594154 Chr.12: 32576347-32576347 on Build GRCh38
rs374980260 Chr.12: 32576376-32576376 on Build GRCh38
rs878977994 Chr.12: 32576541-32576541 on Build GRCh38
rs755013368 Chr.12: 32576318-32576318 on Build GRCh38
rs773986552 Chr.12: 32576498-32576498 on Build GRCh38
rs928202784 Chr.12: 32576481-32576481 on Build GRCh38
rs975646718 Chr.12: 32576388-32576388 on Build GRCh38
rs780841698 Chr.12: 32576345-32576345 on Build GRCh38
rs768231391 Chr.12: 32576359-32576359 on Build GRCh38
rs773908114 Chr.12: 32576482-32576484 on Build GRCh38
rs771978622 Chr.12: 32576491-32576492 on Build GRCh38
rs578120751 Chr.12: 32576641-32576641 on Build GRCh38
rs552158609 Chr.12: 32576630-32576630 on Build GRCh38
rs1037529663 Chr.12: 32576544-32576544 on Build GRCh38
rs866663556 Chr.12: 32576605-32576605 on Build GRCh38
rs751968860 Chr.12: 32576216-32576216 on Build GRCh38
rs779877075 Chr.12: 32576468-32576468 on Build GRCh38
rs958305546 Chr.12: 32576411-32576411 on Build GRCh38
rs889670119 Chr.12: 32576644-32576644 on Build GRCh38
rs897098057 Chr.12: 32576283-32576283 on Build GRCh38
rs989660324 Chr.12: 32576418-32576418 on Build GRCh38
rs556668619 Chr.12: 32576541-32576542 on Build GRCh38
rs371407163 Chr.12: 32576407-32576407 on Build GRCh38
rs553969089 Chr.12: 32576487-32576487 on Build GRCh38
rs537787603 Chr.12: 32576218-32576218 on Build GRCh38
rs775103873 Chr.12: 32576351-32576351 on Build GRCh38
rs759320695 Chr.12: 32576221-32576221 on Build GRCh38
rs756801805 Chr.12: 32576587-32576587 on Build GRCh38
rs765238074 Chr.12: 32576424-32576424 on Build GRCh38
rs267603439 Chr.12: 32576372-32576372 on Build GRCh38
rs780231723 Chr.12: 32576306-32576306 on Build GRCh38
rs755897211 Chr.12: 32576467-32576467 on Build GRCh38
rs574053963 Chr.12: 32576598-32576598 on Build GRCh38
rs774159977 Chr.12: 32576364-32576364 on Build GRCh38
rs763807390 Chr.12: 32576255-32576255 on Build GRCh38
rs1047024051 Chr.12: 32576626-32576626 on Build GRCh38
rs762713946 Chr.12: 32576244-32576244 on Build GRCh38
rs149584314 Chr.12: 32576381-32576381 on Build GRCh38
rs755380148 Chr.12: 32576298-32576298 on Build GRCh38

View Full Product Details