Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 82399131-82399642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL25
Entrez Gene ID
84190
Gene Name
methyltransferase like 25
Gene Aliases
C12orf26
UniGene
Hs.506222
Gene Chromosome Location
Chr.12: 82358497-82479239 on Build GRCh38
Transcripts
NM_001319675.1 NM_032230.2 XM_005269186.4 XM_011538827.2 XM_011538828.2 XM_011538829.2 XM_017020020.1 XM_017020021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5804182 c.1027A>T p.R343* Chr.12: 82399290-82399290 on Build GRCh38
4966918 c.919T>C p.C307R Chr.12: 82399182-82399182 on Build GRCh38
6711688 c.914A>C p.N305T Chr.12: 82399177-82399177 on Build GRCh38
943959 c.1082C>A p.S361Y Chr.12: 82399345-82399345 on Build GRCh38
7206440 c.1121A>C p.K374T Chr.12: 82399384-82399384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145668507 Chr.12: 82399288-82399288 on Build GRCh38
rs767060142 Chr.12: 82399470-82399470 on Build GRCh38
rs376484379 Chr.12: 82399353-82399353 on Build GRCh38
rs751689941 Chr.12: 82399377-82399379 on Build GRCh38
rs904737662 Chr.12: 82399456-82399456 on Build GRCh38
rs762620251 Chr.12: 82399240-82399240 on Build GRCh38
rs756250375 Chr.12: 82399167-82399167 on Build GRCh38
rs866661596 Chr.12: 82399235-82399235 on Build GRCh38
rs1053527757 Chr.12: 82399168-82399168 on Build GRCh38
rs758368862 Chr.12: 82399499-82399499 on Build GRCh38
rs368946462 Chr.12: 82399306-82399306 on Build GRCh38
rs370253053 Chr.12: 82399230-82399230 on Build GRCh38
rs777251083 Chr.12: 82399436-82399436 on Build GRCh38
rs527617328 Chr.12: 82399617-82399617 on Build GRCh38
rs756087182 Chr.12: 82399440-82399440 on Build GRCh38
rs759798144 Chr.12: 82399257-82399257 on Build GRCh38
rs746875884 Chr.12: 82399300-82399300 on Build GRCh38
rs752666077 Chr.12: 82399435-82399435 on Build GRCh38
rs909768027 Chr.12: 82399208-82399208 on Build GRCh38
rs963349541 Chr.12: 82399209-82399209 on Build GRCh38
rs200406987 Chr.12: 82399223-82399223 on Build GRCh38
rs760014184 Chr.12: 82399352-82399352 on Build GRCh38
rs758069802 Chr.12: 82399252-82399252 on Build GRCh38
rs1019776932 Chr.12: 82399212-82399212 on Build GRCh38
rs200598895 Chr.12: 82399386-82399386 on Build GRCh38
rs776163830 Chr.12: 82399374-82399374 on Build GRCh38
rs768683672 Chr.12: 82399225-82399225 on Build GRCh38
rs756275185 Chr.12: 82399261-82399261 on Build GRCh38
rs780327141 Chr.12: 82399255-82399255 on Build GRCh38
rs925974512 Chr.12: 82399479-82399479 on Build GRCh38
rs984351885 Chr.12: 82399315-82399315 on Build GRCh38
rs749469219 Chr.12: 82399195-82399195 on Build GRCh38
rs780918555 Chr.12: 82399434-82399434 on Build GRCh38
rs868675683 Chr.12: 82399371-82399371 on Build GRCh38
rs12821414 Chr.12: 82399396-82399396 on Build GRCh38
rs754759042 Chr.12: 82399304-82399304 on Build GRCh38
rs766687433 Chr.12: 82399253-82399253 on Build GRCh38
rs12816597 Chr.12: 82399490-82399490 on Build GRCh38
rs774821544 Chr.12: 82399255-82399255 on Build GRCh38
rs750548938 Chr.12: 82399390-82399390 on Build GRCh38
rs150163659 Chr.12: 82399203-82399203 on Build GRCh38
rs373368121 Chr.12: 82399328-82399328 on Build GRCh38
rs144861076 Chr.12: 82399233-82399233 on Build GRCh38
rs779857890 Chr.12: 82399221-82399221 on Build GRCh38
rs936028682 Chr.12: 82399526-82399526 on Build GRCh38
rs891574319 Chr.12: 82399586-82399586 on Build GRCh38
rs10506873 Chr.12: 82399163-82399163 on Build GRCh38
rs912063393 Chr.12: 82399370-82399370 on Build GRCh38
rs754785415 Chr.12: 82399424-82399424 on Build GRCh38
rs190335664 Chr.12: 82399375-82399375 on Build GRCh38
rs12821609 Chr.12: 82399453-82399453 on Build GRCh38
rs753996337 Chr.12: 82399377-82399377 on Build GRCh38
rs78590216 Chr.12: 82399275-82399275 on Build GRCh38
rs778978154 Chr.12: 82399294-82399294 on Build GRCh38
rs565370896 Chr.12: 82399469-82399469 on Build GRCh38
rs757536636 Chr.12: 82399290-82399290 on Build GRCh38
rs768108659 Chr.12: 82399162-82399162 on Build GRCh38
rs543602656 Chr.12: 82399463-82399463 on Build GRCh38
rs1052390392 Chr.12: 82399332-82399332 on Build GRCh38
rs1039296219 Chr.12: 82399452-82399452 on Build GRCh38
rs980035026 Chr.12: 82399201-82399201 on Build GRCh38
rs769318670 Chr.12: 82399317-82399317 on Build GRCh38
rs141543492 Chr.12: 82399333-82399333 on Build GRCh38
rs1025325475 Chr.12: 82399238-82399238 on Build GRCh38
rs748025152 Chr.12: 82399313-82399313 on Build GRCh38
rs753187918 Chr.12: 82399259-82399259 on Build GRCh38
rs34765930 Chr.12: 82399170-82399170 on Build GRCh38
rs748716939 Chr.12: 82399330-82399330 on Build GRCh38
rs772866839 Chr.12: 82399236-82399236 on Build GRCh38
rs375196888 Chr.12: 82399376-82399376 on Build GRCh38
rs965199963 Chr.12: 82399228-82399228 on Build GRCh38
rs977364136 Chr.12: 82399460-82399460 on Build GRCh38
rs183242625 Chr.12: 82399601-82399601 on Build GRCh38
rs768198375 Chr.12: 82399258-82399258 on Build GRCh38
rs770667481 Chr.12: 82399350-82399350 on Build GRCh38
rs113859754 Chr.12: 82399305-82399305 on Build GRCh38
rs545179515 Chr.12: 82399577-82399577 on Build GRCh38

View Full Product Details