Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119499968-119500464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC60
Entrez Gene ID
160777
Gene Name
coiled-coil domain containing 60
Gene Aliases
-
UniGene
Hs.98188
Gene Chromosome Location
Chr.12: 119334712-119541047 on Build GRCh38
Transcripts
NM_178499.4 XM_017018911.1 XM_017018912.1 XM_017018913.1 XM_017018914.1 XM_017018916.1

Gene Symbol
LOC105370027
Entrez Gene ID
105370027
Gene Name
uncharacterized LOC105370027
Gene Aliases
-
UniGene
Hs.720596
Gene Chromosome Location
Chr.12: 119387991-119668006 on Build GRCh38
Transcripts
NM_178499.4 XM_017018911.1 XM_017018912.1 XM_017018913.1 XM_017018914.1 XM_017018916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4669975 c.642G>A p.A214A Chr.12: 119500162-119500162 on Build GRCh38
4501641 c.594C>T p.I198I Chr.12: 119500114-119500114 on Build GRCh38
5021777 c.648+10A>C p.? Chr.12: 119500178-119500178 on Build GRCh38
4501204 c.582C>T p.T194T Chr.12: 119500102-119500102 on Build GRCh38
6135639 c.633C>G p.F211L Chr.12: 119500153-119500153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs482802 Chr.12: 119500178-119500178 on Build GRCh38
rs932212435 Chr.12: 119500074-119500074 on Build GRCh38
rs774090572 Chr.12: 119500068-119500068 on Build GRCh38
rs1011634932 Chr.12: 119500240-119500240 on Build GRCh38
rs374562709 Chr.12: 119500187-119500187 on Build GRCh38
rs779918876 Chr.12: 119500195-119500195 on Build GRCh38
rs771991228 Chr.12: 119500325-119500325 on Build GRCh38
rs1055085666 Chr.12: 119500098-119500098 on Build GRCh38
rs977895549 Chr.12: 119500249-119500249 on Build GRCh38
rs924057810 Chr.12: 119500258-119500258 on Build GRCh38
rs141099915 Chr.12: 119500161-119500161 on Build GRCh38
rs573012137 Chr.12: 119500057-119500057 on Build GRCh38
rs759057262 Chr.12: 119500105-119500106 on Build GRCh38
rs771836946 Chr.12: 119500194-119500194 on Build GRCh38
rs570601119 Chr.12: 119500372-119500372 on Build GRCh38
rs887915749 Chr.12: 119500090-119500090 on Build GRCh38
rs536887106 Chr.12: 119500337-119500337 on Build GRCh38
rs1727390 Chr.12: 119500205-119500205 on Build GRCh38
rs765656867 Chr.12: 119500126-119500126 on Build GRCh38
rs894841059 Chr.12: 119500405-119500405 on Build GRCh38
rs930665376 Chr.12: 119500340-119500340 on Build GRCh38
rs756954214 Chr.12: 119500184-119500184 on Build GRCh38
rs1040365546 Chr.12: 119500021-119500021 on Build GRCh38
rs566273271 Chr.12: 119500238-119500238 on Build GRCh38
rs773772906 Chr.12: 119500138-119500138 on Build GRCh38
rs960976027 Chr.12: 119500323-119500323 on Build GRCh38
rs149890067 Chr.12: 119500047-119500047 on Build GRCh38
rs1022211135 Chr.12: 119500438-119500438 on Build GRCh38
rs763543436 Chr.12: 119500209-119500209 on Build GRCh38
rs474748 Chr.12: 119500326-119500326 on Build GRCh38
rs768223723 Chr.12: 119500199-119500199 on Build GRCh38
rs1011851753 Chr.12: 119500426-119500426 on Build GRCh38
rs767127768 Chr.12: 119500106-119500106 on Build GRCh38
rs552488615 Chr.12: 119500222-119500222 on Build GRCh38
rs114007573 Chr.12: 119500404-119500404 on Build GRCh38
rs542109030 Chr.12: 119500084-119500084 on Build GRCh38
rs921929895 Chr.12: 119500305-119500305 on Build GRCh38
rs928131940 Chr.12: 119500308-119500308 on Build GRCh38
rs756287549 Chr.12: 119500181-119500181 on Build GRCh38
rs372681245 Chr.12: 119500162-119500162 on Build GRCh38
rs763167241 Chr.12: 119500143-119500143 on Build GRCh38
rs939595698 Chr.12: 119500093-119500093 on Build GRCh38
rs79268044 Chr.12: 119500185-119500185 on Build GRCh38
rs944771323 Chr.12: 119500279-119500279 on Build GRCh38
rs978262952 Chr.12: 119500301-119500301 on Build GRCh38
rs761219364 Chr.12: 119500335-119500335 on Build GRCh38
rs753829749 Chr.12: 119500183-119500183 on Build GRCh38
rs539930121 Chr.12: 119500394-119500394 on Build GRCh38
rs868642901 Chr.12: 119500136-119500136 on Build GRCh38
rs543112061 Chr.12: 119499995-119499995 on Build GRCh38
rs369442968 Chr.12: 119500083-119500083 on Build GRCh38
rs539859943 Chr.12: 119500338-119500338 on Build GRCh38
rs1727392 Chr.12: 119500439-119500439 on Build GRCh38
rs773798030 Chr.12: 119500066-119500066 on Build GRCh38
rs202186474 Chr.12: 119500182-119500182 on Build GRCh38
rs771463517 Chr.12: 119500217-119500217 on Build GRCh38
rs966921485 Chr.12: 119499996-119499996 on Build GRCh38
rs576951845 Chr.12: 119500011-119500011 on Build GRCh38
rs775117225 Chr.12: 119500103-119500103 on Build GRCh38
rs201674961 Chr.12: 119500116-119500116 on Build GRCh38
rs80155680 Chr.12: 119500243-119500243 on Build GRCh38
rs755987121 Chr.12: 119500055-119500055 on Build GRCh38
rs543711905 Chr.12: 119500387-119500387 on Build GRCh38
rs144893545 Chr.12: 119500280-119500280 on Build GRCh38
rs1022157146 Chr.12: 119500239-119500239 on Build GRCh38
rs545675836 Chr.12: 119500033-119500033 on Build GRCh38
rs777827065 Chr.12: 119500056-119500056 on Build GRCh38
rs767780495 Chr.12: 119500177-119500177 on Build GRCh38
rs907986610 Chr.12: 119500421-119500421 on Build GRCh38
rs556980923 Chr.12: 119500342-119500342 on Build GRCh38
rs932232930 Chr.12: 119500307-119500307 on Build GRCh38

View Full Product Details