Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119701539-119702046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIT
Entrez Gene ID
11113
Gene Name
citron rho-interacting serine/threonine kinase
Gene Aliases
CRIK, STK21
UniGene
Hs.119594
Gene Chromosome Location
Chr.12: 119685790-119877318 on Build GRCh38
Transcripts
NM_001206999.1 NM_007174.2 XM_006719206.2 XM_011537783.1 XM_011537784.1 XM_011537785.1 XM_011537787.1 XM_011537788.1 XM_011537789.1 XM_011537790.2 XM_011537791.2 XM_011537792.2 XM_017018735.1 XM_017018736.1 XM_017018737.1 XM_017018738.1 XM_017018739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1194461 c.5286G>T p.E1762D Chr.12: 119701851-119701851 on Build GRCh38
691827 c.5235C>T p.L1745L Chr.12: 119701902-119701902 on Build GRCh38
5567547 c.5205C>T p.S1735S Chr.12: 119701932-119701932 on Build GRCh38
5552943 c.5306A>T p.E1769V Chr.12: 119701957-119701957 on Build GRCh38
6290046 c.5364C>T p.I1788I Chr.12: 119701676-119701676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752318456 Chr.12: 119701625-119701625 on Build GRCh38
rs766223378 Chr.12: 119701695-119701695 on Build GRCh38
rs763623625 Chr.12: 119701731-119701731 on Build GRCh38
rs35154815 Chr.12: 119701947-119701947 on Build GRCh38
rs766148400 Chr.12: 119701943-119701943 on Build GRCh38
rs751211586 Chr.12: 119701613-119701613 on Build GRCh38
rs561006897 Chr.12: 119701704-119701704 on Build GRCh38
rs778880051 Chr.12: 119701902-119701902 on Build GRCh38
rs138161610 Chr.12: 119701628-119701628 on Build GRCh38
rs745758275 Chr.12: 119701917-119701917 on Build GRCh38
rs372881627 Chr.12: 119701600-119701600 on Build GRCh38
rs920309812 Chr.12: 119701612-119701612 on Build GRCh38
rs375663799 Chr.12: 119701824-119701824 on Build GRCh38
rs763536384 Chr.12: 119701815-119701815 on Build GRCh38
rs201590211 Chr.12: 119701881-119701881 on Build GRCh38
rs374145571 Chr.12: 119701806-119701806 on Build GRCh38
rs756370485 Chr.12: 119701786-119701786 on Build GRCh38
rs768569538 Chr.12: 119701604-119701604 on Build GRCh38
rs778190229 Chr.12: 119701891-119701891 on Build GRCh38
rs758918547 Chr.12: 119701760-119701760 on Build GRCh38
rs543047397 Chr.12: 119701726-119701726 on Build GRCh38
rs371054789 Chr.12: 119701805-119701805 on Build GRCh38
rs1020741303 Chr.12: 119701567-119701567 on Build GRCh38
rs769593950 Chr.12: 119701687-119701687 on Build GRCh38
rs906116174 Chr.12: 119701801-119701801 on Build GRCh38
rs763011355 Chr.12: 119701689-119701689 on Build GRCh38
rs114315762 Chr.12: 119701828-119701828 on Build GRCh38
rs771848800 Chr.12: 119701857-119701857 on Build GRCh38
rs574700924 Chr.12: 119701705-119701705 on Build GRCh38
rs759976682 Chr.12: 119701880-119701880 on Build GRCh38
rs776729880 Chr.12: 119701616-119701616 on Build GRCh38
rs772605323 Chr.12: 119701994-119701994 on Build GRCh38
rs769872098 Chr.12: 119701786-119701786 on Build GRCh38
rs747826680 Chr.12: 119701987-119701987 on Build GRCh38
rs200570302 Chr.12: 119702003-119702003 on Build GRCh38
rs768271650 Chr.12: 119701978-119701978 on Build GRCh38
rs550218744 Chr.12: 119701886-119701886 on Build GRCh38
rs947930495 Chr.12: 119701720-119701720 on Build GRCh38
rs149158991 Chr.12: 119701660-119701660 on Build GRCh38
rs1014720152 Chr.12: 119701570-119701570 on Build GRCh38
rs377716627 Chr.12: 119701774-119701774 on Build GRCh38
rs369065687 Chr.12: 119701617-119701617 on Build GRCh38
rs527864200 Chr.12: 119701870-119701870 on Build GRCh38
rs769238852 Chr.12: 119701991-119701991 on Build GRCh38
rs567861274 Chr.12: 119701878-119701878 on Build GRCh38
rs776101339 Chr.12: 119701871-119701871 on Build GRCh38
rs781435644 Chr.12: 119701829-119701829 on Build GRCh38
rs763442325 Chr.12: 119701819-119701819 on Build GRCh38
rs767968421 Chr.12: 119702004-119702004 on Build GRCh38
rs768924501 Chr.12: 119701675-119701675 on Build GRCh38
rs748315119 Chr.12: 119701678-119701678 on Build GRCh38
rs758273739 Chr.12: 119701931-119701931 on Build GRCh38
rs771060145 Chr.12: 119701804-119701804 on Build GRCh38
rs559057807 Chr.12: 119702006-119702006 on Build GRCh38
rs750880441 Chr.12: 119701756-119701756 on Build GRCh38
rs753349581 Chr.12: 119701818-119701818 on Build GRCh38
rs773246397 Chr.12: 119701688-119701688 on Build GRCh38
rs199581187 Chr.12: 119701807-119701807 on Build GRCh38
rs780327471 Chr.12: 119701761-119701761 on Build GRCh38
rs761584754 Chr.12: 119701606-119701606 on Build GRCh38
rs776302726 Chr.12: 119701986-119701986 on Build GRCh38
rs746834233 Chr.12: 119701958-119701958 on Build GRCh38
rs760306959 Chr.12: 119701862-119701862 on Build GRCh38
rs746198257 Chr.12: 119701656-119701656 on Build GRCh38
rs775167132 Chr.12: 119701812-119701812 on Build GRCh38
rs765381193 Chr.12: 119701713-119701713 on Build GRCh38
rs752901272 Chr.12: 119701784-119701787 on Build GRCh38
rs950222701 Chr.12: 119701998-119701998 on Build GRCh38
rs34140357 Chr.12: 119701896-119701896 on Build GRCh38
rs987781674 Chr.12: 119701595-119701595 on Build GRCh38
rs920257555 Chr.12: 119701882-119701882 on Build GRCh38
rs749175393 Chr.12: 119701790-119701790 on Build GRCh38
rs749437654 Chr.12: 119701851-119701851 on Build GRCh38
rs968249730 Chr.12: 119701655-119701655 on Build GRCh38
rs781522619 Chr.12: 119701776-119701776 on Build GRCh38
rs770972219 Chr.12: 119701853-119701853 on Build GRCh38
rs765283743 Chr.12: 119701608-119701608 on Build GRCh38
rs545201342 Chr.12: 119701593-119701593 on Build GRCh38
rs777508949 Chr.12: 119701730-119701730 on Build GRCh38
rs567298155 Chr.12: 119701661-119701661 on Build GRCh38
rs368957852 Chr.12: 119701603-119701603 on Build GRCh38
rs777996947 Chr.12: 119701839-119701839 on Build GRCh38
rs757373626 Chr.12: 119701900-119701900 on Build GRCh38
rs562984540 Chr.12: 119701784-119701784 on Build GRCh38
rs758358701 Chr.12: 119701706-119701706 on Build GRCh38
rs746291860 Chr.12: 119701588-119701588 on Build GRCh38
rs766838097 Chr.12: 119701875-119701875 on Build GRCh38
rs776565133 Chr.12: 119701676-119701676 on Build GRCh38
rs1018826933 Chr.12: 119701629-119701629 on Build GRCh38
rs201551867 Chr.12: 119701650-119701650 on Build GRCh38
rs776782740 Chr.12: 119701605-119701605 on Build GRCh38
rs756298431 Chr.12: 119701834-119701834 on Build GRCh38
rs760265067 Chr.12: 119701813-119701813 on Build GRCh38
rs764978291 Chr.12: 119701989-119701990 on Build GRCh38
rs1044990143 Chr.12: 119701767-119701767 on Build GRCh38
rs779143986 Chr.12: 119701576-119701576 on Build GRCh38
rs779034565 Chr.12: 119701854-119701854 on Build GRCh38
rs755140359 Chr.12: 119701775-119701775 on Build GRCh38
rs565055860 Chr.12: 119701874-119701874 on Build GRCh38
rs751845241 Chr.12: 119701826-119701826 on Build GRCh38
rs180716474 Chr.12: 119701858-119701858 on Build GRCh38

View Full Product Details