Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120666803-120667304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP1
Entrez Gene ID
9478
Gene Name
calcium binding protein 1
Gene Aliases
CALBRAIN, HCALB_BR
UniGene
Hs.458482
Gene Chromosome Location
Chr.12: 120640552-120680869 on Build GRCh38
Transcripts
NM_001033677.1 NM_004276.4 NM_031205.3 XM_017020235.1 XM_017020236.1 XM_017020237.1 XM_017020238.1 XM_017020239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1677408 c.662T>G p.F221C Chr.12: 120666878-120666878 on Build GRCh38
1677407 c.1091T>G p.F364C Chr.12: 120666878-120666878 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1040629280 Chr.12: 120667229-120667229 on Build GRCh38
rs879513735 Chr.12: 120667192-120667192 on Build GRCh38
rs1040595236 Chr.12: 120667089-120667089 on Build GRCh38
rs763678776 Chr.12: 120666934-120666934 on Build GRCh38
rs959744854 Chr.12: 120666973-120666973 on Build GRCh38
rs768773042 Chr.12: 120666857-120666857 on Build GRCh38
rs377660519 Chr.12: 120666906-120666906 on Build GRCh38
rs1023472132 Chr.12: 120666842-120666842 on Build GRCh38
rs920630552 Chr.12: 120666939-120666939 on Build GRCh38
rs903345553 Chr.12: 120667095-120667095 on Build GRCh38
rs867354771 Chr.12: 120667203-120667203 on Build GRCh38
rs779082988 Chr.12: 120666909-120666909 on Build GRCh38
rs571034033 Chr.12: 120666991-120666991 on Build GRCh38
rs1037975656 Chr.12: 120667232-120667232 on Build GRCh38
rs761725542 Chr.12: 120666860-120666860 on Build GRCh38
rs780213777 Chr.12: 120666825-120666825 on Build GRCh38
rs897514446 Chr.12: 120666876-120666876 on Build GRCh38
rs1010497504 Chr.12: 120667129-120667129 on Build GRCh38
rs772893852 Chr.12: 120666870-120666870 on Build GRCh38
rs573976556 Chr.12: 120667153-120667153 on Build GRCh38
rs372433018 Chr.12: 120666858-120666858 on Build GRCh38
rs756750504 Chr.12: 120666965-120666965 on Build GRCh38
rs981884869 Chr.12: 120666905-120666905 on Build GRCh38
rs779412918 Chr.12: 120666937-120666937 on Build GRCh38
rs934814383 Chr.12: 120667276-120667276 on Build GRCh38
rs751205092 Chr.12: 120666904-120666904 on Build GRCh38
rs1003400316 Chr.12: 120667145-120667145 on Build GRCh38
rs879010161 Chr.12: 120667054-120667054 on Build GRCh38
rs373836365 Chr.12: 120666872-120666872 on Build GRCh38
rs780547361 Chr.12: 120666982-120666982 on Build GRCh38
rs923363907 Chr.12: 120667115-120667115 on Build GRCh38
rs988071974 Chr.12: 120667107-120667107 on Build GRCh38
rs999723357 Chr.12: 120667096-120667096 on Build GRCh38
rs1023524480 Chr.12: 120667133-120667133 on Build GRCh38
rs927799443 Chr.12: 120666931-120666931 on Build GRCh38
rs933366709 Chr.12: 120667150-120667150 on Build GRCh38
rs968951930 Chr.12: 120667058-120667058 on Build GRCh38
rs760064610 Chr.12: 120667277-120667277 on Build GRCh38
rs910544284 Chr.12: 120667204-120667204 on Build GRCh38
rs1012759308 Chr.12: 120667036-120667036 on Build GRCh38
rs898043739 Chr.12: 120667262-120667262 on Build GRCh38
rs1022845746 Chr.12: 120667042-120667042 on Build GRCh38
rs942179482 Chr.12: 120667220-120667220 on Build GRCh38
rs769755752 Chr.12: 120666865-120666865 on Build GRCh38
rs1064001 Chr.12: 120666980-120666980 on Build GRCh38
rs893473899 Chr.12: 120667110-120667110 on Build GRCh38
rs924778772 Chr.12: 120667267-120667267 on Build GRCh38
rs1050679585 Chr.12: 120666985-120666985 on Build GRCh38
rs961901018 Chr.12: 120667158-120667158 on Build GRCh38
rs1052153749 Chr.12: 120667106-120667106 on Build GRCh38
rs920661179 Chr.12: 120667175-120667175 on Build GRCh38
rs942491424 Chr.12: 120667041-120667041 on Build GRCh38
rs751072710 Chr.12: 120666942-120666942 on Build GRCh38
rs200202796 Chr.12: 120666882-120666882 on Build GRCh38
rs933730644 Chr.12: 120667059-120667059 on Build GRCh38
rs944976182 Chr.12: 120667224-120667224 on Build GRCh38
rs750789111 Chr.12: 120666941-120666941 on Build GRCh38
rs778891331 Chr.12: 120666935-120666935 on Build GRCh38
rs986154739 Chr.12: 120667201-120667201 on Build GRCh38
rs536621032 Chr.12: 120667198-120667198 on Build GRCh38
rs969298245 Chr.12: 120667134-120667134 on Build GRCh38
rs756003406 Chr.12: 120667214-120667214 on Build GRCh38
rs952044489 Chr.12: 120667179-120667179 on Build GRCh38
rs35520598 Chr.12: 120667028-120667029 on Build GRCh38
rs758743736 Chr.12: 120666942-120666942 on Build GRCh38
rs1039538494 Chr.12: 120667046-120667046 on Build GRCh38
rs757721882 Chr.12: 120666907-120666907 on Build GRCh38
rs745858237 Chr.12: 120667083-120667083 on Build GRCh38
rs539696796 Chr.12: 120667200-120667200 on Build GRCh38
rs747115203 Chr.12: 120666850-120666850 on Build GRCh38
rs972561355 Chr.12: 120667168-120667168 on Build GRCh38

View Full Product Details