Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21217516-21218007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECE1
Entrez Gene ID
1889
Gene Name
endothelin converting enzyme 1
Gene Aliases
ECE
UniGene
Hs.195080
Gene Chromosome Location
Chr.1: 21217247-21345541 on Build GRCh38
Transcripts
NM_001113347.1 NM_001113348.1 NM_001113349.1 NM_001397.2 XM_006710398.2 XM_011540872.2 XM_011540873.2 XM_017000511.1

SNP Detail

SNP ID Chromosome Location
rs3026915 Chr.1: 21217730-21217730 on Build GRCh38
rs565115846 Chr.1: 21217855-21217855 on Build GRCh38
rs548973942 Chr.1: 21217904-21217904 on Build GRCh38
rs6657745 Chr.1: 21217581-21217581 on Build GRCh38
rs936848332 Chr.1: 21217857-21217862 on Build GRCh38
rs866785001 Chr.1: 21217864-21217864 on Build GRCh38
rs28368058 Chr.1: 21217954-21217954 on Build GRCh38
rs965053264 Chr.1: 21217877-21217877 on Build GRCh38
rs954716882 Chr.1: 21217615-21217615 on Build GRCh38
rs993591007 Chr.1: 21217783-21217783 on Build GRCh38
rs546925071 Chr.1: 21217845-21217845 on Build GRCh38
rs896973123 Chr.1: 21217928-21217928 on Build GRCh38
rs747982102 Chr.1: 21217757-21217757 on Build GRCh38
rs188103782 Chr.1: 21217574-21217574 on Build GRCh38
rs1054018047 Chr.1: 21217886-21217892 on Build GRCh38
rs1036108989 Chr.1: 21217573-21217573 on Build GRCh38
rs1052220112 Chr.1: 21217690-21217690 on Build GRCh38
rs987935786 Chr.1: 21217946-21217946 on Build GRCh38
rs948337248 Chr.1: 21217903-21217903 on Build GRCh38
rs142368680 Chr.1: 21217776-21217776 on Build GRCh38
rs978620087 Chr.1: 21217833-21217833 on Build GRCh38
rs993581697 Chr.1: 21217595-21217595 on Build GRCh38
rs1044512079 Chr.1: 21217691-21217691 on Build GRCh38
rs879165738 Chr.1: 21217943-21217943 on Build GRCh38
rs1026349063 Chr.1: 21217607-21217607 on Build GRCh38
rs1008867347 Chr.1: 21217842-21217842 on Build GRCh38
rs536756133 Chr.1: 21217926-21217926 on Build GRCh38
rs79702808 Chr.1: 21217921-21217921 on Build GRCh38
rs376100722 Chr.1: 21217789-21217789 on Build GRCh38
rs925164688 Chr.1: 21217662-21217662 on Build GRCh38
rs752961302 Chr.1: 21217731-21217731 on Build GRCh38
rs912488590 Chr.1: 21217647-21217647 on Build GRCh38
rs906961628 Chr.1: 21217696-21217696 on Build GRCh38
rs1046692042 Chr.1: 21217548-21217548 on Build GRCh38
rs879145663 Chr.1: 21217958-21217958 on Build GRCh38
rs535645742 Chr.1: 21217986-21217986 on Build GRCh38
rs1002620468 Chr.1: 21217919-21217919 on Build GRCh38
rs905418631 Chr.1: 21217561-21217561 on Build GRCh38
rs74343247 Chr.1: 21217754-21217754 on Build GRCh38
rs1002688143 Chr.1: 21217695-21217695 on Build GRCh38
rs963966611 Chr.1: 21217648-21217648 on Build GRCh38
rs541343019 Chr.1: 21217577-21217577 on Build GRCh38
rs542210812 Chr.1: 21217764-21217764 on Build GRCh38
rs14171 Chr.1: 21217684-21217684 on Build GRCh38
rs925186004 Chr.1: 21217894-21217894 on Build GRCh38
rs181972813 Chr.1: 21217683-21217683 on Build GRCh38
rs965493219 Chr.1: 21217826-21217826 on Build GRCh38
rs936489055 Chr.1: 21217676-21217676 on Build GRCh38
rs148767616 Chr.1: 21217630-21217630 on Build GRCh38
rs912603138 Chr.1: 21217797-21217797 on Build GRCh38
rs532615634 Chr.1: 21217890-21217890 on Build GRCh38
rs1026827767 Chr.1: 21217976-21217976 on Build GRCh38
rs916847355 Chr.1: 21217896-21217896 on Build GRCh38
rs756035609 Chr.1: 21217796-21217796 on Build GRCh38
rs975722752 Chr.1: 21217659-21217659 on Build GRCh38
rs906904130 Chr.1: 21217915-21217915 on Build GRCh38
rs369254256 Chr.1: 21217661-21217661 on Build GRCh38
rs750495682 Chr.1: 21217675-21217675 on Build GRCh38
rs904842038 Chr.1: 21217748-21217748 on Build GRCh38
rs944175158 Chr.1: 21217557-21217557 on Build GRCh38
rs548262816 Chr.1: 21217935-21217935 on Build GRCh38
rs111964570 Chr.1: 21217818-21217818 on Build GRCh38

View Full Product Details