Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41183338-41183867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD1
Entrez Gene ID
161829
Gene Name
exonuclease 3'-5' domain containing 1
Gene Aliases
EXDL1
UniGene
Hs.307999
Gene Chromosome Location
Chr.15: 41182728-41231074 on Build GRCh38
Transcripts
NM_001286441.1 NM_152596.3 XM_006720415.3 XM_011521298.2 XM_011521299.2 XM_011521300.2 XM_011521301.2 XM_011521302.2 XM_017021970.1 XM_017021971.1

SNP Detail

SNP ID Chromosome Location
rs544512068 Chr.15: 41183844-41183844 on Build GRCh38
rs553889292 Chr.15: 41183746-41183746 on Build GRCh38
rs114644953 Chr.15: 41183758-41183758 on Build GRCh38
rs554884967 Chr.15: 41183523-41183523 on Build GRCh38
rs546007212 Chr.15: 41183788-41183788 on Build GRCh38
rs895099663 Chr.15: 41183576-41183576 on Build GRCh38
rs977842947 Chr.15: 41183657-41183657 on Build GRCh38
rs999647771 Chr.15: 41183541-41183541 on Build GRCh38
rs987052221 Chr.15: 41183458-41183458 on Build GRCh38
rs911503164 Chr.15: 41183460-41183460 on Build GRCh38
rs558619725 Chr.15: 41183496-41183496 on Build GRCh38
rs970132754 Chr.15: 41183373-41183373 on Build GRCh38
rs1003311446 Chr.15: 41183533-41183533 on Build GRCh38
rs907179027 Chr.15: 41183532-41183532 on Build GRCh38
rs969723311 Chr.15: 41183646-41183646 on Build GRCh38
rs1031189767 Chr.15: 41183689-41183689 on Build GRCh38
rs898670995 Chr.15: 41183470-41183470 on Build GRCh38
rs565225041 Chr.15: 41183761-41183761 on Build GRCh38
rs914210441 Chr.15: 41183365-41183365 on Build GRCh38
rs931277896 Chr.15: 41183461-41183461 on Build GRCh38
rs1012086361 Chr.15: 41183590-41183590 on Build GRCh38
rs796993731 Chr.15: 41183497-41183497 on Build GRCh38
rs375314138 Chr.15: 41183444-41183444 on Build GRCh38
rs933667987 Chr.15: 41183457-41183457 on Build GRCh38
rs1011119614 Chr.15: 41183609-41183609 on Build GRCh38
rs113643823 Chr.15: 41183460-41183461 on Build GRCh38
rs974304841 Chr.15: 41183774-41183774 on Build GRCh38
rs978252198 Chr.15: 41183433-41183433 on Build GRCh38
rs955054595 Chr.15: 41183734-41183734 on Build GRCh38
rs569138799 Chr.15: 41183466-41183466 on Build GRCh38
rs986905354 Chr.15: 41183749-41183749 on Build GRCh38
rs1024326381 Chr.15: 41183614-41183614 on Build GRCh38
rs959176991 Chr.15: 41183389-41183389 on Build GRCh38
rs113079417 Chr.15: 41183521-41183521 on Build GRCh38
rs1048796235 Chr.15: 41183467-41183470 on Build GRCh38
rs568112144 Chr.15: 41183538-41183538 on Build GRCh38
rs779615327 Chr.15: 41183612-41183612 on Build GRCh38
rs968298087 Chr.15: 41183652-41183652 on Build GRCh38
rs377306350 Chr.15: 41183571-41183571 on Build GRCh38
rs534151735 Chr.15: 41183666-41183666 on Build GRCh38
rs1022047253 Chr.15: 41183426-41183426 on Build GRCh38
rs1003433397 Chr.15: 41183378-41183378 on Build GRCh38
rs552711950 Chr.15: 41183376-41183376 on Build GRCh38
rs757574735 Chr.15: 41183435-41183435 on Build GRCh38
rs1022101338 Chr.15: 41183635-41183635 on Build GRCh38
rs537704255 Chr.15: 41183665-41183665 on Build GRCh38
rs575550820 Chr.15: 41183819-41183819 on Build GRCh38
rs938481772 Chr.15: 41183445-41183445 on Build GRCh38
rs990683415 Chr.15: 41183410-41183410 on Build GRCh38
rs1034953766 Chr.15: 41183385-41183385 on Build GRCh38
rs958458304 Chr.15: 41183588-41183588 on Build GRCh38
rs558769342 Chr.15: 41183807-41183807 on Build GRCh38
rs920066759 Chr.15: 41183777-41183777 on Build GRCh38
rs942862839 Chr.15: 41183465-41183465 on Build GRCh38
rs967830413 Chr.15: 41183428-41183428 on Build GRCh38
rs754180527 Chr.15: 41183411-41183411 on Build GRCh38

View Full Product Details