Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78601809-78602315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA3
Entrez Gene ID
1136
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
Gene Aliases
LNCR2, NACHRA3, PAOD2
UniGene
Hs.89605
Gene Chromosome Location
Chr.15: 78593052-78621295 on Build GRCh38
Transcripts
NM_000743.4 NM_001166694.1 NR_046313.1 XM_006720382.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555121 c.687C>A p.P229P Chr.15: 78601955-78601955 on Build GRCh38
5542745 c.578G>A p.G193D Chr.15: 78602064-78602064 on Build GRCh38
965402 c.503T>C p.F168S Chr.15: 78602139-78602139 on Build GRCh38
458951 c.713G>A p.R238Q Chr.15: 78601929-78601929 on Build GRCh38
965398 c.828C>A p.C276* Chr.15: 78601814-78601814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747512200 Chr.15: 78602240-78602240 on Build GRCh38
rs8192481 Chr.15: 78602066-78602066 on Build GRCh38
rs373218818 Chr.15: 78602082-78602082 on Build GRCh38
rs144257258 Chr.15: 78601955-78601955 on Build GRCh38
rs773346977 Chr.15: 78602290-78602290 on Build GRCh38
rs201993014 Chr.15: 78602090-78602090 on Build GRCh38
rs201203313 Chr.15: 78602177-78602177 on Build GRCh38
rs201693091 Chr.15: 78601898-78601898 on Build GRCh38
rs8192480 Chr.15: 78602078-78602078 on Build GRCh38
rs780434499 Chr.15: 78602109-78602109 on Build GRCh38
rs200161807 Chr.15: 78602155-78602155 on Build GRCh38
rs751388943 Chr.15: 78602029-78602029 on Build GRCh38
rs200129241 Chr.15: 78601885-78601885 on Build GRCh38
rs200244272 Chr.15: 78602014-78602014 on Build GRCh38
rs202207061 Chr.15: 78602183-78602183 on Build GRCh38
rs56371320 Chr.15: 78601941-78601941 on Build GRCh38
rs1012439879 Chr.15: 78602027-78602027 on Build GRCh38
rs201420772 Chr.15: 78602272-78602272 on Build GRCh38
rs112696857 Chr.15: 78602187-78602187 on Build GRCh38
rs200437117 Chr.15: 78602268-78602268 on Build GRCh38
rs759225098 Chr.15: 78602080-78602080 on Build GRCh38
rs934273238 Chr.15: 78602095-78602095 on Build GRCh38
rs199667131 Chr.15: 78601869-78601869 on Build GRCh38
rs75253420 Chr.15: 78602007-78602007 on Build GRCh38
rs529535693 Chr.15: 78601893-78601893 on Build GRCh38
rs79106783 Chr.15: 78601940-78601940 on Build GRCh38
rs778841169 Chr.15: 78602125-78602125 on Build GRCh38
rs757145934 Chr.15: 78602030-78602030 on Build GRCh38
rs756947841 Chr.15: 78601957-78601957 on Build GRCh38
rs201771623 Chr.15: 78602159-78602159 on Build GRCh38
rs201559369 Chr.15: 78602199-78602199 on Build GRCh38
rs61737495 Chr.15: 78601927-78601927 on Build GRCh38
rs752749283 Chr.15: 78602227-78602227 on Build GRCh38
rs763640173 Chr.15: 78601940-78601953 on Build GRCh38
rs976553743 Chr.15: 78601865-78601865 on Build GRCh38
rs143083384 Chr.15: 78602171-78602171 on Build GRCh38
rs202208442 Chr.15: 78601946-78601946 on Build GRCh38
rs201927629 Chr.15: 78601837-78601837 on Build GRCh38
rs201986182 Chr.15: 78602259-78602259 on Build GRCh38
rs761349369 Chr.15: 78602180-78602180 on Build GRCh38
rs759677807 Chr.15: 78601879-78601879 on Build GRCh38
rs766076436 Chr.15: 78601926-78601926 on Build GRCh38
rs577413653 Chr.15: 78602141-78602141 on Build GRCh38
rs750220404 Chr.15: 78602099-78602099 on Build GRCh38
rs1051730 Chr.15: 78601997-78601997 on Build GRCh38
rs781514820 Chr.15: 78602262-78602262 on Build GRCh38
rs200732759 Chr.15: 78601887-78601887 on Build GRCh38
rs771151569 Chr.15: 78601841-78601841 on Build GRCh38
rs555968931 Chr.15: 78602124-78602124 on Build GRCh38
rs79305070 Chr.15: 78602267-78602267 on Build GRCh38
rs147586937 Chr.15: 78602096-78602096 on Build GRCh38
rs201648536 Chr.15: 78602119-78602119 on Build GRCh38
rs984595773 Chr.15: 78602166-78602166 on Build GRCh38
rs200528785 Chr.15: 78601840-78601840 on Build GRCh38
rs74315118 Chr.15: 78602078-78602097 on Build GRCh38
rs200795919 Chr.15: 78602088-78602088 on Build GRCh38
rs539983389 Chr.15: 78601970-78601970 on Build GRCh38
rs768795101 Chr.15: 78602065-78602065 on Build GRCh38
rs774009196 Chr.15: 78602270-78602270 on Build GRCh38
rs199955606 Chr.15: 78601948-78601948 on Build GRCh38
rs72650603 Chr.15: 78601993-78601993 on Build GRCh38
rs79592402 Chr.15: 78601880-78601880 on Build GRCh38
rs771994804 Chr.15: 78602280-78602280 on Build GRCh38
rs575401292 Chr.15: 78602093-78602093 on Build GRCh38
rs368207734 Chr.15: 78601834-78601834 on Build GRCh38
rs767389238 Chr.15: 78602098-78602098 on Build GRCh38
rs570003848 Chr.15: 78602196-78602196 on Build GRCh38
rs771344616 Chr.15: 78602243-78602243 on Build GRCh38
rs764876556 Chr.15: 78602274-78602275 on Build GRCh38
rs775689954 Chr.15: 78601989-78601989 on Build GRCh38
rs199836835 Chr.15: 78602039-78602039 on Build GRCh38
rs200640072 Chr.15: 78602103-78602103 on Build GRCh38
rs200497131 Chr.15: 78601873-78601873 on Build GRCh38
rs542582690 Chr.15: 78602282-78602282 on Build GRCh38
rs761136255 Chr.15: 78601899-78601900 on Build GRCh38
rs551841980 Chr.15: 78601991-78601991 on Build GRCh38
rs71581738 Chr.15: 78601999-78601999 on Build GRCh38
rs760548327 Chr.15: 78602207-78602207 on Build GRCh38
rs759093187 Chr.15: 78602009-78602009 on Build GRCh38
rs748594252 Chr.15: 78602069-78602069 on Build GRCh38
rs55958820 Chr.15: 78602015-78602015 on Build GRCh38
rs144917775 Chr.15: 78602087-78602087 on Build GRCh38
rs201207154 Chr.15: 78601934-78601934 on Build GRCh38
rs762042374 Chr.15: 78601921-78601921 on Build GRCh38
rs764034645 Chr.15: 78602017-78602017 on Build GRCh38
rs750198240 Chr.15: 78602277-78602278 on Build GRCh38
rs761402698 Chr.15: 78602275-78602276 on Build GRCh38
rs746154616 Chr.15: 78602034-78602034 on Build GRCh38
rs772545198 Chr.15: 78602074-78602074 on Build GRCh38
rs780385627 Chr.15: 78602061-78602061 on Build GRCh38
rs78597655 Chr.15: 78601925-78601925 on Build GRCh38
rs201699381 Chr.15: 78601961-78601961 on Build GRCh38
rs139427736 Chr.15: 78601835-78601835 on Build GRCh38
rs372805117 Chr.15: 78601911-78601911 on Build GRCh38
rs766268680 Chr.15: 78602212-78602212 on Build GRCh38
rs748983960 Chr.15: 78602265-78602265 on Build GRCh38
rs76529122 Chr.15: 78602186-78602186 on Build GRCh38
rs758452997 Chr.15: 78602233-78602233 on Build GRCh38
rs144488029 Chr.15: 78602031-78602031 on Build GRCh38
rs753293959 Chr.15: 78601964-78601966 on Build GRCh38
rs755132539 Chr.15: 78602217-78602217 on Build GRCh38
rs71651684 Chr.15: 78602202-78602202 on Build GRCh38
rs72648887 Chr.15: 78602142-78602142 on Build GRCh38
rs748738137 Chr.15: 78602126-78602126 on Build GRCh38
rs200744442 Chr.15: 78602045-78602045 on Build GRCh38
rs980156624 Chr.15: 78602073-78602073 on Build GRCh38
rs76159650 Chr.15: 78601874-78601874 on Build GRCh38
rs200382777 Chr.15: 78601990-78601990 on Build GRCh38
rs76821682 Chr.15: 78601994-78601994 on Build GRCh38
rs199887463 Chr.15: 78601930-78601930 on Build GRCh38
rs754167059 Chr.15: 78602213-78602213 on Build GRCh38
rs149875687 Chr.15: 78602081-78602081 on Build GRCh38
rs778148547 Chr.15: 78602237-78602237 on Build GRCh38
rs762582486 Chr.15: 78602158-78602158 on Build GRCh38
rs149559299 Chr.15: 78601929-78601929 on Build GRCh38
rs754044282 Chr.15: 78602111-78602111 on Build GRCh38
rs772426129 Chr.15: 78602135-78602135 on Build GRCh38
rs200927324 Chr.15: 78602156-78602156 on Build GRCh38
rs777582527 Chr.15: 78601954-78601954 on Build GRCh38
rs146400389 Chr.15: 78601859-78601859 on Build GRCh38
rs183045252 Chr.15: 78602274-78602274 on Build GRCh38
rs764603730 Chr.15: 78601907-78601907 on Build GRCh38
rs770056747 Chr.15: 78601972-78601972 on Build GRCh38

View Full Product Details