Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67538248-67538759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65A
Entrez Gene ID
79567
Gene Name
family with sequence similarity 65 member A
Gene Aliases
-
UniGene
Hs.152717
Gene Chromosome Location
Chr.16: 67528814-67546788 on Build GRCh38
Transcripts
NM_001193522.1 NM_001193523.1 NM_001193524.1 NM_024519.3 XM_011523321.1 XM_011523322.1 XM_011523324.2 XM_011523325.1 XM_011523326.1 XM_017023663.1 XM_017023664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4061875 c.105-1G>A p.? Chr.16: 67538671-67538671 on Build GRCh38
123029 c.65C>G p.S22C Chr.16: 67538511-67538511 on Build GRCh38
6555798 c.15G>T p.S5S Chr.16: 67538461-67538461 on Build GRCh38
5157648 c.145delC p.P50fs*36 Chr.16: 67538700-67538700 on Build GRCh38
7252972 c.20G>T p.R7L Chr.16: 67538466-67538466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765807346 Chr.16: 67538398-67538398 on Build GRCh38
rs920634056 Chr.16: 67538734-67538734 on Build GRCh38
rs774719944 Chr.16: 67538634-67538634 on Build GRCh38
rs775344220 Chr.16: 67538670-67538672 on Build GRCh38
rs778508248 Chr.16: 67538530-67538530 on Build GRCh38
rs542569987 Chr.16: 67538568-67538568 on Build GRCh38
rs779856915 Chr.16: 67538425-67538425 on Build GRCh38
rs906737629 Chr.16: 67538617-67538617 on Build GRCh38
rs760147715 Chr.16: 67538390-67538390 on Build GRCh38
rs199516887 Chr.16: 67538661-67538661 on Build GRCh38
rs751544853 Chr.16: 67538662-67538662 on Build GRCh38
rs953110504 Chr.16: 67538359-67538359 on Build GRCh38
rs200677001 Chr.16: 67538701-67538701 on Build GRCh38
rs548551482 Chr.16: 67538684-67538684 on Build GRCh38
rs776058984 Chr.16: 67538495-67538495 on Build GRCh38
rs763547675 Chr.16: 67538549-67538549 on Build GRCh38
rs372202996 Chr.16: 67538435-67538435 on Build GRCh38
rs745527672 Chr.16: 67538621-67538621 on Build GRCh38
rs767541942 Chr.16: 67538730-67538730 on Build GRCh38
rs773229607 Chr.16: 67538652-67538652 on Build GRCh38
rs759987634 Chr.16: 67538602-67538622 on Build GRCh38
rs528170969 Chr.16: 67538601-67538601 on Build GRCh38
rs949254274 Chr.16: 67538672-67538672 on Build GRCh38
rs958032753 Chr.16: 67538643-67538643 on Build GRCh38
rs780781753 Chr.16: 67538616-67538616 on Build GRCh38
rs888443999 Chr.16: 67538525-67538525 on Build GRCh38
rs544904659 Chr.16: 67538542-67538542 on Build GRCh38
rs775670775 Chr.16: 67538545-67538545 on Build GRCh38
rs376326486 Chr.16: 67538641-67538641 on Build GRCh38
rs757578255 Chr.16: 67538732-67538732 on Build GRCh38
rs113573319 Chr.16: 67538718-67538718 on Build GRCh38
rs1027371496 Chr.16: 67538291-67538291 on Build GRCh38
rs771239489 Chr.16: 67538712-67538712 on Build GRCh38
rs778812178 Chr.16: 67538436-67538436 on Build GRCh38
rs924921124 Chr.16: 67538364-67538364 on Build GRCh38
rs777371446 Chr.16: 67538538-67538538 on Build GRCh38
rs564723641 Chr.16: 67538609-67538609 on Build GRCh38
rs139082757 Chr.16: 67538695-67538695 on Build GRCh38
rs769292995 Chr.16: 67538622-67538622 on Build GRCh38
rs899403366 Chr.16: 67538592-67538592 on Build GRCh38
rs771617887 Chr.16: 67538455-67538455 on Build GRCh38
rs749051541 Chr.16: 67538431-67538431 on Build GRCh38
rs569084656 Chr.16: 67538658-67538658 on Build GRCh38
rs770663713 Chr.16: 67538478-67538478 on Build GRCh38
rs968617667 Chr.16: 67538532-67538532 on Build GRCh38
rs550497737 Chr.16: 67538289-67538289 on Build GRCh38
rs1037922969 Chr.16: 67538397-67538397 on Build GRCh38
rs569075307 Chr.16: 67538306-67538306 on Build GRCh38
rs770755744 Chr.16: 67538541-67538541 on Build GRCh38
rs759736492 Chr.16: 67538723-67538723 on Build GRCh38
rs758489771 Chr.16: 67538408-67538408 on Build GRCh38
rs776153028 Chr.16: 67538712-67538712 on Build GRCh38
rs750011303 Chr.16: 67538735-67538735 on Build GRCh38
rs764258817 Chr.16: 67538499-67538499 on Build GRCh38
rs371323659 Chr.16: 67538477-67538477 on Build GRCh38
rs372941924 Chr.16: 67538726-67538726 on Build GRCh38
rs905238206 Chr.16: 67538273-67538273 on Build GRCh38
rs749732081 Chr.16: 67538697-67538697 on Build GRCh38
rs200947620 Chr.16: 67538554-67538554 on Build GRCh38
rs760394583 Chr.16: 67538476-67538476 on Build GRCh38
rs746466475 Chr.16: 67538539-67538539 on Build GRCh38
rs749772512 Chr.16: 67538404-67538405 on Build GRCh38
rs751348548 Chr.16: 67538728-67538728 on Build GRCh38
rs775964913 Chr.16: 67538625-67538625 on Build GRCh38
rs921980921 Chr.16: 67538341-67538341 on Build GRCh38
rs767704941 Chr.16: 67538516-67538516 on Build GRCh38
rs376811532 Chr.16: 67538555-67538555 on Build GRCh38
rs777171446 Chr.16: 67538488-67538488 on Build GRCh38
rs758397600 Chr.16: 67538587-67538587 on Build GRCh38
rs117467133 Chr.16: 67538631-67538631 on Build GRCh38
rs987711522 Chr.16: 67538696-67538696 on Build GRCh38
rs754767058 Chr.16: 67538528-67538528 on Build GRCh38
rs757374021 Chr.16: 67538413-67538413 on Build GRCh38
rs113729483 Chr.16: 67538330-67538330 on Build GRCh38
rs776712527 Chr.16: 67538716-67538716 on Build GRCh38
rs554667300 Chr.16: 67538347-67538347 on Build GRCh38
rs772525243 Chr.16: 67538706-67538706 on Build GRCh38
rs770285491 Chr.16: 67538543-67538543 on Build GRCh38
rs774539490 Chr.16: 67538561-67538561 on Build GRCh38
rs559190281 Chr.16: 67538395-67538395 on Build GRCh38
rs201157660 Chr.16: 67538691-67538691 on Build GRCh38
rs765902594 Chr.16: 67538493-67538493 on Build GRCh38
rs746575229 Chr.16: 67538600-67538600 on Build GRCh38
rs201528706 Chr.16: 67538692-67538692 on Build GRCh38
rs747683810 Chr.16: 67538447-67538447 on Build GRCh38
rs765737270 Chr.16: 67538460-67538460 on Build GRCh38
rs200327287 Chr.16: 67538588-67538588 on Build GRCh38
rs956150115 Chr.16: 67538278-67538278 on Build GRCh38
rs751554489 Chr.16: 67538500-67538500 on Build GRCh38
rs763365894 Chr.16: 67538577-67538577 on Build GRCh38
rs974725133 Chr.16: 67538689-67538689 on Build GRCh38
rs755576251 Chr.16: 67538738-67538738 on Build GRCh38
rs539969878 Chr.16: 67538603-67538603 on Build GRCh38
rs575053065 Chr.16: 67538563-67538563 on Build GRCh38
rs760718190 Chr.16: 67538711-67538711 on Build GRCh38
rs759852393 Chr.16: 67538376-67538376 on Build GRCh38
rs772536081 Chr.16: 67538470-67538470 on Build GRCh38
rs749923451 Chr.16: 67538690-67538690 on Build GRCh38
rs550384936 Chr.16: 67538654-67538654 on Build GRCh38
rs914468421 Chr.16: 67538531-67538531 on Build GRCh38
rs760915192 Chr.16: 67538653-67538653 on Build GRCh38
rs1032123812 Chr.16: 67538638-67538638 on Build GRCh38
rs762638930 Chr.16: 67538660-67538660 on Build GRCh38
rs759957663 Chr.16: 67538489-67538489 on Build GRCh38
rs771421309 Chr.16: 67538405-67538405 on Build GRCh38
rs570973377 Chr.16: 67538707-67538707 on Build GRCh38
rs949285059 Chr.16: 67538473-67538473 on Build GRCh38
rs761883642 Chr.16: 67538729-67538729 on Build GRCh38
rs756822638 Chr.16: 67538614-67538614 on Build GRCh38
rs752391414 Chr.16: 67538586-67538586 on Build GRCh38
rs768910968 Chr.16: 67538699-67538699 on Build GRCh38
rs767797161 Chr.16: 67538564-67538564 on Build GRCh38
rs965921457 Chr.16: 67538335-67538335 on Build GRCh38
rs763705850 Chr.16: 67538711-67538712 on Build GRCh38
rs772544743 Chr.16: 67538645-67538648 on Build GRCh38
rs752767590 Chr.16: 67538406-67538406 on Build GRCh38
rs534328632 Chr.16: 67538360-67538360 on Build GRCh38
rs760403210 Chr.16: 67538700-67538700 on Build GRCh38
rs566914396 Chr.16: 67538353-67538353 on Build GRCh38
rs140402307 Chr.16: 67538703-67538703 on Build GRCh38
rs560970042 Chr.16: 67538668-67538668 on Build GRCh38
rs866431554 Chr.16: 67538308-67538308 on Build GRCh38
rs1038859701 Chr.16: 67538335-67538336 on Build GRCh38
rs753694604 Chr.16: 67538521-67538521 on Build GRCh38
rs577540820 Chr.16: 67538515-67538515 on Build GRCh38
rs369978963 Chr.16: 67538628-67538628 on Build GRCh38
rs748133124 Chr.16: 67538536-67538536 on Build GRCh38
rs952773537 Chr.16: 67538297-67538297 on Build GRCh38

View Full Product Details