Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71382628-71383126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALB2
Entrez Gene ID
794
Gene Name
calbindin 2
Gene Aliases
CAB29, CAL2, CR
UniGene
Hs.106857
Gene Chromosome Location
Chr.16: 71358713-71390438 on Build GRCh38
Transcripts
NM_001740.4 NM_007088.3 NR_027910.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1379600 c.368G>A p.R123K Chr.16: 71382744-71382744 on Build GRCh38
973273 c.381C>T p.I127I Chr.16: 71382757-71382757 on Build GRCh38
973272 c.358G>A p.D120N Chr.16: 71382734-71382734 on Build GRCh38
7400425 c.347G>A p.W116* Chr.16: 71382723-71382723 on Build GRCh38
6998690 c.350G>A p.R117Q Chr.16: 71382726-71382726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749562670 Chr.16: 71382747-71382747 on Build GRCh38
rs747892011 Chr.16: 71382725-71382725 on Build GRCh38
rs778732378 Chr.16: 71382720-71382720 on Build GRCh38
rs140667050 Chr.16: 71382726-71382726 on Build GRCh38
rs1044796403 Chr.16: 71382826-71382826 on Build GRCh38
rs757188848 Chr.16: 71382819-71382819 on Build GRCh38
rs776468322 Chr.16: 71382660-71382660 on Build GRCh38
rs758135773 Chr.16: 71382813-71382813 on Build GRCh38
rs759662297 Chr.16: 71383041-71383041 on Build GRCh38
rs946851093 Chr.16: 71383015-71383015 on Build GRCh38
rs905546238 Chr.16: 71382806-71382806 on Build GRCh38
rs550300231 Chr.16: 71382907-71382907 on Build GRCh38
rs370740060 Chr.16: 71382687-71382687 on Build GRCh38
rs71709684 Chr.16: 71382967-71382967 on Build GRCh38
rs751517775 Chr.16: 71382815-71382815 on Build GRCh38
rs189281944 Chr.16: 71382983-71382983 on Build GRCh38
rs935569116 Chr.16: 71382750-71382750 on Build GRCh38
rs766234364 Chr.16: 71382680-71382681 on Build GRCh38
rs377617012 Chr.16: 71382812-71382812 on Build GRCh38
rs1056857543 Chr.16: 71383080-71383080 on Build GRCh38
rs142590569 Chr.16: 71382934-71382934 on Build GRCh38
rs765895649 Chr.16: 71382703-71382703 on Build GRCh38
rs755517347 Chr.16: 71382701-71382701 on Build GRCh38
rs983312083 Chr.16: 71382661-71382661 on Build GRCh38
rs564593187 Chr.16: 71382891-71382891 on Build GRCh38
rs777700059 Chr.16: 71383050-71383050 on Build GRCh38
rs80320734 Chr.16: 71382817-71382817 on Build GRCh38
rs762955797 Chr.16: 71382890-71382890 on Build GRCh38
rs926481301 Chr.16: 71383091-71383091 on Build GRCh38
rs762431344 Chr.16: 71382683-71382683 on Build GRCh38
rs752739435 Chr.16: 71382782-71382782 on Build GRCh38
rs965957744 Chr.16: 71382666-71382666 on Build GRCh38
rs886797883 Chr.16: 71382869-71382869 on Build GRCh38
rs1014798494 Chr.16: 71382878-71382878 on Build GRCh38
rs749339191 Chr.16: 71382827-71382827 on Build GRCh38
rs745974357 Chr.16: 71382733-71382733 on Build GRCh38
rs573954879 Chr.16: 71382757-71382757 on Build GRCh38
rs398100276 Chr.16: 71382969-71382969 on Build GRCh38
rs921962326 Chr.16: 71382697-71382697 on Build GRCh38
rs769823419 Chr.16: 71382734-71382734 on Build GRCh38
rs1004799644 Chr.16: 71382872-71382872 on Build GRCh38
rs751286683 Chr.16: 71382789-71382790 on Build GRCh38
rs911892429 Chr.16: 71382994-71382994 on Build GRCh38
rs147616775 Chr.16: 71383003-71383003 on Build GRCh38
rs576200028 Chr.16: 71382651-71382651 on Build GRCh38
rs749952537 Chr.16: 71382694-71382694 on Build GRCh38
rs542260348 Chr.16: 71382777-71382777 on Build GRCh38
rs768151389 Chr.16: 71382767-71382767 on Build GRCh38
rs895475525 Chr.16: 71383092-71383092 on Build GRCh38
rs777509291 Chr.16: 71382727-71382727 on Build GRCh38
rs997424964 Chr.16: 71382910-71382910 on Build GRCh38
rs759212504 Chr.16: 71382799-71382799 on Build GRCh38
rs745900487 Chr.16: 71382669-71382669 on Build GRCh38
rs201261434 Chr.16: 71382754-71382754 on Build GRCh38
rs764832101 Chr.16: 71382801-71382801 on Build GRCh38
rs777612114 Chr.16: 71382814-71382814 on Build GRCh38
rs572449666 Chr.16: 71382855-71382855 on Build GRCh38
rs150422245 Chr.16: 71382769-71382769 on Build GRCh38
rs777937544 Chr.16: 71382684-71382684 on Build GRCh38
rs772828170 Chr.16: 71382871-71382871 on Build GRCh38
rs924651550 Chr.16: 71382713-71382713 on Build GRCh38
rs867906130 Chr.16: 71382758-71382758 on Build GRCh38
rs34063691 Chr.16: 71382705-71382706 on Build GRCh38
rs565386466 Chr.16: 71382985-71382985 on Build GRCh38
rs981022788 Chr.16: 71383082-71383082 on Build GRCh38
rs935621395 Chr.16: 71383001-71383001 on Build GRCh38
rs376259009 Chr.16: 71382670-71382670 on Build GRCh38
rs755579458 Chr.16: 71382664-71382665 on Build GRCh38
rs904946577 Chr.16: 71382848-71382848 on Build GRCh38
rs753499091 Chr.16: 71382707-71382707 on Build GRCh38
rs987458898 Chr.16: 71382974-71382974 on Build GRCh38
rs569688207 Chr.16: 71383060-71383060 on Build GRCh38
rs551198232 Chr.16: 71383004-71383004 on Build GRCh38
rs761236517 Chr.16: 71382775-71382775 on Build GRCh38
rs1001266046 Chr.16: 71383065-71383065 on Build GRCh38
rs754591244 Chr.16: 71382716-71382716 on Build GRCh38
rs768950133 Chr.16: 71382751-71382751 on Build GRCh38
rs763223572 Chr.16: 71382680-71382680 on Build GRCh38
rs754756463 Chr.16: 71382797-71382798 on Build GRCh38
rs1052605135 Chr.16: 71383002-71383002 on Build GRCh38
rs780048549 Chr.16: 71382824-71382824 on Build GRCh38
rs1043161376 Chr.16: 71382792-71382792 on Build GRCh38
rs1048087477 Chr.16: 71382864-71382864 on Build GRCh38
rs775517731 Chr.16: 71382676-71382676 on Build GRCh38
rs529579588 Chr.16: 71382965-71382965 on Build GRCh38
rs775637563 Chr.16: 71382743-71382743 on Build GRCh38
rs192997572 Chr.16: 71382682-71382682 on Build GRCh38
rs751682954 Chr.16: 71382973-71382973 on Build GRCh38
rs546025663 Chr.16: 71382863-71382863 on Build GRCh38
rs560504358 Chr.16: 71382783-71382783 on Build GRCh38
rs184824013 Chr.16: 71382894-71382894 on Build GRCh38

View Full Product Details