Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35984250-35984746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL14
Entrez Gene ID
6358
Gene Name
C-C motif chemokine ligand 14
Gene Aliases
CC-1, CC-3, CKB1, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, MCIF, NCC-2, NCC2, SCYA14, SCYL2, SY14
UniGene
Hs.272493
Gene Chromosome Location
Chr.17: 35983656-35986728 on Build GRCh38
Transcripts
NM_032962.4 NM_032963.3 NR_027921.3 NR_027922.3

Gene Symbol
CCL15-CCL14
Entrez Gene ID
348249
Gene Name
CCL15-CCL14 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.17: 35983656-36002038 on Build GRCh38
Transcripts
NM_032962.4 NM_032963.3 NR_027921.3 NR_027922.3

SNP Detail

SNP ID Chromosome Location
rs1033246578 Chr.17: 35984450-35984450 on Build GRCh38
rs533731552 Chr.17: 35984687-35984687 on Build GRCh38
rs758247502 Chr.17: 35984484-35984484 on Build GRCh38
rs762190316 Chr.17: 35984438-35984438 on Build GRCh38
rs894293188 Chr.17: 35984700-35984700 on Build GRCh38
rs758615336 Chr.17: 35984305-35984305 on Build GRCh38
rs111856180 Chr.17: 35984587-35984587 on Build GRCh38
rs1003950167 Chr.17: 35984423-35984423 on Build GRCh38
rs377214495 Chr.17: 35984354-35984354 on Build GRCh38
rs1041380821 Chr.17: 35984619-35984619 on Build GRCh38
rs766779146 Chr.17: 35984706-35984706 on Build GRCh38
rs1039968972 Chr.17: 35984602-35984602 on Build GRCh38
rs532266388 Chr.17: 35984364-35984364 on Build GRCh38
rs773777488 Chr.17: 35984561-35984561 on Build GRCh38
rs148605501 Chr.17: 35984397-35984397 on Build GRCh38
rs763317253 Chr.17: 35984411-35984411 on Build GRCh38
rs756962710 Chr.17: 35984504-35984504 on Build GRCh38
rs144903710 Chr.17: 35984395-35984395 on Build GRCh38
rs112801225 Chr.17: 35984337-35984337 on Build GRCh38
rs769035837 Chr.17: 35984556-35984556 on Build GRCh38
rs368560391 Chr.17: 35984473-35984473 on Build GRCh38
rs767642183 Chr.17: 35984585-35984585 on Build GRCh38
rs776345194 Chr.17: 35984651-35984651 on Build GRCh38
rs749222447 Chr.17: 35984274-35984274 on Build GRCh38
rs778927371 Chr.17: 35984376-35984376 on Build GRCh38
rs984248322 Chr.17: 35984562-35984562 on Build GRCh38
rs370183552 Chr.17: 35984307-35984307 on Build GRCh38
rs775132854 Chr.17: 35984298-35984298 on Build GRCh38
rs372808751 Chr.17: 35984396-35984396 on Build GRCh38
rs768680560 Chr.17: 35984312-35984312 on Build GRCh38
rs760207812 Chr.17: 35984332-35984332 on Build GRCh38
rs745481745 Chr.17: 35984517-35984517 on Build GRCh38
rs752792314 Chr.17: 35984301-35984301 on Build GRCh38
rs960427631 Chr.17: 35984398-35984398 on Build GRCh38
rs750620028 Chr.17: 35984446-35984446 on Build GRCh38
rs920605410 Chr.17: 35984343-35984343 on Build GRCh38
rs765132575 Chr.17: 35984600-35984600 on Build GRCh38
rs747972780 Chr.17: 35984383-35984383 on Build GRCh38
rs199677295 Chr.17: 35984419-35984419 on Build GRCh38
rs769854730 Chr.17: 35984449-35984449 on Build GRCh38
rs16971802 Chr.17: 35984351-35984351 on Build GRCh38
rs771429403 Chr.17: 35984326-35984326 on Build GRCh38
rs911919263 Chr.17: 35984498-35984498 on Build GRCh38
rs771308300 Chr.17: 35984341-35984341 on Build GRCh38
rs746518735 Chr.17: 35984495-35984495 on Build GRCh38
rs747684916 Chr.17: 35984321-35984321 on Build GRCh38
rs764281468 Chr.17: 35984418-35984418 on Build GRCh38
rs1000335053 Chr.17: 35984683-35984683 on Build GRCh38
rs572762228 Chr.17: 35984424-35984424 on Build GRCh38
rs77812652 Chr.17: 35984365-35984365 on Build GRCh38
rs780826457 Chr.17: 35984516-35984516 on Build GRCh38
rs772924502 Chr.17: 35984327-35984327 on Build GRCh38
rs751828166 Chr.17: 35984360-35984360 on Build GRCh38
rs753086467 Chr.17: 35984352-35984352 on Build GRCh38
rs376131777 Chr.17: 35984406-35984406 on Build GRCh38
rs372776364 Chr.17: 35984311-35984311 on Build GRCh38
rs749838384 Chr.17: 35984555-35984555 on Build GRCh38
rs201876342 Chr.17: 35984389-35984389 on Build GRCh38
rs775693566 Chr.17: 35984549-35984549 on Build GRCh38
rs760762528 Chr.17: 35984593-35984593 on Build GRCh38
rs778350631 Chr.17: 35984315-35984315 on Build GRCh38
rs1056047257 Chr.17: 35984634-35984634 on Build GRCh38
rs375335603 Chr.17: 35984356-35984356 on Build GRCh38
rs368511619 Chr.17: 35984363-35984363 on Build GRCh38
rs113271725 Chr.17: 35984559-35984559 on Build GRCh38
rs752706416 Chr.17: 35984477-35984477 on Build GRCh38
rs764801757 Chr.17: 35984560-35984560 on Build GRCh38
rs144024677 Chr.17: 35984390-35984390 on Build GRCh38
rs756806176 Chr.17: 35984483-35984484 on Build GRCh38
rs778815719 Chr.17: 35984345-35984345 on Build GRCh38
rs922814238 Chr.17: 35984603-35984603 on Build GRCh38
rs986512912 Chr.17: 35984497-35984497 on Build GRCh38
rs113264703 Chr.17: 35984453-35984453 on Build GRCh38
rs200944789 Chr.17: 35984372-35984372 on Build GRCh38
rs750105376 Chr.17: 35984606-35984608 on Build GRCh38
rs763421716 Chr.17: 35984662-35984662 on Build GRCh38
rs146230281 Chr.17: 35984346-35984346 on Build GRCh38
rs892667038 Chr.17: 35984378-35984378 on Build GRCh38
rs934232213 Chr.17: 35984614-35984614 on Build GRCh38
rs771646249 Chr.17: 35984387-35984387 on Build GRCh38
rs960458894 Chr.17: 35984306-35984306 on Build GRCh38
rs756002151 Chr.17: 35984308-35984308 on Build GRCh38
rs752405532 Chr.17: 35984624-35984624 on Build GRCh38
rs777385639 Chr.17: 35984485-35984485 on Build GRCh38
rs918607281 Chr.17: 35984560-35984560 on Build GRCh38
rs770215339 Chr.17: 35984530-35984530 on Build GRCh38
rs914067946 Chr.17: 35984552-35984552 on Build GRCh38
rs962902663 Chr.17: 35984439-35984439 on Build GRCh38

View Full Product Details