Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64053194-64053699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERN1
Entrez Gene ID
2081
Gene Name
endoplasmic reticulum to nucleus signaling 1
Gene Aliases
IRE1, IRE1P, IRE1a, hIRE1p
UniGene
Hs.133982
Gene Chromosome Location
Chr.17: 64039142-64130819 on Build GRCh38
Transcripts
NM_001433.3 XM_017024347.1 XM_017024348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5668778 c.2026_2053+6del34 p.? Chr.17: 64053266-64053299 on Build GRCh38
6235443 c.2024G>A p.G675D Chr.17: 64053301-64053301 on Build GRCh38
4405398 c.1999A>G p.T667A Chr.17: 64053326-64053326 on Build GRCh38
1161200 c.1969G>A p.D657N Chr.17: 64053356-64053356 on Build GRCh38
3520931 c.2042C>T p.S681F Chr.17: 64053283-64053283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781259623 Chr.17: 64053398-64053398 on Build GRCh38
rs775854687 Chr.17: 64053369-64053369 on Build GRCh38
rs188511906 Chr.17: 64053370-64053370 on Build GRCh38
rs774733130 Chr.17: 64053348-64053348 on Build GRCh38
rs902264538 Chr.17: 64053260-64053260 on Build GRCh38
rs754718350 Chr.17: 64053394-64053394 on Build GRCh38
rs548146614 Chr.17: 64053657-64053657 on Build GRCh38
rs753918612 Chr.17: 64053386-64053386 on Build GRCh38
rs923575184 Chr.17: 64053673-64053673 on Build GRCh38
rs756804970 Chr.17: 64053308-64053311 on Build GRCh38
rs760061001 Chr.17: 64053250-64053250 on Build GRCh38
rs372670758 Chr.17: 64053304-64053304 on Build GRCh38
rs768664089 Chr.17: 64053227-64053227 on Build GRCh38
rs780017080 Chr.17: 64053291-64053291 on Build GRCh38
rs1034645671 Chr.17: 64053489-64053489 on Build GRCh38
rs371123787 Chr.17: 64053365-64053365 on Build GRCh38
rs746759316 Chr.17: 64053296-64053296 on Build GRCh38
rs951121474 Chr.17: 64053584-64053584 on Build GRCh38
rs541222628 Chr.17: 64053427-64053427 on Build GRCh38
rs964013696 Chr.17: 64053641-64053641 on Build GRCh38
rs754009385 Chr.17: 64053273-64053273 on Build GRCh38
rs748589397 Chr.17: 64053484-64053484 on Build GRCh38
rs764093163 Chr.17: 64053379-64053379 on Build GRCh38
rs765524076 Chr.17: 64053392-64053392 on Build GRCh38
rs577472148 Chr.17: 64053216-64053216 on Build GRCh38
rs556887858 Chr.17: 64053239-64053239 on Build GRCh38
rs1015304455 Chr.17: 64053516-64053516 on Build GRCh38
rs201648652 Chr.17: 64053261-64053261 on Build GRCh38
rs139208803 Chr.17: 64053234-64053234 on Build GRCh38
rs911128842 Chr.17: 64053242-64053242 on Build GRCh38
rs763379163 Chr.17: 64053244-64053244 on Build GRCh38
rs771364129 Chr.17: 64053597-64053597 on Build GRCh38
rs196929 Chr.17: 64053577-64053577 on Build GRCh38
rs764440786 Chr.17: 64053272-64053272 on Build GRCh38
rs757494318 Chr.17: 64053279-64053279 on Build GRCh38
rs373493203 Chr.17: 64053389-64053389 on Build GRCh38
rs942292175 Chr.17: 64053315-64053315 on Build GRCh38
rs200136146 Chr.17: 64053332-64053332 on Build GRCh38
rs770525435 Chr.17: 64053342-64053342 on Build GRCh38
rs747754405 Chr.17: 64053324-64053324 on Build GRCh38
rs536005555 Chr.17: 64053311-64053311 on Build GRCh38
rs748955378 Chr.17: 64053341-64053341 on Build GRCh38
rs774643237 Chr.17: 64053249-64053249 on Build GRCh38
rs531627528 Chr.17: 64053649-64053649 on Build GRCh38
rs149950313 Chr.17: 64053446-64053446 on Build GRCh38
rs554046227 Chr.17: 64053215-64053215 on Build GRCh38
rs922151043 Chr.17: 64053661-64053661 on Build GRCh38
rs376161056 Chr.17: 64053248-64053248 on Build GRCh38
rs535941406 Chr.17: 64053608-64053608 on Build GRCh38
rs561780829 Chr.17: 64053349-64053349 on Build GRCh38
rs778635155 Chr.17: 64053312-64053313 on Build GRCh38
rs772726313 Chr.17: 64053243-64053243 on Build GRCh38
rs145940718 Chr.17: 64053635-64053635 on Build GRCh38
rs922127107 Chr.17: 64053340-64053340 on Build GRCh38
rs983786071 Chr.17: 64053626-64053627 on Build GRCh38
rs756132682 Chr.17: 64053271-64053271 on Build GRCh38
rs533138574 Chr.17: 64053533-64053533 on Build GRCh38
rs201095170 Chr.17: 64053303-64053303 on Build GRCh38
rs769535746 Chr.17: 64053241-64053241 on Build GRCh38
rs931234915 Chr.17: 64053238-64053238 on Build GRCh38
rs770219153 Chr.17: 64053517-64053517 on Build GRCh38
rs959912071 Chr.17: 64053520-64053520 on Build GRCh38
rs1027720448 Chr.17: 64053598-64053598 on Build GRCh38
rs35591377 Chr.17: 64053631-64053631 on Build GRCh38
rs376687803 Chr.17: 64053393-64053393 on Build GRCh38
rs904478409 Chr.17: 64053451-64053451 on Build GRCh38
rs752583693 Chr.17: 64053399-64053399 on Build GRCh38
rs1001461817 Chr.17: 64053465-64053465 on Build GRCh38
rs1012353947 Chr.17: 64053334-64053334 on Build GRCh38

View Full Product Details