Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65013046-65013593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA13
Entrez Gene ID
10672
Gene Name
G protein subunit alpha 13
Gene Aliases
G13
UniGene
Hs.515018
Gene Chromosome Location
Chr.17: 65009289-65056802 on Build GRCh38
Transcripts
NM_001282425.1 NM_006572.5

SNP Detail

SNP ID Chromosome Location
rs369259098 Chr.17: 65013321-65013323 on Build GRCh38
rs952251595 Chr.17: 65013096-65013096 on Build GRCh38
rs540566694 Chr.17: 65013165-65013165 on Build GRCh38
rs985044199 Chr.17: 65013151-65013151 on Build GRCh38
rs569929430 Chr.17: 65013148-65013148 on Build GRCh38
rs753506530 Chr.17: 65013483-65013483 on Build GRCh38
rs147489674 Chr.17: 65013561-65013561 on Build GRCh38
rs532417508 Chr.17: 65013194-65013194 on Build GRCh38
rs975803781 Chr.17: 65013512-65013512 on Build GRCh38
rs1056931887 Chr.17: 65013436-65013436 on Build GRCh38
rs369137197 Chr.17: 65013104-65013106 on Build GRCh38
rs1044296112 Chr.17: 65013335-65013335 on Build GRCh38
rs988966674 Chr.17: 65013538-65013538 on Build GRCh38
rs914407078 Chr.17: 65013251-65013251 on Build GRCh38
rs535767638 Chr.17: 65013182-65013182 on Build GRCh38
rs773178034 Chr.17: 65013218-65013218 on Build GRCh38
rs549810307 Chr.17: 65013113-65013113 on Build GRCh38
rs549176863 Chr.17: 65013270-65013270 on Build GRCh38
rs565901599 Chr.17: 65013299-65013299 on Build GRCh38
rs1036090721 Chr.17: 65013340-65013340 on Build GRCh38
rs1010083923 Chr.17: 65013476-65013476 on Build GRCh38
rs934477658 Chr.17: 65013199-65013199 on Build GRCh38
rs914370447 Chr.17: 65013542-65013542 on Build GRCh38
rs552163495 Chr.17: 65013342-65013342 on Build GRCh38
rs374947814 Chr.17: 65013488-65013488 on Build GRCh38
rs994008214 Chr.17: 65013458-65013458 on Build GRCh38
rs756873609 Chr.17: 65013496-65013496 on Build GRCh38
rs1045421633 Chr.17: 65013402-65013402 on Build GRCh38
rs892274103 Chr.17: 65013473-65013473 on Build GRCh38
rs1026445095 Chr.17: 65013478-65013478 on Build GRCh38
rs763904482 Chr.17: 65013425-65013425 on Build GRCh38
rs952914246 Chr.17: 65013281-65013281 on Build GRCh38
rs947255921 Chr.17: 65013543-65013543 on Build GRCh38
rs947264177 Chr.17: 65013263-65013263 on Build GRCh38
rs975828833 Chr.17: 65013180-65013180 on Build GRCh38
rs755846772 Chr.17: 65013248-65013248 on Build GRCh38
rs955791294 Chr.17: 65013537-65013537 on Build GRCh38
rs979986351 Chr.17: 65013557-65013557 on Build GRCh38
rs114591136 Chr.17: 65013361-65013361 on Build GRCh38
rs1017247687 Chr.17: 65013153-65013153 on Build GRCh38
rs988630530 Chr.17: 65013243-65013243 on Build GRCh38
rs865967909 Chr.17: 65013320-65013320 on Build GRCh38
rs78170638 Chr.17: 65013440-65013440 on Build GRCh38
rs778233939 Chr.17: 65013499-65013499 on Build GRCh38
rs887953394 Chr.17: 65013484-65013484 on Build GRCh38
rs1030423110 Chr.17: 65013528-65013528 on Build GRCh38
rs1032468895 Chr.17: 65013125-65013125 on Build GRCh38
rs965788188 Chr.17: 65013493-65013493 on Build GRCh38
rs1020044562 Chr.17: 65013214-65013214 on Build GRCh38
rs905108532 Chr.17: 65013403-65013403 on Build GRCh38

View Full Product Details