Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65851632-65852161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP112
Entrez Gene ID
201134
Gene Name
centrosomal protein 112
Gene Aliases
CCDC46, MACOCO
UniGene
Hs.408676
Gene Chromosome Location
Chr.17: 65635540-66192131 on Build GRCh38
Transcripts
NM_001199165.2 NM_001302891.1 XM_005257119.4 XM_005257125.3 XM_005257126.4 XM_006721740.2 XM_011524461.2 XM_011524462.2 XM_011524463.2 XM_011524464.2 XM_011524465.2 XM_011524466.2 XM_011524467.2 XM_017024302.1 XM_017024303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
144118 c.2271G>T p.E757D Chr.17: 65851927-65851927 on Build GRCh38
983089 c.2251C>A p.L751I Chr.17: 65851947-65851947 on Build GRCh38
6866565 c.2164-1G>A p.? Chr.17: 65852035-65852035 on Build GRCh38
5781203 c.2324C>G p.A775G Chr.17: 65851874-65851874 on Build GRCh38
5781202 c.2324C>G p.A775G Chr.17: 65851874-65851874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755149525 Chr.17: 65851795-65851795 on Build GRCh38
rs367585528 Chr.17: 65851792-65851792 on Build GRCh38
rs113286836 Chr.17: 65852020-65852020 on Build GRCh38
rs138625347 Chr.17: 65851860-65851860 on Build GRCh38
rs561006900 Chr.17: 65851762-65851762 on Build GRCh38
rs776259687 Chr.17: 65851780-65851781 on Build GRCh38
rs368676308 Chr.17: 65852009-65852009 on Build GRCh38
rs747685992 Chr.17: 65851935-65851937 on Build GRCh38
rs766721317 Chr.17: 65851778-65851778 on Build GRCh38
rs866906487 Chr.17: 65851864-65851864 on Build GRCh38
rs753860478 Chr.17: 65851812-65851812 on Build GRCh38
rs377414265 Chr.17: 65851968-65851968 on Build GRCh38
rs373589628 Chr.17: 65851934-65851934 on Build GRCh38
rs751505996 Chr.17: 65852026-65852026 on Build GRCh38
rs201402783 Chr.17: 65851982-65851982 on Build GRCh38
rs763469247 Chr.17: 65851891-65851891 on Build GRCh38
rs767597431 Chr.17: 65851919-65851919 on Build GRCh38
rs759510365 Chr.17: 65851909-65851909 on Build GRCh38
rs757132311 Chr.17: 65851927-65851927 on Build GRCh38
rs754598651 Chr.17: 65852048-65852048 on Build GRCh38
rs374657590 Chr.17: 65852034-65852034 on Build GRCh38
rs137923595 Chr.17: 65851890-65851890 on Build GRCh38
rs745679581 Chr.17: 65851846-65851846 on Build GRCh38
rs765645780 Chr.17: 65851761-65851761 on Build GRCh38
rs531004028 Chr.17: 65852097-65852097 on Build GRCh38
rs985739663 Chr.17: 65852126-65852126 on Build GRCh38
rs769497207 Chr.17: 65852051-65852052 on Build GRCh38
rs952666442 Chr.17: 65852119-65852119 on Build GRCh38
rs772197906 Chr.17: 65852016-65852016 on Build GRCh38
rs911136553 Chr.17: 65852134-65852134 on Build GRCh38
rs760811143 Chr.17: 65852025-65852025 on Build GRCh38
rs749638969 Chr.17: 65852000-65852000 on Build GRCh38
rs372409188 Chr.17: 65851875-65851875 on Build GRCh38
rs1053257455 Chr.17: 65851858-65851858 on Build GRCh38
rs765371162 Chr.17: 65851741-65851741 on Build GRCh38
rs775535059 Chr.17: 65852021-65852021 on Build GRCh38
rs762903928 Chr.17: 65852035-65852035 on Build GRCh38
rs763199697 Chr.17: 65851777-65851777 on Build GRCh38
rs201276594 Chr.17: 65851868-65851868 on Build GRCh38
rs779823432 Chr.17: 65851852-65851852 on Build GRCh38
rs748623448 Chr.17: 65851884-65851884 on Build GRCh38
rs369172278 Chr.17: 65851674-65851674 on Build GRCh38
rs761990802 Chr.17: 65851704-65851704 on Build GRCh38
rs750711306 Chr.17: 65851743-65851743 on Build GRCh38
rs201042942 Chr.17: 65852037-65852037 on Build GRCh38
rs778615857 Chr.17: 65851838-65851838 on Build GRCh38
rs774599354 Chr.17: 65852003-65852003 on Build GRCh38
rs759738653 Chr.17: 65852005-65852005 on Build GRCh38
rs752726681 Chr.17: 65851923-65851923 on Build GRCh38
rs751313319 Chr.17: 65852043-65852043 on Build GRCh38
rs779551655 Chr.17: 65851947-65851947 on Build GRCh38
rs369383445 Chr.17: 65851895-65851895 on Build GRCh38
rs762063202 Chr.17: 65851756-65851756 on Build GRCh38
rs754616289 Chr.17: 65851967-65851967 on Build GRCh38
rs919729894 Chr.17: 65852106-65852106 on Build GRCh38
rs774670507 Chr.17: 65851897-65851897 on Build GRCh38
rs532040833 Chr.17: 65851940-65851940 on Build GRCh38
rs539816538 Chr.17: 65851832-65851832 on Build GRCh38
rs778400898 Chr.17: 65852082-65852082 on Build GRCh38
rs150567970 Chr.17: 65851810-65851810 on Build GRCh38
rs757220269 Chr.17: 65851828-65851828 on Build GRCh38
rs758037715 Chr.17: 65851849-65851849 on Build GRCh38
rs773741553 Chr.17: 65851765-65851765 on Build GRCh38
rs750214916 Chr.17: 65851935-65851935 on Build GRCh38
rs771374539 Chr.17: 65852002-65852002 on Build GRCh38
rs776668982 Chr.17: 65851703-65851703 on Build GRCh38
rs746624928 Chr.17: 65851962-65851962 on Build GRCh38
rs778486600 Chr.17: 65851989-65851989 on Build GRCh38
rs752245892 Chr.17: 65852064-65852064 on Build GRCh38
rs780762036 Chr.17: 65852049-65852049 on Build GRCh38
rs373330595 Chr.17: 65852022-65852022 on Build GRCh38
rs755725443 Chr.17: 65852080-65852080 on Build GRCh38

View Full Product Details