Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69084298-69084825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA6
Entrez Gene ID
23460
Gene Name
ATP binding cassette subfamily A member 6
Gene Aliases
EST155051
UniGene
Hs.709514
Gene Chromosome Location
Chr.17: 69062044-69142188 on Build GRCh38
Transcripts
NM_080284.2 XM_011524574.2 XM_017024404.1 XM_017024405.1 XM_017024406.1 XM_017024407.1 XM_017024408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970262 c.4259A>G p.K1420R Chr.17: 69084433-69084433 on Build GRCh38
3521405 c.4286G>A p.G1429E Chr.17: 69084330-69084330 on Build GRCh38
1710689 c.4279C>T p.L1427F Chr.17: 69084337-69084337 on Build GRCh38
4658117 c.4249A>C p.I1417L Chr.17: 69084443-69084443 on Build GRCh38
3820380 c.4206G>A p.L1402L Chr.17: 69084486-69084486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760068384 Chr.17: 69084393-69084393 on Build GRCh38
rs375435290 Chr.17: 69084474-69084474 on Build GRCh38
rs1045417403 Chr.17: 69084575-69084575 on Build GRCh38
rs937298252 Chr.17: 69084621-69084621 on Build GRCh38
rs770650236 Chr.17: 69084528-69084528 on Build GRCh38
rs149935430 Chr.17: 69084790-69084790 on Build GRCh38
rs902606688 Chr.17: 69084796-69084796 on Build GRCh38
rs1032864392 Chr.17: 69084411-69084411 on Build GRCh38
rs1004163203 Chr.17: 69084394-69084394 on Build GRCh38
rs750668985 Chr.17: 69084413-69084413 on Build GRCh38
rs777103028 Chr.17: 69084405-69084405 on Build GRCh38
rs773629916 Chr.17: 69084526-69084526 on Build GRCh38
rs911156855 Chr.17: 69084584-69084584 on Build GRCh38
rs115212891 Chr.17: 69084424-69084424 on Build GRCh38
rs745588043 Chr.17: 69084450-69084450 on Build GRCh38
rs867863983 Chr.17: 69084441-69084441 on Build GRCh38
rs776027982 Chr.17: 69084419-69084420 on Build GRCh38
rs771791554 Chr.17: 69084459-69084459 on Build GRCh38
rs949773131 Chr.17: 69084566-69084566 on Build GRCh38
rs763185730 Chr.17: 69084534-69084534 on Build GRCh38
rs1054768466 Chr.17: 69084627-69084627 on Build GRCh38
rs113163376 Chr.17: 69084569-69084569 on Build GRCh38
rs112487844 Chr.17: 69084628-69084629 on Build GRCh38
rs895425937 Chr.17: 69084739-69084739 on Build GRCh38
rs751641714 Chr.17: 69084610-69084610 on Build GRCh38
rs34016682 Chr.17: 69084800-69084800 on Build GRCh38
rs371905122 Chr.17: 69084427-69084427 on Build GRCh38
rs567756126 Chr.17: 69084740-69084740 on Build GRCh38
rs1047810174 Chr.17: 69084676-69084677 on Build GRCh38
rs905505815 Chr.17: 69084582-69084582 on Build GRCh38
rs139185299 Chr.17: 69084439-69084439 on Build GRCh38
rs72013736 Chr.17: 69084671-69084671 on Build GRCh38
rs762466556 Chr.17: 69084374-69084374 on Build GRCh38
rs200777076 Chr.17: 69084343-69084343 on Build GRCh38
rs939548559 Chr.17: 69084717-69084717 on Build GRCh38
rs1021951639 Chr.17: 69084775-69084775 on Build GRCh38
rs11657954 Chr.17: 69084629-69084629 on Build GRCh38
rs866525764 Chr.17: 69084766-69084766 on Build GRCh38
rs766646980 Chr.17: 69084542-69084542 on Build GRCh38
rs71144653 Chr.17: 69084684-69084684 on Build GRCh38
rs751708179 Chr.17: 69084546-69084546 on Build GRCh38
rs1010517020 Chr.17: 69084759-69084760 on Build GRCh38
rs145416811 Chr.17: 69084438-69084438 on Build GRCh38
rs371125739 Chr.17: 69084355-69084355 on Build GRCh38
rs879218164 Chr.17: 69084641-69084641 on Build GRCh38
rs374050782 Chr.17: 69084519-69084519 on Build GRCh38
rs943331029 Chr.17: 69084611-69084611 on Build GRCh38
rs759727121 Chr.17: 69084547-69084547 on Build GRCh38
rs147432024 Chr.17: 69084420-69084420 on Build GRCh38
rs771535694 Chr.17: 69084377-69084377 on Build GRCh38
rs920668438 Chr.17: 69084635-69084635 on Build GRCh38
rs929414213 Chr.17: 69084668-69084668 on Build GRCh38
rs572455822 Chr.17: 69084692-69084692 on Build GRCh38
rs765830637 Chr.17: 69084377-69084378 on Build GRCh38
rs765583808 Chr.17: 69084408-69084408 on Build GRCh38
rs774961165 Chr.17: 69084470-69084470 on Build GRCh38
rs774184540 Chr.17: 69084364-69084364 on Build GRCh38
rs941516981 Chr.17: 69084532-69084532 on Build GRCh38
rs745525222 Chr.17: 69084376-69084376 on Build GRCh38
rs764151379 Chr.17: 69084477-69084477 on Build GRCh38
rs770823360 Chr.17: 69084362-69084362 on Build GRCh38
rs754391455 Chr.17: 69084456-69084456 on Build GRCh38
rs966463014 Chr.17: 69084430-69084430 on Build GRCh38
rs918340267 Chr.17: 69084562-69084562 on Build GRCh38
rs767697246 Chr.17: 69084556-69084556 on Build GRCh38
rs768126641 Chr.17: 69084476-69084476 on Build GRCh38
rs869083758 Chr.17: 69084682-69084684 on Build GRCh38
rs750713531 Chr.17: 69084713-69084713 on Build GRCh38
rs778834022 Chr.17: 69084444-69084444 on Build GRCh38
rs544800942 Chr.17: 69084382-69084382 on Build GRCh38
rs775073259 Chr.17: 69084386-69084386 on Build GRCh38
rs1055761348 Chr.17: 69084718-69084718 on Build GRCh38
rs1020089318 Chr.17: 69084732-69084732 on Build GRCh38
rs535257875 Chr.17: 69084782-69084782 on Build GRCh38
rs566912016 Chr.17: 69084589-69084589 on Build GRCh38
rs79289091 Chr.17: 69084683-69084683 on Build GRCh38
rs756081786 Chr.17: 69084349-69084349 on Build GRCh38
rs1013875267 Chr.17: 69084322-69084322 on Build GRCh38
rs910072234 Chr.17: 69084530-69084530 on Build GRCh38
rs764498049 Chr.17: 69084514-69084515 on Build GRCh38
rs761126890 Chr.17: 69084465-69084465 on Build GRCh38
rs201306793 Chr.17: 69084323-69084323 on Build GRCh38
rs9675128 Chr.17: 69084659-69084659 on Build GRCh38
rs749116692 Chr.17: 69084358-69084358 on Build GRCh38
rs777188865 Chr.17: 69084483-69084483 on Build GRCh38
rs755057441 Chr.17: 69084428-69084428 on Build GRCh38

View Full Product Details