Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10137004-10137508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT1
Entrez Gene ID
1786
Gene Name
DNA (cytosine-5-)-methyltransferase 1
Gene Aliases
ADCADN, AIM, CXXC9, DNMT, HSN1E, MCMT, m.HsaI
UniGene
Hs.202672
Gene Chromosome Location
Chr.19: 10133344-10195135 on Build GRCh38
Transcripts
NM_001130823.2 NM_001318730.1 NM_001318731.1 NM_001379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3528092 c.4246-1G>A p.? Chr.19: 10137281-10137281 on Build GRCh38
990435 c.4358G>T p.R1453M Chr.19: 10137168-10137168 on Build GRCh38
6312937 c.4367C>A p.P1456H Chr.19: 10137207-10137207 on Build GRCh38
3148750 c.4395C>T p.G1465G Chr.19: 10137131-10137131 on Build GRCh38
1204294 c.4283T>A p.I1428N Chr.19: 10137243-10137243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796174193 Chr.19: 10137374-10137374 on Build GRCh38
rs200416276 Chr.19: 10137182-10137182 on Build GRCh38
rs754351497 Chr.19: 10137143-10137143 on Build GRCh38
rs770535972 Chr.19: 10137129-10137129 on Build GRCh38
rs935622922 Chr.19: 10137442-10137442 on Build GRCh38
rs904011369 Chr.19: 10137402-10137402 on Build GRCh38
rs781301028 Chr.19: 10137091-10137091 on Build GRCh38
rs536916632 Chr.19: 10137115-10137115 on Build GRCh38
rs762360656 Chr.19: 10137283-10137283 on Build GRCh38
rs35012154 Chr.19: 10137353-10137354 on Build GRCh38
rs866729887 Chr.19: 10137112-10137112 on Build GRCh38
rs938326130 Chr.19: 10137378-10137378 on Build GRCh38
rs1000743307 Chr.19: 10137346-10137346 on Build GRCh38
rs1055183996 Chr.19: 10137058-10137058 on Build GRCh38
rs745789285 Chr.19: 10137108-10137108 on Build GRCh38
rs779878371 Chr.19: 10137059-10137059 on Build GRCh38
rs554185167 Chr.19: 10137060-10137060 on Build GRCh38
rs764531629 Chr.19: 10137135-10137135 on Build GRCh38
rs749134980 Chr.19: 10137242-10137242 on Build GRCh38
rs750653853 Chr.19: 10137041-10137041 on Build GRCh38
rs532634715 Chr.19: 10137411-10137411 on Build GRCh38
rs772660711 Chr.19: 10137111-10137111 on Build GRCh38
rs909222539 Chr.19: 10137294-10137294 on Build GRCh38
rs137949464 Chr.19: 10137310-10137310 on Build GRCh38
rs755314598 Chr.19: 10137313-10137313 on Build GRCh38
rs769347864 Chr.19: 10137096-10137096 on Build GRCh38
rs1031456207 Chr.19: 10137277-10137277 on Build GRCh38
rs774010132 Chr.19: 10137232-10137232 on Build GRCh38
rs117579193 Chr.19: 10137042-10137042 on Build GRCh38
rs1008205469 Chr.19: 10137483-10137483 on Build GRCh38
rs770571074 Chr.19: 10137229-10137229 on Build GRCh38
rs752585204 Chr.19: 10137327-10137327 on Build GRCh38
rs577313883 Chr.19: 10137272-10137272 on Build GRCh38
rs143429853 Chr.19: 10137092-10137092 on Build GRCh38
rs959884336 Chr.19: 10137465-10137465 on Build GRCh38
rs560935745 Chr.19: 10137382-10137382 on Build GRCh38
rs758616927 Chr.19: 10137158-10137158 on Build GRCh38
rs549998179 Chr.19: 10137400-10137400 on Build GRCh38
rs547826400 Chr.19: 10137435-10137435 on Build GRCh38
rs147984942 Chr.19: 10137130-10137130 on Build GRCh38
rs947118403 Chr.19: 10137399-10137399 on Build GRCh38
rs750474625 Chr.19: 10137298-10137298 on Build GRCh38
rs761052410 Chr.19: 10137134-10137134 on Build GRCh38
rs527614200 Chr.19: 10137324-10137324 on Build GRCh38
rs765347113 Chr.19: 10137147-10137147 on Build GRCh38
rs769675693 Chr.19: 10137193-10137193 on Build GRCh38
rs985045812 Chr.19: 10137461-10137461 on Build GRCh38
rs1013461054 Chr.19: 10137074-10137074 on Build GRCh38
rs778937086 Chr.19: 10137030-10137030 on Build GRCh38
rs982716298 Chr.19: 10137367-10137367 on Build GRCh38
rs542862981 Chr.19: 10137336-10137336 on Build GRCh38
rs780170131 Chr.19: 10137159-10137159 on Build GRCh38
rs757066879 Chr.19: 10137296-10137297 on Build GRCh38
rs748089711 Chr.19: 10137192-10137192 on Build GRCh38
rs950828379 Chr.19: 10137443-10137443 on Build GRCh38
rs140884982 Chr.19: 10137481-10137481 on Build GRCh38
rs1006590380 Chr.19: 10137381-10137381 on Build GRCh38
rs184657709 Chr.19: 10137478-10137478 on Build GRCh38
rs142647321 Chr.19: 10137146-10137146 on Build GRCh38
rs555878888 Chr.19: 10137258-10137258 on Build GRCh38
rs754833277 Chr.19: 10137181-10137181 on Build GRCh38
rs192224727 Chr.19: 10137392-10137392 on Build GRCh38
rs2229858 Chr.19: 10137206-10137206 on Build GRCh38
rs755902999 Chr.19: 10137140-10137140 on Build GRCh38
rs1053592193 Chr.19: 10137355-10137355 on Build GRCh38
rs370908833 Chr.19: 10137410-10137410 on Build GRCh38
rs139918621 Chr.19: 10137250-10137250 on Build GRCh38
rs2290683 Chr.19: 10137299-10137299 on Build GRCh38
rs79585069 Chr.19: 10137297-10137297 on Build GRCh38
rs566277834 Chr.19: 10137104-10137104 on Build GRCh38
rs28679320 Chr.19: 10137025-10137025 on Build GRCh38
rs748782322 Chr.19: 10137215-10137215 on Build GRCh38
rs751350395 Chr.19: 10137161-10137161 on Build GRCh38
rs1026801149 Chr.19: 10137331-10137331 on Build GRCh38
rs775110717 Chr.19: 10137462-10137462 on Build GRCh38
rs750520436 Chr.19: 10137154-10137154 on Build GRCh38
rs561220532 Chr.19: 10137409-10137410 on Build GRCh38
rs571798381 Chr.19: 10137408-10137408 on Build GRCh38
rs368411249 Chr.19: 10137329-10137329 on Build GRCh38
rs376184130 Chr.19: 10137328-10137328 on Build GRCh38
rs768672451 Chr.19: 10137088-10137088 on Build GRCh38
rs891130568 Chr.19: 10137482-10137482 on Build GRCh38
rs915636148 Chr.19: 10137393-10137393 on Build GRCh38

View Full Product Details