Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39505395-39505930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLL3
Entrez Gene ID
10683
Gene Name
delta like canonical Notch ligand 3
Gene Aliases
SCDO1
UniGene
Hs.127792
Gene Chromosome Location
Chr.19: 39498917-39508481 on Build GRCh38
Transcripts
NM_016941.3 NM_203486.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6085024 c.1087C>T p.R363C Chr.19: 39505445-39505445 on Build GRCh38
1612235 c.1077G>A p.L359L Chr.19: 39505435-39505435 on Build GRCh38
1734599 c.1088G>A p.R363H Chr.19: 39505446-39505446 on Build GRCh38
6085023 c.1087C>T p.R363C Chr.19: 39505445-39505445 on Build GRCh38
5364925 c.1088G>A p.R363H Chr.19: 39505446-39505446 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1018242774 Chr.19: 39505784-39505784 on Build GRCh38
rs774977551 Chr.19: 39505476-39505476 on Build GRCh38
rs533555534 Chr.19: 39505539-39505539 on Build GRCh38
rs893297355 Chr.19: 39505705-39505705 on Build GRCh38
rs921758065 Chr.19: 39505660-39505660 on Build GRCh38
rs1020953038 Chr.19: 39505721-39505721 on Build GRCh38
rs974129947 Chr.19: 39505812-39505812 on Build GRCh38
rs1026783777 Chr.19: 39505820-39505820 on Build GRCh38
rs142732751 Chr.19: 39505577-39505577 on Build GRCh38
rs371137037 Chr.19: 39505475-39505475 on Build GRCh38
rs189451637 Chr.19: 39505531-39505531 on Build GRCh38
rs766943085 Chr.19: 39505510-39505510 on Build GRCh38
rs755265197 Chr.19: 39505532-39505532 on Build GRCh38
rs566919204 Chr.19: 39505794-39505794 on Build GRCh38
rs990063059 Chr.19: 39505681-39505681 on Build GRCh38
rs758572049 Chr.19: 39505575-39505575 on Build GRCh38
rs976329070 Chr.19: 39505655-39505655 on Build GRCh38
rs951697382 Chr.19: 39505545-39505545 on Build GRCh38
rs181454550 Chr.19: 39505554-39505554 on Build GRCh38
rs931908127 Chr.19: 39505667-39505667 on Build GRCh38
rs747037416 Chr.19: 39505578-39505578 on Build GRCh38
rs752597096 Chr.19: 39505561-39505561 on Build GRCh38
rs373673891 Chr.19: 39505460-39505460 on Build GRCh38
rs748298755 Chr.19: 39505443-39505443 on Build GRCh38
rs996184893 Chr.19: 39505815-39505815 on Build GRCh38
rs117616223 Chr.19: 39505520-39505520 on Build GRCh38
rs951280365 Chr.19: 39505839-39505839 on Build GRCh38
rs773927539 Chr.19: 39505506-39505506 on Build GRCh38
rs749901469 Chr.19: 39505549-39505549 on Build GRCh38
rs763505157 Chr.19: 39505502-39505502 on Build GRCh38
rs745604453 Chr.19: 39505473-39505473 on Build GRCh38
rs891401538 Chr.19: 39505762-39505762 on Build GRCh38
rs1044777472 Chr.19: 39505526-39505526 on Build GRCh38
rs917809701 Chr.19: 39505866-39505866 on Build GRCh38
rs964072992 Chr.19: 39505796-39505796 on Build GRCh38
rs762579188 Chr.19: 39505484-39505484 on Build GRCh38
rs139372003 Chr.19: 39505434-39505434 on Build GRCh38
rs982678475 Chr.19: 39505856-39505856 on Build GRCh38
rs1014623582 Chr.19: 39505587-39505587 on Build GRCh38
rs1027181146 Chr.19: 39505857-39505857 on Build GRCh38
rs562761654 Chr.19: 39505653-39505653 on Build GRCh38
rs1056382666 Chr.19: 39505804-39505804 on Build GRCh38
rs755250115 Chr.19: 39505429-39505429 on Build GRCh38
rs777748147 Chr.19: 39505452-39505452 on Build GRCh38
rs761274470 Chr.19: 39505507-39505507 on Build GRCh38
rs146065522 Chr.19: 39505764-39505764 on Build GRCh38
rs919771434 Chr.19: 39505446-39505446 on Build GRCh38
rs781119337 Chr.19: 39505425-39505425 on Build GRCh38
rs113883332 Chr.19: 39505707-39505707 on Build GRCh38
rs949312031 Chr.19: 39505870-39505870 on Build GRCh38
rs965780070 Chr.19: 39505880-39505880 on Build GRCh38
rs146274789 Chr.19: 39505424-39505424 on Build GRCh38
rs975832844 Chr.19: 39505886-39505888 on Build GRCh38
rs917877372 Chr.19: 39505482-39505482 on Build GRCh38
rs907290833 Chr.19: 39505774-39505774 on Build GRCh38
rs770788839 Chr.19: 39505471-39505471 on Build GRCh38
rs776428035 Chr.19: 39505472-39505472 on Build GRCh38
rs757212237 Chr.19: 39505589-39505589 on Build GRCh38
rs756610034 Chr.19: 39505418-39505418 on Build GRCh38
rs142597040 Chr.19: 39505449-39505449 on Build GRCh38
rs1014508562 Chr.19: 39505877-39505877 on Build GRCh38

View Full Product Details