Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51235354-51235791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD33
Entrez Gene ID
945
Gene Name
CD33 molecule
Gene Aliases
SIGLEC-3, SIGLEC3, p67
UniGene
Hs.83731
Gene Chromosome Location
Chr.19: 51211054-51240019 on Build GRCh38
Transcripts
NM_001082618.1 NM_001177608.1 NM_001772.3 XM_011527531.2 XM_011527532.2 XM_017027508.1 XM_017027509.1 XM_017027510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4648319 c.895A>G p.T299A Chr.19: 51235647-51235647 on Build GRCh38
5390495 c.861G>A p.R287R Chr.19: 51235613-51235613 on Build GRCh38
5390497 c.863A>T p.K288I Chr.19: 51235615-51235615 on Build GRCh38
6425282 c.896C>A p.T299N Chr.19: 51235648-51235648 on Build GRCh38
6465897 c.868G>A p.A290T Chr.19: 51235620-51235620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976972082 Chr.19: 51235626-51235626 on Build GRCh38
rs769327558 Chr.19: 51235636-51235636 on Build GRCh38
rs772741411 Chr.19: 51235741-51235741 on Build GRCh38
rs138721648 Chr.19: 51235634-51235634 on Build GRCh38
rs751618546 Chr.19: 51235683-51235683 on Build GRCh38
rs937857596 Chr.19: 51235732-51235732 on Build GRCh38
rs897764645 Chr.19: 51235477-51235477 on Build GRCh38
rs374978260 Chr.19: 51235675-51235675 on Build GRCh38
rs754792639 Chr.19: 51235693-51235693 on Build GRCh38
rs758437739 Chr.19: 51235620-51235620 on Build GRCh38
rs976989594 Chr.19: 51235416-51235416 on Build GRCh38
rs760819350 Chr.19: 51235583-51235583 on Build GRCh38
rs760083270 Chr.19: 51235563-51235563 on Build GRCh38
rs780507431 Chr.19: 51235736-51235736 on Build GRCh38
rs566640100 Chr.19: 51235535-51235535 on Build GRCh38
rs201364242 Chr.19: 51235650-51235650 on Build GRCh38
rs752348760 Chr.19: 51235704-51235704 on Build GRCh38
rs1012627769 Chr.19: 51235497-51235497 on Build GRCh38
rs570781433 Chr.19: 51235589-51235589 on Build GRCh38
rs746335149 Chr.19: 51235649-51235649 on Build GRCh38
rs548309496 Chr.19: 51235517-51235517 on Build GRCh38
rs35112940 Chr.19: 51235662-51235662 on Build GRCh38
rs894097774 Chr.19: 51235390-51235390 on Build GRCh38
rs564870559 Chr.19: 51235382-51235382 on Build GRCh38
rs754074519 Chr.19: 51235593-51235593 on Build GRCh38
rs273621 Chr.19: 51235678-51235678 on Build GRCh38
rs776889535 Chr.19: 51235664-51235664 on Build GRCh38
rs1015533237 Chr.19: 51235751-51235751 on Build GRCh38
rs777271933 Chr.19: 51235730-51235730 on Build GRCh38
rs765281112 Chr.19: 51235605-51235605 on Build GRCh38
rs902752109 Chr.19: 51235673-51235673 on Build GRCh38
rs558681033 Chr.19: 51235569-51235569 on Build GRCh38
rs111722079 Chr.19: 51235676-51235676 on Build GRCh38
rs750324224 Chr.19: 51235613-51235613 on Build GRCh38
rs1012632619 Chr.19: 51235697-51235697 on Build GRCh38
rs1042538116 Chr.19: 51235399-51235399 on Build GRCh38
rs1046762848 Chr.19: 51235724-51235724 on Build GRCh38
rs868629031 Chr.19: 51235639-51235639 on Build GRCh38
rs984012913 Chr.19: 51235539-51235539 on Build GRCh38
rs908383796 Chr.19: 51235555-51235555 on Build GRCh38
rs532697662 Chr.19: 51235398-51235398 on Build GRCh38
rs377686525 Chr.19: 51235742-51235742 on Build GRCh38
rs993820030 Chr.19: 51235507-51235507 on Build GRCh38
rs776934953 Chr.19: 51235686-51235686 on Build GRCh38
rs761921300 Chr.19: 51235603-51235603 on Build GRCh38
rs2271652 Chr.19: 51235632-51235632 on Build GRCh38
rs961262085 Chr.19: 51235565-51235565 on Build GRCh38
rs1046277001 Chr.19: 51235516-51235516 on Build GRCh38
rs61736475 Chr.19: 51235665-51235665 on Build GRCh38
rs368958943 Chr.19: 51235644-51235644 on Build GRCh38
rs759177587 Chr.19: 51235749-51235749 on Build GRCh38
rs141835096 Chr.19: 51235660-51235660 on Build GRCh38
rs779924950 Chr.19: 51235621-51235621 on Build GRCh38
rs751409712 Chr.19: 51235627-51235627 on Build GRCh38
rs768776683 Chr.19: 51235460-51235460 on Build GRCh38
rs932799799 Chr.19: 51235439-51235439 on Build GRCh38
rs754882490 Chr.19: 51235630-51235630 on Build GRCh38
rs1010989721 Chr.19: 51235745-51235745 on Build GRCh38
rs551217090 Chr.19: 51235420-51235420 on Build GRCh38
rs775774290 Chr.19: 51235651-51235651 on Build GRCh38
rs1051199 Chr.19: 51235640-51235640 on Build GRCh38
rs747563205 Chr.19: 51235738-51235738 on Build GRCh38
rs113714504 Chr.19: 51235575-51235575 on Build GRCh38
rs772372749 Chr.19: 51235566-51235566 on Build GRCh38
rs756817874 Chr.19: 51235731-51235731 on Build GRCh38
rs534001899 Chr.19: 51235538-51235538 on Build GRCh38
rs755755670 Chr.19: 51235716-51235716 on Build GRCh38
rs1010937245 Chr.19: 51235648-51235648 on Build GRCh38
rs142485148 Chr.19: 51235672-51235672 on Build GRCh38
rs141793680 Chr.19: 51235456-51235456 on Build GRCh38
rs188461905 Chr.19: 51235446-51235446 on Build GRCh38
rs767267297 Chr.19: 51235700-51235700 on Build GRCh38

View Full Product Details