Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54173748-54174238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5RL
Entrez Gene ID
606495
Gene Name
cytochrome b5 reductase like
Gene Aliases
-
UniGene
Hs.591426
Gene Chromosome Location
Chr.1: 54172336-54200073 on Build GRCh38
Transcripts
NM_001031672.2

SNP Detail

SNP ID Chromosome Location
rs932897325 Chr.1: 54174201-54174201 on Build GRCh38
rs368469673 Chr.1: 54174106-54174106 on Build GRCh38
rs922810717 Chr.1: 54174181-54174181 on Build GRCh38
rs1035323394 Chr.1: 54174131-54174131 on Build GRCh38
rs993417062 Chr.1: 54173954-54173954 on Build GRCh38
rs181693976 Chr.1: 54173922-54173922 on Build GRCh38
rs983183895 Chr.1: 54173825-54173825 on Build GRCh38
rs538211626 Chr.1: 54173980-54173980 on Build GRCh38
rs922090930 Chr.1: 54174214-54174214 on Build GRCh38
rs983621481 Chr.1: 54174202-54174202 on Build GRCh38
rs957192711 Chr.1: 54173782-54173782 on Build GRCh38
rs922758320 Chr.1: 54173838-54173838 on Build GRCh38
rs974970691 Chr.1: 54174212-54174212 on Build GRCh38
rs56195740 Chr.1: 54174136-54174136 on Build GRCh38
rs924623398 Chr.1: 54173809-54173809 on Build GRCh38
rs897509977 Chr.1: 54173946-54173946 on Build GRCh38
rs189726242 Chr.1: 54174093-54174093 on Build GRCh38
rs956268306 Chr.1: 54174135-54174135 on Build GRCh38
rs17389969 Chr.1: 54173817-54173817 on Build GRCh38
rs1023207735 Chr.1: 54174107-54174107 on Build GRCh38
rs922205944 Chr.1: 54173907-54173907 on Build GRCh38
rs72908043 Chr.1: 54174211-54174211 on Build GRCh38
rs950355365 Chr.1: 54174176-54174176 on Build GRCh38
rs550463204 Chr.1: 54174113-54174113 on Build GRCh38
rs751530222 Chr.1: 54174216-54174216 on Build GRCh38
rs1022764973 Chr.1: 54174022-54174022 on Build GRCh38
rs975295940 Chr.1: 54173888-54173888 on Build GRCh38
rs773972339 Chr.1: 54174065-54174065 on Build GRCh38
rs969213789 Chr.1: 54173790-54173790 on Build GRCh38
rs1053846651 Chr.1: 54173990-54173990 on Build GRCh38
rs544945729 Chr.1: 54173796-54173796 on Build GRCh38
rs567510021 Chr.1: 54174114-54174114 on Build GRCh38
rs888834691 Chr.1: 54173897-54173897 on Build GRCh38
rs892978737 Chr.1: 54174038-54174038 on Build GRCh38
rs185191672 Chr.1: 54173945-54173945 on Build GRCh38
rs879781871 Chr.1: 54174200-54174200 on Build GRCh38
rs969930152 Chr.1: 54173805-54173805 on Build GRCh38
rs776914653 Chr.1: 54173802-54173802 on Build GRCh38
rs1031658323 Chr.1: 54174087-54174087 on Build GRCh38
rs968807267 Chr.1: 54174067-54174067 on Build GRCh38
rs148382396 Chr.1: 54174040-54174040 on Build GRCh38
rs574945605 Chr.1: 54173810-54173810 on Build GRCh38
rs1012667397 Chr.1: 54173988-54173988 on Build GRCh38
rs977029750 Chr.1: 54173829-54173829 on Build GRCh38
rs114787371 Chr.1: 54173957-54173957 on Build GRCh38
rs572631963 Chr.1: 54173928-54173928 on Build GRCh38
rs376206030 Chr.1: 54173797-54173797 on Build GRCh38
rs928297042 Chr.1: 54173814-54173814 on Build GRCh38
rs765449058 Chr.1: 54173826-54173826 on Build GRCh38
rs979277727 Chr.1: 54174083-54174083 on Build GRCh38
rs1022858038 Chr.1: 54173803-54173803 on Build GRCh38
rs933550260 Chr.1: 54173933-54173933 on Build GRCh38
rs143220154 Chr.1: 54173856-54173856 on Build GRCh38
rs767012426 Chr.1: 54173972-54173972 on Build GRCh38
rs1002745172 Chr.1: 54174128-54174128 on Build GRCh38
rs1051803671 Chr.1: 54173934-54173934 on Build GRCh38
rs909381548 Chr.1: 54173853-54173853 on Build GRCh38
rs758475403 Chr.1: 54174207-54174207 on Build GRCh38
rs530397438 Chr.1: 54174057-54174057 on Build GRCh38

View Full Product Details