Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19196174-19196675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLTCL1
Entrez Gene ID
8218
Gene Name
clathrin heavy chain like 1
Gene Aliases
CHC22, CLH22, CLTCL, CLTD
UniGene
Hs.368266
Gene Chromosome Location
Chr.22: 19179473-19291716 on Build GRCh38
Transcripts
NM_001835.3 NM_007098.3 XM_011530401.2 XM_017028953.1 XM_017028954.1 XM_017028955.1 XM_017028956.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
461163 c.4177G>T p.D1393Y Chr.22: 19196280-19196280 on Build GRCh38
4672964 c.4031A>G p.N1344S Chr.22: 19196499-19196499 on Build GRCh38
4645436 c.4120G>A p.A1374T Chr.22: 19196337-19196337 on Build GRCh38
4672963 c.4031A>G p.N1344S Chr.22: 19196499-19196499 on Build GRCh38
1032040 c.3943G>A p.G1315S Chr.22: 19196587-19196587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776070983 Chr.22: 19196413-19196413 on Build GRCh38
rs371322004 Chr.22: 19196451-19196451 on Build GRCh38
rs781795037 Chr.22: 19196243-19196243 on Build GRCh38
rs782682662 Chr.22: 19196381-19196381 on Build GRCh38
rs2066732 Chr.22: 19196484-19196484 on Build GRCh38
rs782508706 Chr.22: 19196438-19196438 on Build GRCh38
rs375139032 Chr.22: 19196380-19196380 on Build GRCh38
rs782105034 Chr.22: 19196348-19196348 on Build GRCh38
rs781958676 Chr.22: 19196219-19196219 on Build GRCh38
rs782512890 Chr.22: 19196253-19196253 on Build GRCh38
rs781887587 Chr.22: 19196450-19196450 on Build GRCh38
rs536151684 Chr.22: 19196506-19196506 on Build GRCh38
rs781886709 Chr.22: 19196631-19196631 on Build GRCh38
rs539977681 Chr.22: 19196521-19196521 on Build GRCh38
rs148493715 Chr.22: 19196216-19196217 on Build GRCh38
rs201613157 Chr.22: 19196544-19196544 on Build GRCh38
rs781966797 Chr.22: 19196343-19196343 on Build GRCh38
rs548132819 Chr.22: 19196302-19196302 on Build GRCh38
rs781918864 Chr.22: 19196427-19196427 on Build GRCh38
rs947512112 Chr.22: 19196418-19196418 on Build GRCh38
rs117703854 Chr.22: 19196422-19196422 on Build GRCh38
rs782567839 Chr.22: 19196452-19196452 on Build GRCh38
rs376919709 Chr.22: 19196268-19196268 on Build GRCh38
rs143358242 Chr.22: 19196437-19196437 on Build GRCh38
rs200525646 Chr.22: 19196615-19196615 on Build GRCh38
rs781805918 Chr.22: 19196653-19196653 on Build GRCh38
rs782224246 Chr.22: 19196402-19196402 on Build GRCh38
rs782097067 Chr.22: 19196221-19196221 on Build GRCh38
rs782394698 Chr.22: 19196468-19196468 on Build GRCh38
rs781882719 Chr.22: 19196225-19196225 on Build GRCh38
rs200600002 Chr.22: 19196635-19196635 on Build GRCh38
rs782206829 Chr.22: 19196590-19196590 on Build GRCh38
rs572567479 Chr.22: 19196511-19196511 on Build GRCh38
rs782042627 Chr.22: 19196332-19196332 on Build GRCh38
rs781901074 Chr.22: 19196573-19196573 on Build GRCh38
rs782407097 Chr.22: 19196614-19196614 on Build GRCh38
rs782718897 Chr.22: 19196335-19196335 on Build GRCh38
rs782516151 Chr.22: 19196563-19196563 on Build GRCh38
rs782103755 Chr.22: 19196616-19196616 on Build GRCh38
rs781993061 Chr.22: 19196415-19196415 on Build GRCh38
rs111922411 Chr.22: 19196382-19196382 on Build GRCh38
rs1061325 Chr.22: 19196584-19196584 on Build GRCh38
rs782302023 Chr.22: 19196383-19196383 on Build GRCh38
rs782763855 Chr.22: 19196223-19196223 on Build GRCh38
rs782211704 Chr.22: 19196281-19196281 on Build GRCh38
rs373225202 Chr.22: 19196624-19196624 on Build GRCh38
rs782793738 Chr.22: 19196442-19196442 on Build GRCh38
rs563773335 Chr.22: 19196397-19196397 on Build GRCh38
rs556579608 Chr.22: 19196499-19196499 on Build GRCh38
rs376981962 Chr.22: 19196592-19196592 on Build GRCh38
rs781951943 Chr.22: 19196501-19196501 on Build GRCh38
rs782245729 Chr.22: 19196376-19196376 on Build GRCh38
rs201311337 Chr.22: 19196373-19196373 on Build GRCh38
rs782553621 Chr.22: 19196363-19196363 on Build GRCh38
rs782362240 Chr.22: 19196283-19196283 on Build GRCh38
rs369429598 Chr.22: 19196293-19196293 on Build GRCh38
rs782318173 Chr.22: 19196216-19196216 on Build GRCh38
rs782098593 Chr.22: 19196503-19196503 on Build GRCh38
rs782104259 Chr.22: 19196300-19196301 on Build GRCh38
rs782614257 Chr.22: 19196375-19196375 on Build GRCh38
rs782315545 Chr.22: 19196400-19196400 on Build GRCh38
rs994611963 Chr.22: 19196508-19196508 on Build GRCh38
rs782408947 Chr.22: 19196599-19196599 on Build GRCh38
rs374643955 Chr.22: 19196436-19196436 on Build GRCh38
rs782137134 Chr.22: 19196424-19196424 on Build GRCh38
rs782295560 Chr.22: 19196259-19196259 on Build GRCh38
rs555682535 Chr.22: 19196510-19196510 on Build GRCh38
rs782157960 Chr.22: 19196230-19196230 on Build GRCh38
rs568202121 Chr.22: 19196352-19196352 on Build GRCh38
rs782684713 Chr.22: 19196475-19196475 on Build GRCh38
rs782058726 Chr.22: 19196334-19196334 on Build GRCh38
rs782699047 Chr.22: 19196371-19196371 on Build GRCh38
rs375087295 Chr.22: 19196296-19196296 on Build GRCh38
rs369672659 Chr.22: 19196553-19196553 on Build GRCh38
rs781848408 Chr.22: 19196247-19196247 on Build GRCh38
rs782037652 Chr.22: 19196612-19196612 on Build GRCh38
rs1633399 Chr.22: 19196276-19196276 on Build GRCh38
rs1015119625 Chr.22: 19196201-19196201 on Build GRCh38
rs369197143 Chr.22: 19196467-19196467 on Build GRCh38
rs782660281 Chr.22: 19196569-19196569 on Build GRCh38
rs782734868 Chr.22: 19196334-19196335 on Build GRCh38
rs377562489 Chr.22: 19196353-19196353 on Build GRCh38
rs372686427 Chr.22: 19196407-19196407 on Build GRCh38
rs782802805 Chr.22: 19196425-19196425 on Build GRCh38
rs372375390 Chr.22: 19196256-19196256 on Build GRCh38
rs74677129 Chr.22: 19196549-19196549 on Build GRCh38
rs782693638 Chr.22: 19196641-19196641 on Build GRCh38
rs376402561 Chr.22: 19196505-19196505 on Build GRCh38
rs782630647 Chr.22: 19196460-19196460 on Build GRCh38
rs782815796 Chr.22: 19196232-19196232 on Build GRCh38
rs374830800 Chr.22: 19196458-19196458 on Build GRCh38
rs368439865 Chr.22: 19196401-19196401 on Build GRCh38
rs35428656 Chr.22: 19196281-19196282 on Build GRCh38
rs200802426 Chr.22: 19196652-19196652 on Build GRCh38
rs782476266 Chr.22: 19196459-19196459 on Build GRCh38
rs782576583 Chr.22: 19196585-19196585 on Build GRCh38
rs781985012 Chr.22: 19196292-19196292 on Build GRCh38

View Full Product Details