Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75599860-75600373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL2
Entrez Gene ID
8999
Gene Name
cyclin dependent kinase like 2
Gene Aliases
KKIAMRE, P56
UniGene
Hs.591698
Gene Chromosome Location
Chr.4: 75576494-75630537 on Build GRCh38
Transcripts
NM_003948.3 XM_005263335.2 XM_006714406.2 XM_017008809.1 XM_017008810.1 XM_017008811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1057552 c.819C>A p.D273E Chr.4: 75600346-75600346 on Build GRCh38
98880 c.817G>A p.D273N Chr.4: 75600348-75600348 on Build GRCh38
2831677 c.849C>T p.H283H Chr.4: 75600316-75600316 on Build GRCh38
5814339 c.864A>G p.Q288Q Chr.4: 75600301-75600301 on Build GRCh38
4561341 c.880G>A p.E294K Chr.4: 75600285-75600285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890734858 Chr.4: 75599947-75599947 on Build GRCh38
rs977288552 Chr.4: 75600057-75600057 on Build GRCh38
rs777120581 Chr.4: 75600329-75600329 on Build GRCh38
rs773438679 Chr.4: 75600341-75600341 on Build GRCh38
rs982545121 Chr.4: 75600215-75600215 on Build GRCh38
rs764884812 Chr.4: 75600338-75600338 on Build GRCh38
rs557325819 Chr.4: 75600021-75600021 on Build GRCh38
rs555236758 Chr.4: 75600155-75600155 on Build GRCh38
rs943061630 Chr.4: 75600134-75600134 on Build GRCh38
rs1017767804 Chr.4: 75600054-75600054 on Build GRCh38
rs990652352 Chr.4: 75600286-75600286 on Build GRCh38
rs540417721 Chr.4: 75600257-75600257 on Build GRCh38
rs761309425 Chr.4: 75600322-75600322 on Build GRCh38
rs752183586 Chr.4: 75600247-75600247 on Build GRCh38
rs577121830 Chr.4: 75600311-75600311 on Build GRCh38
rs865840727 Chr.4: 75600260-75600260 on Build GRCh38
rs142575330 Chr.4: 75600283-75600283 on Build GRCh38
rs959370642 Chr.4: 75600001-75600001 on Build GRCh38
rs576031608 Chr.4: 75600028-75600028 on Build GRCh38
rs79921527 Chr.4: 75600348-75600348 on Build GRCh38
rs757015885 Chr.4: 75600267-75600267 on Build GRCh38
rs773821131 Chr.4: 75600320-75600320 on Build GRCh38
rs1040002687 Chr.4: 75600154-75600154 on Build GRCh38
rs752283195 Chr.4: 75600056-75600056 on Build GRCh38
rs55750224 Chr.4: 75600278-75600278 on Build GRCh38
rs185913812 Chr.4: 75599910-75599910 on Build GRCh38
rs144803383 Chr.4: 75600330-75600330 on Build GRCh38
rs190390856 Chr.4: 75600187-75600187 on Build GRCh38
rs1054347210 Chr.4: 75600071-75600071 on Build GRCh38
rs772307019 Chr.4: 75600318-75600318 on Build GRCh38
rs755722434 Chr.4: 75600067-75600067 on Build GRCh38
rs924466511 Chr.4: 75600066-75600066 on Build GRCh38
rs757857199 Chr.4: 75600252-75600252 on Build GRCh38
rs144805549 Chr.4: 75599894-75599894 on Build GRCh38
rs537859934 Chr.4: 75600018-75600018 on Build GRCh38
rs941057350 Chr.4: 75600316-75600316 on Build GRCh38
rs767003411 Chr.4: 75600111-75600111 on Build GRCh38
rs148959397 Chr.4: 75600349-75600349 on Build GRCh38
rs770375391 Chr.4: 75600249-75600249 on Build GRCh38
rs771405423 Chr.4: 75600327-75600327 on Build GRCh38
rs570395203 Chr.4: 75599979-75599979 on Build GRCh38
rs768992377 Chr.4: 75600295-75600295 on Build GRCh38
rs767401264 Chr.4: 75600038-75600038 on Build GRCh38
rs966755035 Chr.4: 75600211-75600211 on Build GRCh38
rs868545441 Chr.4: 75600202-75600202 on Build GRCh38
rs909721494 Chr.4: 75600296-75600296 on Build GRCh38
rs1036799184 Chr.4: 75600317-75600317 on Build GRCh38
rs536809057 Chr.4: 75600086-75600086 on Build GRCh38
rs1034102779 Chr.4: 75599924-75599924 on Build GRCh38
rs1021345612 Chr.4: 75600173-75600173 on Build GRCh38
rs779010784 Chr.4: 75600297-75600297 on Build GRCh38
rs777640018 Chr.4: 75600253-75600253 on Build GRCh38
rs910224226 Chr.4: 75600103-75600103 on Build GRCh38
rs777303634 Chr.4: 75600158-75600158 on Build GRCh38

View Full Product Details