Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83049486-83049991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS4
Entrez Gene ID
51138
Gene Name
COP9 signalosome subunit 4
Gene Aliases
CSN4
UniGene
Hs.190384
Gene Chromosome Location
Chr.4: 83035086-83075818 on Build GRCh38
Transcripts
NM_001258006.1 NM_016129.2 XM_011532019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3606579 c.314C>T p.S105F Chr.4: 83049888-83049888 on Build GRCh38
1486123 c.410+1G>A p.? Chr.4: 83049985-83049985 on Build GRCh38
260258 c.349G>T p.E117* Chr.4: 83049923-83049923 on Build GRCh38
1753856 c.332C>T p.A111V Chr.4: 83049906-83049906 on Build GRCh38
30531 c.385G>C p.G129R Chr.4: 83049959-83049959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776566721 Chr.4: 83049904-83049904 on Build GRCh38
rs749271635 Chr.4: 83049696-83049696 on Build GRCh38
rs199554765 Chr.4: 83049834-83049834 on Build GRCh38
rs534613489 Chr.4: 83049760-83049760 on Build GRCh38
rs970727128 Chr.4: 83049629-83049629 on Build GRCh38
rs1049121492 Chr.4: 83049751-83049751 on Build GRCh38
rs988799685 Chr.4: 83049571-83049571 on Build GRCh38
rs966357895 Chr.4: 83049539-83049539 on Build GRCh38
rs930501011 Chr.4: 83049749-83049749 on Build GRCh38
rs529766614 Chr.4: 83049611-83049611 on Build GRCh38
rs1009791043 Chr.4: 83049793-83049793 on Build GRCh38
rs143003537 Chr.4: 83049773-83049773 on Build GRCh38
rs530854891 Chr.4: 83049779-83049779 on Build GRCh38
rs766363793 Chr.4: 83049890-83049890 on Build GRCh38
rs968197608 Chr.4: 83049840-83049840 on Build GRCh38
rs1036701120 Chr.4: 83049743-83049743 on Build GRCh38
rs546656356 Chr.4: 83049745-83049745 on Build GRCh38
rs112158482 Chr.4: 83049516-83049516 on Build GRCh38
rs775610157 Chr.4: 83049692-83049692 on Build GRCh38
rs13105527 Chr.4: 83049652-83049652 on Build GRCh38
rs767702565 Chr.4: 83049913-83049913 on Build GRCh38
rs773037270 Chr.4: 83049864-83049864 on Build GRCh38
rs148127219 Chr.4: 83049720-83049720 on Build GRCh38
rs1018905795 Chr.4: 83049772-83049772 on Build GRCh38
rs769832908 Chr.4: 83049574-83049574 on Build GRCh38
rs891594580 Chr.4: 83049937-83049937 on Build GRCh38
rs184918544 Chr.4: 83049712-83049712 on Build GRCh38
rs767453225 Chr.4: 83049931-83049931 on Build GRCh38
rs570622720 Chr.4: 83049759-83049759 on Build GRCh38
rs1003972070 Chr.4: 83049659-83049659 on Build GRCh38
rs1021468051 Chr.4: 83049567-83049567 on Build GRCh38
rs548301879 Chr.4: 83049623-83049623 on Build GRCh38
rs916904823 Chr.4: 83049718-83049718 on Build GRCh38
rs759023199 Chr.4: 83049930-83049930 on Build GRCh38
rs146149758 Chr.4: 83049871-83049871 on Build GRCh38
rs565175316 Chr.4: 83049541-83049541 on Build GRCh38
rs559397459 Chr.4: 83049566-83049566 on Build GRCh38
rs1049364657 Chr.4: 83049778-83049778 on Build GRCh38

View Full Product Details