Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172994538-172995036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0E1
Entrez Gene ID
8992
Gene Name
ATPase H+ transporting V0 subunit e1
Gene Aliases
ATP6H, ATP6V0E, M9.2, Vma21, Vma21p
UniGene
Hs.484188
Gene Chromosome Location
Chr.5: 172983760-173034897 on Build GRCh38
Transcripts
NM_003945.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6504424 c.134C>T p.S45L Chr.5: 172994804-172994804 on Build GRCh38
6504425 c.152+2T>C p.? Chr.5: 172994824-172994824 on Build GRCh38
3827696 c.148C>T p.L50F Chr.5: 172994818-172994818 on Build GRCh38
5947163 c.105A>T p.G35G Chr.5: 172994775-172994775 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs559097222 Chr.5: 172994575-172994575 on Build GRCh38
rs372332112 Chr.5: 172994816-172994816 on Build GRCh38
rs758958080 Chr.5: 172994830-172994831 on Build GRCh38
rs779207427 Chr.5: 172994758-172994759 on Build GRCh38
rs546526484 Chr.5: 172994760-172994761 on Build GRCh38
rs866408340 Chr.5: 172994773-172994773 on Build GRCh38
rs1030523481 Chr.5: 172994823-172994826 on Build GRCh38
rs746297351 Chr.5: 172994781-172994781 on Build GRCh38
rs1037752868 Chr.5: 172994631-172994631 on Build GRCh38
rs1048179019 Chr.5: 172994617-172994617 on Build GRCh38
rs796772002 Chr.5: 172994772-172994772 on Build GRCh38
rs781113695 Chr.5: 172994955-172994955 on Build GRCh38
rs757561313 Chr.5: 172994935-172994935 on Build GRCh38
rs754264892 Chr.5: 172994747-172994747 on Build GRCh38
rs373430932 Chr.5: 172994859-172994859 on Build GRCh38
rs545355668 Chr.5: 172994945-172994945 on Build GRCh38
rs917150523 Chr.5: 172994604-172994604 on Build GRCh38
rs769081978 Chr.5: 172994757-172994758 on Build GRCh38
rs207466625 Chr.5: 172994818-172994818 on Build GRCh38
rs904556713 Chr.5: 172994681-172994681 on Build GRCh38
rs745395149 Chr.5: 172994864-172994865 on Build GRCh38
rs746480890 Chr.5: 172994822-172994822 on Build GRCh38
rs919142680 Chr.5: 172994587-172994587 on Build GRCh38
rs150038275 Chr.5: 172994610-172994610 on Build GRCh38
rs369931007 Chr.5: 172994854-172994854 on Build GRCh38
rs755861161 Chr.5: 172994912-172994912 on Build GRCh38
rs762750129 Chr.5: 172994864-172994864 on Build GRCh38
rs879137806 Chr.5: 172994759-172994759 on Build GRCh38
rs934625362 Chr.5: 172994716-172994716 on Build GRCh38
rs574712782 Chr.5: 172994749-172994749 on Build GRCh38
rs762857671 Chr.5: 172994586-172994586 on Build GRCh38
rs927019669 Chr.5: 172994949-172994949 on Build GRCh38
rs772496917 Chr.5: 172994758-172994761 on Build GRCh38
rs765194999 Chr.5: 172994588-172994588 on Build GRCh38
rs780282910 Chr.5: 172994757-172994757 on Build GRCh38
rs200726685 Chr.5: 172994861-172994861 on Build GRCh38
rs199527939 Chr.5: 172994871-172994871 on Build GRCh38
rs757828997 Chr.5: 172994761-172994761 on Build GRCh38
rs374828372 Chr.5: 172994860-172994860 on Build GRCh38
rs866454996 Chr.5: 172994705-172994705 on Build GRCh38
rs377134642 Chr.5: 172994872-172994872 on Build GRCh38
rs543260071 Chr.5: 172994759-172994760 on Build GRCh38
rs17075485 Chr.5: 172994996-172994996 on Build GRCh38
rs776262965 Chr.5: 172994561-172994561 on Build GRCh38
rs540994131 Chr.5: 172994760-172994760 on Build GRCh38
rs756206815 Chr.5: 172994727-172994727 on Build GRCh38
rs1053299969 Chr.5: 172994721-172994721 on Build GRCh38
rs937163859 Chr.5: 172994584-172994584 on Build GRCh38
rs372851853 Chr.5: 172994635-172994635 on Build GRCh38
rs11296906 Chr.5: 172994758-172994758 on Build GRCh38
rs60940868 Chr.5: 172994620-172994620 on Build GRCh38
rs963040002 Chr.5: 172994577-172994577 on Build GRCh38
rs968377674 Chr.5: 172994919-172994919 on Build GRCh38
rs774415300 Chr.5: 172994775-172994775 on Build GRCh38
rs533025872 Chr.5: 172994671-172994671 on Build GRCh38
rs368834743 Chr.5: 172994870-172994870 on Build GRCh38
rs764620595 Chr.5: 172994744-172994744 on Build GRCh38
rs554784310 Chr.5: 172994621-172994621 on Build GRCh38
rs779879867 Chr.5: 172994787-172994787 on Build GRCh38
rs996746178 Chr.5: 172994691-172994691 on Build GRCh38
rs958520064 Chr.5: 172994994-172994994 on Build GRCh38
rs963048293 Chr.5: 172994948-172994948 on Build GRCh38
rs192962545 Chr.5: 172994991-172994991 on Build GRCh38
rs539745397 Chr.5: 172994758-172994760 on Build GRCh38
rs779129450 Chr.5: 172994765-172994765 on Build GRCh38
rs199914104 Chr.5: 172994839-172994839 on Build GRCh38

View Full Product Details