Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176575197-176575695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR2
Entrez Gene ID
54825
Gene Name
cadherin related family member 2
Gene Aliases
PCDH24, PCLKC
UniGene
Hs.4205
Gene Chromosome Location
Chr.5: 176542511-176595768 on Build GRCh38
Transcripts
NM_001171976.1 NM_017675.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1696778 c.653C>T p.T218I Chr.5: 176575311-176575311 on Build GRCh38
328683 c.635T>A p.M212K Chr.5: 176575293-176575293 on Build GRCh38
4671041 c.637T>C p.Y213H Chr.5: 176575295-176575295 on Build GRCh38
1066045 c.840C>T p.I280I Chr.5: 176575577-176575577 on Build GRCh38
1066044 c.836G>T p.S279I Chr.5: 176575573-176575573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558396996 Chr.5: 176575444-176575444 on Build GRCh38
rs1054352350 Chr.5: 176575613-176575613 on Build GRCh38
rs776904636 Chr.5: 176575476-176575476 on Build GRCh38
rs770930688 Chr.5: 176575460-176575460 on Build GRCh38
rs779629169 Chr.5: 176575282-176575282 on Build GRCh38
rs780470108 Chr.5: 176575381-176575381 on Build GRCh38
rs751985605 Chr.5: 176575435-176575435 on Build GRCh38
rs748050927 Chr.5: 176575295-176575295 on Build GRCh38
rs1033139078 Chr.5: 176575289-176575289 on Build GRCh38
rs144549731 Chr.5: 176575579-176575579 on Build GRCh38
rs777917560 Chr.5: 176575397-176575397 on Build GRCh38
rs748321882 Chr.5: 176575569-176575569 on Build GRCh38
rs775644383 Chr.5: 176575321-176575321 on Build GRCh38
rs371315136 Chr.5: 176575497-176575497 on Build GRCh38
rs41275281 Chr.5: 176575541-176575541 on Build GRCh38
rs763852877 Chr.5: 176575617-176575617 on Build GRCh38
rs565382865 Chr.5: 176575398-176575398 on Build GRCh38
rs948810643 Chr.5: 176575630-176575630 on Build GRCh38
rs766605701 Chr.5: 176575433-176575433 on Build GRCh38
rs752802702 Chr.5: 176575259-176575259 on Build GRCh38
rs755850705 Chr.5: 176575521-176575521 on Build GRCh38
rs776774282 Chr.5: 176575243-176575243 on Build GRCh38
rs953791028 Chr.5: 176575537-176575537 on Build GRCh38
rs111331750 Chr.5: 176575652-176575652 on Build GRCh38
rs754682591 Chr.5: 176575387-176575387 on Build GRCh38
rs545087058 Chr.5: 176575625-176575625 on Build GRCh38
rs763331211 Chr.5: 176575506-176575506 on Build GRCh38
rs1015296928 Chr.5: 176575483-176575483 on Build GRCh38
rs985995103 Chr.5: 176575672-176575672 on Build GRCh38
rs757549117 Chr.5: 176575224-176575224 on Build GRCh38
rs35763816 Chr.5: 176575294-176575294 on Build GRCh38
rs749434791 Chr.5: 176575582-176575582 on Build GRCh38
rs371884538 Chr.5: 176575225-176575225 on Build GRCh38
rs757117381 Chr.5: 176575539-176575539 on Build GRCh38
rs769805729 Chr.5: 176575494-176575494 on Build GRCh38
rs375004151 Chr.5: 176575273-176575273 on Build GRCh38
rs745470073 Chr.5: 176575228-176575228 on Build GRCh38
rs752245779 Chr.5: 176575440-176575440 on Build GRCh38
rs576687748 Chr.5: 176575446-176575446 on Build GRCh38
rs781043740 Chr.5: 176575437-176575437 on Build GRCh38
rs765727050 Chr.5: 176575414-176575414 on Build GRCh38
rs777398718 Chr.5: 176575434-176575435 on Build GRCh38
rs767276440 Chr.5: 176575514-176575514 on Build GRCh38
rs752333727 Chr.5: 176575516-176575516 on Build GRCh38
rs766951572 Chr.5: 176575509-176575509 on Build GRCh38
rs375152272 Chr.5: 176575432-176575432 on Build GRCh38
rs767769498 Chr.5: 176575314-176575314 on Build GRCh38
rs981716435 Chr.5: 176575628-176575628 on Build GRCh38
rs765067737 Chr.5: 176575247-176575247 on Build GRCh38
rs780808780 Chr.5: 176575390-176575390 on Build GRCh38
rs761937446 Chr.5: 176575410-176575410 on Build GRCh38
rs750772140 Chr.5: 176575370-176575370 on Build GRCh38
rs377539876 Chr.5: 176575246-176575246 on Build GRCh38
rs767986869 Chr.5: 176575288-176575288 on Build GRCh38
rs974177342 Chr.5: 176575422-176575422 on Build GRCh38
rs757934024 Chr.5: 176575390-176575390 on Build GRCh38
rs369712069 Chr.5: 176575637-176575637 on Build GRCh38
rs765782132 Chr.5: 176575361-176575361 on Build GRCh38
rs758754545 Chr.5: 176575373-176575373 on Build GRCh38
rs1046344120 Chr.5: 176575632-176575632 on Build GRCh38
rs930449004 Chr.5: 176575664-176575664 on Build GRCh38
rs762708653 Chr.5: 176575310-176575310 on Build GRCh38
rs764236286 Chr.5: 176575268-176575268 on Build GRCh38
rs907468822 Chr.5: 176575465-176575465 on Build GRCh38
rs760298460 Chr.5: 176575655-176575655 on Build GRCh38
rs773115328 Chr.5: 176575306-176575306 on Build GRCh38
rs974121000 Chr.5: 176575643-176575643 on Build GRCh38
rs768624111 Chr.5: 176575236-176575236 on Build GRCh38
rs577130376 Chr.5: 176575462-176575462 on Build GRCh38
rs746522469 Chr.5: 176575286-176575286 on Build GRCh38
rs769836635 Chr.5: 176575395-176575395 on Build GRCh38
rs779836636 Chr.5: 176575554-176575554 on Build GRCh38
rs533394997 Chr.5: 176575590-176575590 on Build GRCh38
rs962364465 Chr.5: 176575491-176575491 on Build GRCh38
rs902880028 Chr.5: 176575467-176575467 on Build GRCh38
rs762025143 Chr.5: 176575482-176575482 on Build GRCh38
rs758121533 Chr.5: 176575548-176575548 on Build GRCh38
rs747704946 Chr.5: 176575394-176575394 on Build GRCh38
rs573333919 Chr.5: 176575636-176575636 on Build GRCh38
rs754647304 Chr.5: 176575436-176575436 on Build GRCh38
rs776025203 Chr.5: 176575594-176575594 on Build GRCh38
rs574174277 Chr.5: 176575571-176575571 on Build GRCh38
rs201292271 Chr.5: 176575250-176575250 on Build GRCh38
rs962476862 Chr.5: 176575232-176575232 on Build GRCh38
rs936007685 Chr.5: 176575612-176575612 on Build GRCh38
rs768475972 Chr.5: 176575523-176575523 on Build GRCh38
rs760039691 Chr.5: 176575513-176575513 on Build GRCh38
rs753270430 Chr.5: 176575674-176575674 on Build GRCh38
rs894526417 Chr.5: 176575424-176575424 on Build GRCh38
rs762647021 Chr.5: 176575449-176575449 on Build GRCh38
rs754282100 Chr.5: 176575272-176575272 on Build GRCh38
rs776669151 Chr.5: 176575409-176575409 on Build GRCh38
rs377337438 Chr.5: 176575275-176575275 on Build GRCh38
rs773164368 Chr.5: 176575249-176575249 on Build GRCh38
rs920411750 Chr.5: 176575428-176575428 on Build GRCh38
rs753946454 Chr.5: 176575348-176575348 on Build GRCh38
rs932479241 Chr.5: 176575260-176575260 on Build GRCh38
rs916179816 Chr.5: 176575320-176575320 on Build GRCh38
rs539729372 Chr.5: 176575400-176575400 on Build GRCh38
rs762273064 Chr.5: 176575349-176575349 on Build GRCh38
rs760963659 Chr.5: 176575329-176575329 on Build GRCh38
rs746271492 Chr.5: 176575276-176575276 on Build GRCh38
rs770570875 Chr.5: 176575399-176575399 on Build GRCh38
rs745658793 Chr.5: 176575542-176575542 on Build GRCh38
rs764108247 Chr.5: 176575341-176575341 on Build GRCh38
rs769555114 Chr.5: 176575366-176575366 on Build GRCh38
rs771649886 Chr.5: 176575229-176575229 on Build GRCh38
rs760370477 Chr.5: 176575616-176575616 on Build GRCh38
rs41275283 Chr.5: 176575442-176575442 on Build GRCh38
rs374219258 Chr.5: 176575522-176575522 on Build GRCh38
rs760127288 Chr.5: 176575593-176575593 on Build GRCh38
rs199964393 Chr.5: 176575382-176575382 on Build GRCh38
rs541587987 Chr.5: 176575575-176575575 on Build GRCh38
rs1040002803 Chr.5: 176575452-176575452 on Build GRCh38
rs981446652 Chr.5: 176575354-176575354 on Build GRCh38
rs769883949 Chr.5: 176575304-176575304 on Build GRCh38
rs558686748 Chr.5: 176575461-176575461 on Build GRCh38
rs1027968095 Chr.5: 176575671-176575671 on Build GRCh38
rs774560711 Chr.5: 176575510-176575510 on Build GRCh38
rs746714639 Chr.5: 176575523-176575525 on Build GRCh38
rs201250874 Chr.5: 176575468-176575468 on Build GRCh38
rs368970243 Chr.5: 176575598-176575598 on Build GRCh38

View Full Product Details