Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 94535215-94535757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASD1
Entrez Gene ID
64921
Gene Name
CAS1 domain containing 1
Gene Aliases
C7orf12, NBLA04196
UniGene
Hs.260041
Gene Chromosome Location
Chr.7: 94509858-94633465 on Build GRCh38
Transcripts
NM_022900.4 XM_006716092.3 XM_006716093.3 XM_011516495.2 XM_017012546.1 XM_017012547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620432 c.798G>A p.M266I Chr.7: 94535478-94535478 on Build GRCh38
5700083 c.665A>C p.K222T Chr.7: 94535345-94535345 on Build GRCh38
4938297 c.826G>C p.E276Q Chr.7: 94535506-94535506 on Build GRCh38
1313435 c.640G>A p.E214K Chr.7: 94535320-94535320 on Build GRCh38
1093277 c.646C>A p.L216I Chr.7: 94535326-94535326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751971394 Chr.7: 94535510-94535510 on Build GRCh38
rs778693082 Chr.7: 94535478-94535478 on Build GRCh38
rs1029932822 Chr.7: 94535541-94535541 on Build GRCh38
rs753255825 Chr.7: 94535466-94535466 on Build GRCh38
rs778666168 Chr.7: 94535279-94535279 on Build GRCh38
rs768459446 Chr.7: 94535535-94535535 on Build GRCh38
rs111970288 Chr.7: 94535597-94535597 on Build GRCh38
rs996420872 Chr.7: 94535657-94535657 on Build GRCh38
rs1004861077 Chr.7: 94535248-94535248 on Build GRCh38
rs756615615 Chr.7: 94535263-94535263 on Build GRCh38
rs1029160639 Chr.7: 94535306-94535306 on Build GRCh38
rs781233390 Chr.7: 94535545-94535545 on Build GRCh38
rs991805104 Chr.7: 94535497-94535497 on Build GRCh38
rs991094131 Chr.7: 94535682-94535682 on Build GRCh38
rs146953436 Chr.7: 94535475-94535475 on Build GRCh38
rs777103468 Chr.7: 94535266-94535266 on Build GRCh38
rs759764626 Chr.7: 94535336-94535336 on Build GRCh38
rs546884916 Chr.7: 94535589-94535589 on Build GRCh38
rs78922839 Chr.7: 94535511-94535511 on Build GRCh38
rs566783380 Chr.7: 94535645-94535645 on Build GRCh38
rs773121846 Chr.7: 94535312-94535312 on Build GRCh38
rs771030601 Chr.7: 94535324-94535324 on Build GRCh38
rs539230958 Chr.7: 94535696-94535697 on Build GRCh38
rs755150089 Chr.7: 94535568-94535568 on Build GRCh38
rs764709737 Chr.7: 94535430-94535430 on Build GRCh38
rs925476060 Chr.7: 94535637-94535637 on Build GRCh38
rs753407472 Chr.7: 94535269-94535269 on Build GRCh38
rs34850251 Chr.7: 94535508-94535508 on Build GRCh38
rs200618939 Chr.7: 94535302-94535302 on Build GRCh38
rs372582422 Chr.7: 94535567-94535567 on Build GRCh38
rs375425016 Chr.7: 94535354-94535354 on Build GRCh38
rs917899429 Chr.7: 94535684-94535684 on Build GRCh38
rs746877572 Chr.7: 94535305-94535305 on Build GRCh38
rs1029191628 Chr.7: 94535679-94535679 on Build GRCh38
rs1048214114 Chr.7: 94535714-94535714 on Build GRCh38
rs767799178 Chr.7: 94535415-94535415 on Build GRCh38
rs529939069 Chr.7: 94535580-94535580 on Build GRCh38
rs776702521 Chr.7: 94535251-94535251 on Build GRCh38
rs570300352 Chr.7: 94535718-94535718 on Build GRCh38
rs745836412 Chr.7: 94535252-94535252 on Build GRCh38
rs17325 Chr.7: 94535526-94535526 on Build GRCh38
rs537182364 Chr.7: 94535705-94535705 on Build GRCh38
rs551529374 Chr.7: 94535670-94535670 on Build GRCh38
rs779955770 Chr.7: 94535521-94535521 on Build GRCh38
rs902133485 Chr.7: 94535654-94535654 on Build GRCh38
rs774416932 Chr.7: 94535560-94535560 on Build GRCh38
rs148670741 Chr.7: 94535427-94535427 on Build GRCh38
rs886488226 Chr.7: 94535719-94535719 on Build GRCh38
rs996085615 Chr.7: 94535529-94535529 on Build GRCh38
rs979562019 Chr.7: 94535626-94535626 on Build GRCh38
rs370376513 Chr.7: 94535308-94535308 on Build GRCh38
rs756837505 Chr.7: 94535278-94535278 on Build GRCh38
rs534971107 Chr.7: 94535649-94535649 on Build GRCh38
rs1055122124 Chr.7: 94535394-94535394 on Build GRCh38
rs757228618 Chr.7: 94535378-94535378 on Build GRCh38
rs369229758 Chr.7: 94535658-94535658 on Build GRCh38
rs753087488 Chr.7: 94535421-94535421 on Build GRCh38
rs377364571 Chr.7: 94535303-94535303 on Build GRCh38
rs968874111 Chr.7: 94535622-94535622 on Build GRCh38
rs79949295 Chr.7: 94535704-94535704 on Build GRCh38
rs182846765 Chr.7: 94535286-94535286 on Build GRCh38
rs774625028 Chr.7: 94535325-94535325 on Build GRCh38
rs372925684 Chr.7: 94535547-94535547 on Build GRCh38
rs151135732 Chr.7: 94535317-94535317 on Build GRCh38

View Full Product Details