Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130270586-130271076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA2
Entrez Gene ID
1358
Gene Name
carboxypeptidase A2
Gene Aliases
-
UniGene
Hs.490038
Gene Chromosome Location
Chr.7: 130266863-130289798 on Build GRCh38
Transcripts
NM_001869.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5529115 c.359T>C p.F120S Chr.7: 130270763-130270763 on Build GRCh38
4402557 c.381A>G p.E127E Chr.7: 130270785-130270785 on Build GRCh38
3633250 c.310G>A p.E104K Chr.7: 130270714-130270714 on Build GRCh38
6438718 c.382G>A p.E128K Chr.7: 130270786-130270786 on Build GRCh38
6634393 c.375C>A p.T125T Chr.7: 130270779-130270779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775642770 Chr.7: 130270780-130270780 on Build GRCh38
rs552354274 Chr.7: 130270995-130270995 on Build GRCh38
rs1006467804 Chr.7: 130270885-130270885 on Build GRCh38
rs770006358 Chr.7: 130270778-130270778 on Build GRCh38
rs763951828 Chr.7: 130270698-130270698 on Build GRCh38
rs767830590 Chr.7: 130270820-130270820 on Build GRCh38
rs368862290 Chr.7: 130270692-130270692 on Build GRCh38
rs114740756 Chr.7: 130270616-130270616 on Build GRCh38
rs3816830 Chr.7: 130270978-130270978 on Build GRCh38
rs148785035 Chr.7: 130271022-130271022 on Build GRCh38
rs967013278 Chr.7: 130270950-130270950 on Build GRCh38
rs745690064 Chr.7: 130270941-130270941 on Build GRCh38
rs888012613 Chr.7: 130270882-130270882 on Build GRCh38
rs1038444486 Chr.7: 130271055-130271055 on Build GRCh38
rs771042644 Chr.7: 130270673-130270673 on Build GRCh38
rs967979443 Chr.7: 130270654-130270654 on Build GRCh38
rs769029499 Chr.7: 130270805-130270805 on Build GRCh38
rs373263899 Chr.7: 130270814-130270814 on Build GRCh38
rs1022081400 Chr.7: 130271021-130271021 on Build GRCh38
rs747319767 Chr.7: 130270723-130270724 on Build GRCh38
rs758206660 Chr.7: 130270741-130270741 on Build GRCh38
rs777694068 Chr.7: 130270744-130270744 on Build GRCh38
rs373103770 Chr.7: 130270693-130270693 on Build GRCh38
rs1021351518 Chr.7: 130270825-130270825 on Build GRCh38
rs748336140 Chr.7: 130271030-130271030 on Build GRCh38
rs377750328 Chr.7: 130270709-130270709 on Build GRCh38
rs764680630 Chr.7: 130270736-130270736 on Build GRCh38
rs781593522 Chr.7: 130270763-130270763 on Build GRCh38
rs979254541 Chr.7: 130270664-130270664 on Build GRCh38
rs752382777 Chr.7: 130270613-130270613 on Build GRCh38
rs763308939 Chr.7: 130270695-130270695 on Build GRCh38
rs774574693 Chr.7: 130270717-130270717 on Build GRCh38
rs113636305 Chr.7: 130270775-130270775 on Build GRCh38
rs757534743 Chr.7: 130270758-130270758 on Build GRCh38
rs776895505 Chr.7: 130270681-130270681 on Build GRCh38
rs760435283 Chr.7: 130270719-130270719 on Build GRCh38
rs1050544061 Chr.7: 130270832-130270832 on Build GRCh38
rs2291457 Chr.7: 130270623-130270623 on Build GRCh38
rs746034700 Chr.7: 130270766-130270766 on Build GRCh38
rs759654834 Chr.7: 130270686-130270686 on Build GRCh38
rs1036480935 Chr.7: 130270934-130270934 on Build GRCh38
rs769951455 Chr.7: 130270691-130270691 on Build GRCh38
rs1035748361 Chr.7: 130270973-130270973 on Build GRCh38
rs752696326 Chr.7: 130270739-130270739 on Build GRCh38
rs749872671 Chr.7: 130270792-130270792 on Build GRCh38
rs200236688 Chr.7: 130270813-130270813 on Build GRCh38
rs753893064 Chr.7: 130270960-130270960 on Build GRCh38
rs753388349 Chr.7: 130270722-130270722 on Build GRCh38
rs774828034 Chr.7: 130270810-130270810 on Build GRCh38
rs923034168 Chr.7: 130270819-130270819 on Build GRCh38
rs994193992 Chr.7: 130270750-130270750 on Build GRCh38
rs532337123 Chr.7: 130270961-130270963 on Build GRCh38
rs780028421 Chr.7: 130270890-130270890 on Build GRCh38
rs911407139 Chr.7: 130270981-130270981 on Build GRCh38
rs776830237 Chr.7: 130270820-130270820 on Build GRCh38
rs751598143 Chr.7: 130270730-130270730 on Build GRCh38
rs757209473 Chr.7: 130270787-130270787 on Build GRCh38
rs765993385 Chr.7: 130270720-130270720 on Build GRCh38
rs778262032 Chr.7: 130270652-130270677 on Build GRCh38
rs755172992 Chr.7: 130270962-130270962 on Build GRCh38
rs1011266379 Chr.7: 130270822-130270822 on Build GRCh38
rs746754566 Chr.7: 130270745-130270745 on Build GRCh38
rs1040086552 Chr.7: 130270749-130270749 on Build GRCh38
rs991911609 Chr.7: 130270617-130270617 on Build GRCh38
rs1012899653 Chr.7: 130271010-130271010 on Build GRCh38

View Full Product Details