Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137877590-137878135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L2
Entrez Gene ID
64764
Gene Name
cAMP responsive element binding protein 3 like 2
Gene Aliases
BBF2H7
UniGene
Hs.490273
Gene Chromosome Location
Chr.7: 137874979-138002101 on Build GRCh38
Transcripts
NM_001318246.1 NM_194071.3 XM_005250541.4 XM_017012540.1 XM_017012541.1

SNP Detail

SNP ID Chromosome Location
rs572393874 Chr.7: 137877833-137877833 on Build GRCh38
rs770927796 Chr.7: 137878100-137878100 on Build GRCh38
rs1039553191 Chr.7: 137878105-137878105 on Build GRCh38
rs909462472 Chr.7: 137877978-137877978 on Build GRCh38
rs900984976 Chr.7: 137878108-137878108 on Build GRCh38
rs891437300 Chr.7: 137877721-137877721 on Build GRCh38
rs575997708 Chr.7: 137877984-137877984 on Build GRCh38
rs73444616 Chr.7: 137877936-137877936 on Build GRCh38
rs941064020 Chr.7: 137877771-137877771 on Build GRCh38
rs938451507 Chr.7: 137878038-137878038 on Build GRCh38
rs938397576 Chr.7: 137877694-137877694 on Build GRCh38
rs909396339 Chr.7: 137877649-137877649 on Build GRCh38
rs3735020 Chr.7: 137878088-137878088 on Build GRCh38
rs997626512 Chr.7: 137878109-137878109 on Build GRCh38
rs894605512 Chr.7: 137877789-137877789 on Build GRCh38
rs1017644902 Chr.7: 137877905-137877905 on Build GRCh38
rs1007186464 Chr.7: 137878089-137878089 on Build GRCh38
rs146255111 Chr.7: 137877630-137877630 on Build GRCh38
rs1046301340 Chr.7: 137878074-137878074 on Build GRCh38
rs1043406748 Chr.7: 137878076-137878076 on Build GRCh38
rs560587831 Chr.7: 137878107-137878107 on Build GRCh38
rs1024513831 Chr.7: 137877824-137877824 on Build GRCh38
rs77924208 Chr.7: 137877837-137877837 on Build GRCh38
rs1046749919 Chr.7: 137877761-137877761 on Build GRCh38
rs1013501035 Chr.7: 137877806-137877806 on Build GRCh38
rs3735018 Chr.7: 137877998-137877998 on Build GRCh38
rs1038137282 Chr.7: 137877783-137877783 on Build GRCh38
rs964799077 Chr.7: 137877836-137877836 on Build GRCh38
rs6976928 Chr.7: 137877861-137877861 on Build GRCh38
rs990214204 Chr.7: 137877672-137877672 on Build GRCh38
rs79222065 Chr.7: 137878053-137878053 on Build GRCh38
rs975644168 Chr.7: 137877924-137877924 on Build GRCh38
rs990160414 Chr.7: 137877898-137877898 on Build GRCh38
rs73729087 Chr.7: 137877922-137877922 on Build GRCh38
rs542393229 Chr.7: 137877933-137877933 on Build GRCh38
rs78165089 Chr.7: 137877817-137877817 on Build GRCh38
rs907387057 Chr.7: 137878075-137878075 on Build GRCh38
rs189544691 Chr.7: 137877647-137877647 on Build GRCh38
rs3735019 Chr.7: 137878052-137878052 on Build GRCh38
rs941010113 Chr.7: 137878007-137878007 on Build GRCh38
rs980301733 Chr.7: 137877996-137877996 on Build GRCh38
rs942235010 Chr.7: 137877986-137877986 on Build GRCh38
rs6959012 Chr.7: 137877758-137877758 on Build GRCh38
rs933957345 Chr.7: 137877760-137877760 on Build GRCh38
rs908178317 Chr.7: 137877999-137877999 on Build GRCh38
rs1006722713 Chr.7: 137877775-137877775 on Build GRCh38
rs572268193 Chr.7: 137877939-137877939 on Build GRCh38
rs1010133029 Chr.7: 137877723-137877723 on Build GRCh38
rs377403417 Chr.7: 137878045-137878045 on Build GRCh38
rs1044575859 Chr.7: 137877669-137877669 on Build GRCh38
rs945706777 Chr.7: 137878056-137878056 on Build GRCh38
rs6958842 Chr.7: 137877651-137877651 on Build GRCh38
rs749988619 Chr.7: 137877727-137877727 on Build GRCh38
rs933880723 Chr.7: 137877966-137877966 on Build GRCh38
rs886794786 Chr.7: 137877769-137877769 on Build GRCh38
rs1018486215 Chr.7: 137877832-137877832 on Build GRCh38
rs896396537 Chr.7: 137877681-137877681 on Build GRCh38
rs201726177 Chr.7: 137877853-137877853 on Build GRCh38
rs923234157 Chr.7: 137877615-137877615 on Build GRCh38
rs757936281 Chr.7: 137877752-137877752 on Build GRCh38
rs537790248 Chr.7: 137877695-137877695 on Build GRCh38

View Full Product Details