Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137878417-137878913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L2
Entrez Gene ID
64764
Gene Name
cAMP responsive element binding protein 3 like 2
Gene Aliases
BBF2H7
UniGene
Hs.490273
Gene Chromosome Location
Chr.7: 137874979-138002101 on Build GRCh38
Transcripts
NM_001318246.1 NM_194071.3 XM_005250541.4 XM_017012540.1 XM_017012541.1

SNP Detail

SNP ID Chromosome Location
rs1015467051 Chr.7: 137878509-137878509 on Build GRCh38
rs963787631 Chr.7: 137878826-137878826 on Build GRCh38
rs1005346542 Chr.7: 137878472-137878472 on Build GRCh38
rs3735021 Chr.7: 137878688-137878688 on Build GRCh38
rs145471641 Chr.7: 137878579-137878580 on Build GRCh38
rs575936992 Chr.7: 137878562-137878562 on Build GRCh38
rs940727403 Chr.7: 137878839-137878840 on Build GRCh38
rs900466294 Chr.7: 137878612-137878612 on Build GRCh38
rs538077075 Chr.7: 137878565-137878565 on Build GRCh38
rs975364333 Chr.7: 137878633-137878633 on Build GRCh38
rs886528108 Chr.7: 137878458-137878458 on Build GRCh38
rs995349995 Chr.7: 137878884-137878884 on Build GRCh38
rs975080768 Chr.7: 137878868-137878868 on Build GRCh38
rs1053203354 Chr.7: 137878580-137878580 on Build GRCh38
rs992960676 Chr.7: 137878564-137878564 on Build GRCh38
rs551292494 Chr.7: 137878744-137878744 on Build GRCh38
rs978564953 Chr.7: 137878810-137878810 on Build GRCh38
rs915533827 Chr.7: 137878536-137878536 on Build GRCh38
rs995796774 Chr.7: 137878654-137878654 on Build GRCh38
rs967168137 Chr.7: 137878573-137878573 on Build GRCh38
rs997527488 Chr.7: 137878635-137878635 on Build GRCh38
rs542743670 Chr.7: 137878710-137878710 on Build GRCh38
rs1037797959 Chr.7: 137878854-137878854 on Build GRCh38
rs940809478 Chr.7: 137878705-137878705 on Build GRCh38
rs899199084 Chr.7: 137878715-137878715 on Build GRCh38
rs969759496 Chr.7: 137878546-137878546 on Build GRCh38
rs1023253403 Chr.7: 137878888-137878888 on Build GRCh38
rs540619591 Chr.7: 137878682-137878682 on Build GRCh38
rs567302656 Chr.7: 137878821-137878821 on Build GRCh38
rs184738995 Chr.7: 137878610-137878610 on Build GRCh38
rs886916238 Chr.7: 137878693-137878693 on Build GRCh38
rs1034275000 Chr.7: 137878760-137878760 on Build GRCh38
rs1046941994 Chr.7: 137878689-137878689 on Build GRCh38
rs754982006 Chr.7: 137878645-137878645 on Build GRCh38
rs960236623 Chr.7: 137878555-137878555 on Build GRCh38
rs780950169 Chr.7: 137878648-137878648 on Build GRCh38
rs563277056 Chr.7: 137878521-137878521 on Build GRCh38
rs914765773 Chr.7: 137878583-137878583 on Build GRCh38
rs1052234277 Chr.7: 137878639-137878639 on Build GRCh38
rs201445696 Chr.7: 137878707-137878707 on Build GRCh38
rs886511737 Chr.7: 137878645-137878646 on Build GRCh38
rs896079482 Chr.7: 137878765-137878765 on Build GRCh38
rs777971642 Chr.7: 137878776-137878776 on Build GRCh38
rs981121233 Chr.7: 137878699-137878699 on Build GRCh38
rs947613541 Chr.7: 137878589-137878589 on Build GRCh38
rs149671840 Chr.7: 137878568-137878568 on Build GRCh38
rs974004920 Chr.7: 137878529-137878529 on Build GRCh38
rs886460788 Chr.7: 137878839-137878839 on Build GRCh38
rs547357347 Chr.7: 137878802-137878802 on Build GRCh38
rs922513266 Chr.7: 137878646-137878646 on Build GRCh38
rs370768188 Chr.7: 137878587-137878587 on Build GRCh38
rs561945831 Chr.7: 137878757-137878757 on Build GRCh38
rs573783249 Chr.7: 137878692-137878692 on Build GRCh38
rs1014431386 Chr.7: 137878774-137878774 on Build GRCh38
rs914711761 Chr.7: 137878741-137878741 on Build GRCh38
rs963690180 Chr.7: 137878514-137878514 on Build GRCh38
rs527633508 Chr.7: 137878775-137878775 on Build GRCh38
rs1001822135 Chr.7: 137878520-137878520 on Build GRCh38
rs988538950 Chr.7: 137878732-137878732 on Build GRCh38
rs942349208 Chr.7: 137878615-137878615 on Build GRCh38
rs562666348 Chr.7: 137878711-137878711 on Build GRCh38
rs1016282981 Chr.7: 137878483-137878483 on Build GRCh38
rs565137814 Chr.7: 137878767-137878767 on Build GRCh38
rs531371413 Chr.7: 137878735-137878735 on Build GRCh38

View Full Product Details