Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11982229-11982731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB135
Entrez Gene ID
613209
Gene Name
defensin beta 135
Gene Aliases
DEFB136
UniGene
Hs.558284
Gene Chromosome Location
Chr.8: 11982321-11984590 on Build GRCh38
Transcripts
NM_001033017.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1454469 c.24G>A p.L8L Chr.8: 11982344-11982344 on Build GRCh38
4386880 c.30C>A p.L10L Chr.8: 11982350-11982350 on Build GRCh38
338059 c.18C>G p.V6V Chr.8: 11982338-11982338 on Build GRCh38
6850880 c.12G>A p.R4R Chr.8: 11982332-11982332 on Build GRCh38
1203323 c.4G>A p.A2T Chr.8: 11982324-11982324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6991081 Chr.8: 11982290-11982290 on Build GRCh38
rs756860694 Chr.8: 11982355-11982355 on Build GRCh38
rs545693021 Chr.8: 11982483-11982483 on Build GRCh38
rs892230079 Chr.8: 11982705-11982705 on Build GRCh38
rs1000659324 Chr.8: 11982467-11982467 on Build GRCh38
rs568583244 Chr.8: 11982657-11982657 on Build GRCh38
rs58052842 Chr.8: 11982454-11982454 on Build GRCh38
rs756488141 Chr.8: 11982397-11982397 on Build GRCh38
rs537618464 Chr.8: 11982686-11982686 on Build GRCh38
rs371741237 Chr.8: 11982330-11982330 on Build GRCh38
rs774081729 Chr.8: 11982386-11982386 on Build GRCh38
rs1044749567 Chr.8: 11982667-11982667 on Build GRCh38
rs746286344 Chr.8: 11982386-11982387 on Build GRCh38
rs896419752 Chr.8: 11982407-11982407 on Build GRCh38
rs751217843 Chr.8: 11982354-11982354 on Build GRCh38
rs6985349 Chr.8: 11982562-11982562 on Build GRCh38
rs13256786 Chr.8: 11982586-11982586 on Build GRCh38
rs74479897 Chr.8: 11982488-11982488 on Build GRCh38
rs368759037 Chr.8: 11982393-11982393 on Build GRCh38
rs1032916654 Chr.8: 11982472-11982472 on Build GRCh38
rs755634575 Chr.8: 11982402-11982402 on Build GRCh38
rs755475708 Chr.8: 11982370-11982370 on Build GRCh38
rs373596001 Chr.8: 11982331-11982331 on Build GRCh38
rs889568306 Chr.8: 11982522-11982522 on Build GRCh38
rs377674176 Chr.8: 11982253-11982253 on Build GRCh38
rs916708884 Chr.8: 11982255-11982255 on Build GRCh38
rs569484376 Chr.8: 11982257-11982257 on Build GRCh38
rs555214844 Chr.8: 11982520-11982520 on Build GRCh38
rs1036207958 Chr.8: 11982642-11982642 on Build GRCh38
rs753281116 Chr.8: 11982415-11982415 on Build GRCh38
rs770353542 Chr.8: 11982270-11982270 on Build GRCh38
rs150752309 Chr.8: 11982485-11982485 on Build GRCh38
rs761501856 Chr.8: 11982387-11982387 on Build GRCh38
rs780613880 Chr.8: 11982316-11982316 on Build GRCh38
rs367637816 Chr.8: 11982306-11982306 on Build GRCh38
rs960668214 Chr.8: 11982589-11982589 on Build GRCh38
rs184595299 Chr.8: 11982352-11982352 on Build GRCh38
rs940510209 Chr.8: 11982631-11982631 on Build GRCh38
rs552113718 Chr.8: 11982624-11982624 on Build GRCh38
rs948240567 Chr.8: 11982256-11982256 on Build GRCh38
rs1006706317 Chr.8: 11982525-11982525 on Build GRCh38
rs13257076 Chr.8: 11982663-11982663 on Build GRCh38
rs549580353 Chr.8: 11982669-11982669 on Build GRCh38
rs1013229232 Chr.8: 11982440-11982440 on Build GRCh38
rs755462498 Chr.8: 11982332-11982332 on Build GRCh38
rs1013571312 Chr.8: 11982602-11982602 on Build GRCh38
rs969917547 Chr.8: 11982635-11982635 on Build GRCh38
rs201414504 Chr.8: 11982362-11982362 on Build GRCh38
rs1053564554 Chr.8: 11982698-11982698 on Build GRCh38
rs752516521 Chr.8: 11982309-11982309 on Build GRCh38
rs751068181 Chr.8: 11982273-11982276 on Build GRCh38
rs767751521 Chr.8: 11982692-11982692 on Build GRCh38
rs747259778 Chr.8: 11982376-11982376 on Build GRCh38
rs556905439 Chr.8: 11982453-11982453 on Build GRCh38
rs1014589563 Chr.8: 11982263-11982263 on Build GRCh38
rs544322528 Chr.8: 11982553-11982553 on Build GRCh38
rs956895424 Chr.8: 11982690-11982690 on Build GRCh38
rs761960747 Chr.8: 11982287-11982287 on Build GRCh38
rs112652650 Chr.8: 11982313-11982313 on Build GRCh38
rs917805876 Chr.8: 11982661-11982661 on Build GRCh38
rs764674804 Chr.8: 11982505-11982505 on Build GRCh38
rs1044759561 Chr.8: 11982441-11982441 on Build GRCh38
rs141141605 Chr.8: 11982351-11982351 on Build GRCh38
rs199948670 Chr.8: 11982336-11982336 on Build GRCh38
rs781737715 Chr.8: 11982380-11982380 on Build GRCh38
rs775214867 Chr.8: 11982350-11982350 on Build GRCh38
rs776383133 Chr.8: 11982341-11982341 on Build GRCh38
rs931423685 Chr.8: 11982367-11982367 on Build GRCh38
rs371688476 Chr.8: 11982687-11982687 on Build GRCh38
rs774736015 Chr.8: 11982284-11982284 on Build GRCh38
rs533129512 Chr.8: 11982600-11982600 on Build GRCh38
rs765191912 Chr.8: 11982347-11982347 on Build GRCh38
rs1025534964 Chr.8: 11982572-11982572 on Build GRCh38
rs758798688 Chr.8: 11982373-11982373 on Build GRCh38
rs535135999 Chr.8: 11982443-11982443 on Build GRCh38
rs763907732 Chr.8: 11982385-11982385 on Build GRCh38
rs200406777 Chr.8: 11982272-11982272 on Build GRCh38
rs769678026 Chr.8: 11982381-11982381 on Build GRCh38
rs960697873 Chr.8: 11982358-11982358 on Build GRCh38
rs552051969 Chr.8: 11982277-11982277 on Build GRCh38
rs979614877 Chr.8: 11982636-11982636 on Build GRCh38
rs561874985 Chr.8: 11982515-11982515 on Build GRCh38
rs377050782 Chr.8: 11982383-11982383 on Build GRCh38
rs776806742 Chr.8: 11982343-11982343 on Build GRCh38
rs778335297 Chr.8: 11982428-11982428 on Build GRCh38
rs925406915 Chr.8: 11982666-11982666 on Build GRCh38
rs368712206 Chr.8: 11982288-11982288 on Build GRCh38
rs369982497 Chr.8: 11982374-11982374 on Build GRCh38
rs757854541 Chr.8: 11982329-11982329 on Build GRCh38
rs187549250 Chr.8: 11982654-11982654 on Build GRCh38
rs994452320 Chr.8: 11982569-11982569 on Build GRCh38
rs759509398 Chr.8: 11982301-11982301 on Build GRCh38
rs370946939 Chr.8: 11982271-11982271 on Build GRCh38
rs904890068 Chr.8: 11982670-11982670 on Build GRCh38
rs771170568 Chr.8: 11982342-11982342 on Build GRCh38
rs1001011821 Chr.8: 11982693-11982693 on Build GRCh38
rs751945199 Chr.8: 11982435-11982435 on Build GRCh38
rs771258699 Chr.8: 11982377-11982377 on Build GRCh38
rs7004833 Chr.8: 11982502-11982502 on Build GRCh38
rs199632993 Chr.8: 11982378-11982378 on Build GRCh38
rs748516239 Chr.8: 11982372-11982372 on Build GRCh38
rs756772657 Chr.8: 11982314-11982314 on Build GRCh38
rs758028817 Chr.8: 11982311-11982311 on Build GRCh38
rs374933805 Chr.8: 11982335-11982335 on Build GRCh38
rs766512729 Chr.8: 11982291-11982291 on Build GRCh38
rs375877371 Chr.8: 11982371-11982371 on Build GRCh38
rs773200010 Chr.8: 11982289-11982289 on Build GRCh38
rs768904866 Chr.8: 11982279-11982279 on Build GRCh38
rs555222009 Chr.8: 11982630-11982630 on Build GRCh38
rs749997823 Chr.8: 11982395-11982395 on Build GRCh38
rs777477530 Chr.8: 11982312-11982312 on Build GRCh38
rs984248464 Chr.8: 11982601-11982601 on Build GRCh38
rs1038645799 Chr.8: 11982536-11982536 on Build GRCh38
rs896181066 Chr.8: 11982268-11982268 on Build GRCh38
rs759597375 Chr.8: 11982344-11982344 on Build GRCh38
rs374212432 Chr.8: 11982300-11982300 on Build GRCh38

View Full Product Details