Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 67057088-67057604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS5
Entrez Gene ID
10987
Gene Name
COP9 signalosome subunit 5
Gene Aliases
CSN5, JAB1, MOV-34, SGN5
UniGene
Hs.491912
Gene Chromosome Location
Chr.8: 67043079-67062327 on Build GRCh38
Transcripts
NM_006837.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038891 c.553G>T p.A185S Chr.8: 67057400-67057400 on Build GRCh38
3650357 c.531C>T p.S177S Chr.8: 67057422-67057422 on Build GRCh38
5723935 c.573+2T>G p.? Chr.8: 67057378-67057378 on Build GRCh38
5409783 c.548T>C p.L183P Chr.8: 67057405-67057405 on Build GRCh38
1700167 c.512A>T p.D171V Chr.8: 67057441-67057441 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs951219755 Chr.8: 67057399-67057399 on Build GRCh38
rs375721290 Chr.8: 67057347-67057347 on Build GRCh38
rs772562898 Chr.8: 67057362-67057365 on Build GRCh38
rs769509975 Chr.8: 67057375-67057375 on Build GRCh38
rs555533389 Chr.8: 67057545-67057545 on Build GRCh38
rs924901882 Chr.8: 67057457-67057457 on Build GRCh38
rs762186633 Chr.8: 67057424-67057424 on Build GRCh38
rs758189607 Chr.8: 67057428-67057428 on Build GRCh38
rs566954787 Chr.8: 67057239-67057239 on Build GRCh38
rs564731932 Chr.8: 67057167-67057167 on Build GRCh38
rs748060462 Chr.8: 67057458-67057458 on Build GRCh38
rs1035703598 Chr.8: 67057488-67057488 on Build GRCh38
rs1022656037 Chr.8: 67057171-67057171 on Build GRCh38
rs779258978 Chr.8: 67057362-67057363 on Build GRCh38
rs748016533 Chr.8: 67057374-67057374 on Build GRCh38
rs369948060 Chr.8: 67057506-67057506 on Build GRCh38
rs777436670 Chr.8: 67057469-67057469 on Build GRCh38
rs112718029 Chr.8: 67057278-67057278 on Build GRCh38
rs1007501428 Chr.8: 67057305-67057305 on Build GRCh38
rs530377403 Chr.8: 67057301-67057301 on Build GRCh38
rs560500656 Chr.8: 67057237-67057237 on Build GRCh38
rs779714353 Chr.8: 67057350-67057350 on Build GRCh38
rs764901154 Chr.8: 67057343-67057343 on Build GRCh38
rs765529888 Chr.8: 67057426-67057426 on Build GRCh38
rs566514545 Chr.8: 67057491-67057491 on Build GRCh38
rs570152372 Chr.8: 67057401-67057401 on Build GRCh38
rs567835733 Chr.8: 67057277-67057277 on Build GRCh38
rs980426293 Chr.8: 67057453-67057453 on Build GRCh38
rs369630651 Chr.8: 67057427-67057427 on Build GRCh38
rs766079032 Chr.8: 67057145-67057145 on Build GRCh38
rs547940234 Chr.8: 67057242-67057242 on Build GRCh38
rs1018179034 Chr.8: 67057359-67057359 on Build GRCh38
rs533660130 Chr.8: 67057200-67057200 on Build GRCh38
rs1035186490 Chr.8: 67057209-67057209 on Build GRCh38
rs535662317 Chr.8: 67057510-67057510 on Build GRCh38
rs746088961 Chr.8: 67057474-67057474 on Build GRCh38
rs746936878 Chr.8: 67057354-67057354 on Build GRCh38
rs765974215 Chr.8: 67057431-67057431 on Build GRCh38
rs199708915 Chr.8: 67057430-67057430 on Build GRCh38
rs973980088 Chr.8: 67057369-67057369 on Build GRCh38
rs780827219 Chr.8: 67057373-67057373 on Build GRCh38
rs754841094 Chr.8: 67057455-67057455 on Build GRCh38
rs201634747 Chr.8: 67057433-67057433 on Build GRCh38
rs780118660 Chr.8: 67057339-67057339 on Build GRCh38
rs753484577 Chr.8: 67057322-67057322 on Build GRCh38
rs772342363 Chr.8: 67057479-67057479 on Build GRCh38
rs540971942 Chr.8: 67057236-67057236 on Build GRCh38
rs749363440 Chr.8: 67057362-67057362 on Build GRCh38
rs933484063 Chr.8: 67057573-67057573 on Build GRCh38
rs936915418 Chr.8: 67057489-67057489 on Build GRCh38
rs111888399 Chr.8: 67057130-67057130 on Build GRCh38
rs755881183 Chr.8: 67057467-67057467 on Build GRCh38
rs866340690 Chr.8: 67057442-67057442 on Build GRCh38
rs185271502 Chr.8: 67057349-67057349 on Build GRCh38
rs372916530 Chr.8: 67057400-67057400 on Build GRCh38
rs962601338 Chr.8: 67057361-67057361 on Build GRCh38
rs982778961 Chr.8: 67057207-67057207 on Build GRCh38
rs755180551 Chr.8: 67057361-67057362 on Build GRCh38
rs796237799 Chr.8: 67057503-67057503 on Build GRCh38
rs368146534 Chr.8: 67057360-67057360 on Build GRCh38
rs939865299 Chr.8: 67057364-67057364 on Build GRCh38
rs561076482 Chr.8: 67057494-67057497 on Build GRCh38
rs773770131 Chr.8: 67057493-67057493 on Build GRCh38
rs1038224344 Chr.8: 67057371-67057371 on Build GRCh38
rs368706550 Chr.8: 67057398-67057398 on Build GRCh38
rs764699097 Chr.8: 67057421-67057421 on Build GRCh38
rs1051493426 Chr.8: 67057357-67057357 on Build GRCh38
rs539349512 Chr.8: 67057416-67057416 on Build GRCh38
rs975358774 Chr.8: 67057326-67057326 on Build GRCh38
rs756825396 Chr.8: 67057337-67057337 on Build GRCh38
rs780089428 Chr.8: 67057486-67057486 on Build GRCh38
rs375417871 Chr.8: 67057422-67057422 on Build GRCh38
rs373023270 Chr.8: 67057456-67057456 on Build GRCh38

View Full Product Details