Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22690140-22690678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR3
Entrez Gene ID
1960
Gene Name
early growth response 3
Gene Aliases
EGR-3, PILOT
UniGene
Hs.534313
Gene Chromosome Location
Chr.8: 22687661-22693302 on Build GRCh38
Transcripts
NM_001199880.1 NM_001199881.1 NM_004430.2 XM_005273425.3 XM_005273426.3 XM_011544429.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3432273 c.1100G>T p.G367V Chr.8: 22690537-22690537 on Build GRCh38
4680428 c.1049G>A p.R350H Chr.8: 22690588-22690588 on Build GRCh38
6652356 c.1015G>A p.G339R Chr.8: 22690622-22690622 on Build GRCh38
4680427 c.1128_1129insC p.V377fs*>12 Chr.8: 22690508-22690509 on Build GRCh38
3647313 c.1133C>T p.S378L Chr.8: 22690504-22690504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040542533 Chr.8: 22690166-22690166 on Build GRCh38
rs992871204 Chr.8: 22690323-22690323 on Build GRCh38
rs780490011 Chr.8: 22690494-22690494 on Build GRCh38
rs34614586 Chr.8: 22690384-22690385 on Build GRCh38
rs772657378 Chr.8: 22690429-22690429 on Build GRCh38
rs768743962 Chr.8: 22690511-22690511 on Build GRCh38
rs966187768 Chr.8: 22690326-22690326 on Build GRCh38
rs762696657 Chr.8: 22690455-22690455 on Build GRCh38
rs930441810 Chr.8: 22690361-22690361 on Build GRCh38
rs966572530 Chr.8: 22690539-22690539 on Build GRCh38
rs952203925 Chr.8: 22690306-22690306 on Build GRCh38
rs770751717 Chr.8: 22690535-22690535 on Build GRCh38
rs760574595 Chr.8: 22690478-22690478 on Build GRCh38
rs200159562 Chr.8: 22690587-22690587 on Build GRCh38
rs771889037 Chr.8: 22690554-22690554 on Build GRCh38
rs201598642 Chr.8: 22690516-22690516 on Build GRCh38
rs921614228 Chr.8: 22690634-22690634 on Build GRCh38
rs1034724924 Chr.8: 22690245-22690245 on Build GRCh38
rs1028527665 Chr.8: 22690256-22690256 on Build GRCh38
rs1012506834 Chr.8: 22690257-22690257 on Build GRCh38
rs753994058 Chr.8: 22690482-22690482 on Build GRCh38
rs775699693 Chr.8: 22690433-22690433 on Build GRCh38
rs747444731 Chr.8: 22690438-22690438 on Build GRCh38
rs762200054 Chr.8: 22690524-22690532 on Build GRCh38
rs537313070 Chr.8: 22690386-22690386 on Build GRCh38
rs149624410 Chr.8: 22690605-22690605 on Build GRCh38
rs1000451981 Chr.8: 22690289-22690289 on Build GRCh38
rs547856396 Chr.8: 22690297-22690297 on Build GRCh38
rs767530238 Chr.8: 22690472-22690472 on Build GRCh38
rs773060036 Chr.8: 22690444-22690444 on Build GRCh38
rs931951261 Chr.8: 22690355-22690355 on Build GRCh38
rs900280615 Chr.8: 22690249-22690249 on Build GRCh38
rs762000735 Chr.8: 22690593-22690593 on Build GRCh38
rs769189657 Chr.8: 22690537-22690547 on Build GRCh38
rs867051535 Chr.8: 22690598-22690598 on Build GRCh38
rs915137293 Chr.8: 22690369-22690369 on Build GRCh38
rs894817902 Chr.8: 22690434-22690434 on Build GRCh38
rs769568286 Chr.8: 22690443-22690443 on Build GRCh38
rs200707775 Chr.8: 22690620-22690620 on Build GRCh38
rs773703007 Chr.8: 22690548-22690548 on Build GRCh38
rs566030472 Chr.8: 22690325-22690325 on Build GRCh38
rs568431773 Chr.8: 22690274-22690274 on Build GRCh38
rs968837532 Chr.8: 22690278-22690278 on Build GRCh38
rs749768945 Chr.8: 22690508-22690509 on Build GRCh38
rs1027873202 Chr.8: 22690292-22690292 on Build GRCh38
rs541187152 Chr.8: 22690232-22690232 on Build GRCh38
rs780680811 Chr.8: 22690515-22690515 on Build GRCh38
rs576597699 Chr.8: 22690380-22690380 on Build GRCh38
rs560246514 Chr.8: 22690411-22690411 on Build GRCh38
rs983595495 Chr.8: 22690311-22690311 on Build GRCh38
rs539954184 Chr.8: 22690352-22690352 on Build GRCh38
rs1043644607 Chr.8: 22690590-22690591 on Build GRCh38
rs759151796 Chr.8: 22690549-22690549 on Build GRCh38
rs757963779 Chr.8: 22690348-22690348 on Build GRCh38
rs138509871 Chr.8: 22690509-22690509 on Build GRCh38
rs750467634 Chr.8: 22690608-22690608 on Build GRCh38
rs373801955 Chr.8: 22690210-22690210 on Build GRCh38
rs996324450 Chr.8: 22690253-22690253 on Build GRCh38
rs774877683 Chr.8: 22690551-22690553 on Build GRCh38
rs141587813 Chr.8: 22690517-22690517 on Build GRCh38
rs763838150 Chr.8: 22690569-22690569 on Build GRCh38
rs765943708 Chr.8: 22690465-22690465 on Build GRCh38
rs753655773 Chr.8: 22690579-22690579 on Build GRCh38
rs150892020 Chr.8: 22690518-22690518 on Build GRCh38
rs757503730 Chr.8: 22690489-22690489 on Build GRCh38
rs1031829352 Chr.8: 22690479-22690479 on Build GRCh38
rs535861312 Chr.8: 22690296-22690296 on Build GRCh38
rs555649419 Chr.8: 22690392-22690392 on Build GRCh38
rs959293282 Chr.8: 22690252-22690252 on Build GRCh38
rs759365414 Chr.8: 22690468-22690468 on Build GRCh38
rs1000336113 Chr.8: 22690439-22690439 on Build GRCh38
rs142198577 Chr.8: 22690557-22690557 on Build GRCh38
rs970207530 Chr.8: 22690336-22690336 on Build GRCh38
rs1042249889 Chr.8: 22690381-22690381 on Build GRCh38
rs746321351 Chr.8: 22690428-22690428 on Build GRCh38
rs976571798 Chr.8: 22690333-22690333 on Build GRCh38
rs774016879 Chr.8: 22690467-22690467 on Build GRCh38
rs760612686 Chr.8: 22690567-22690567 on Build GRCh38
rs900158174 Chr.8: 22690418-22690418 on Build GRCh38
rs147637858 Chr.8: 22690368-22690368 on Build GRCh38
rs778916249 Chr.8: 22690498-22690498 on Build GRCh38
rs34545192 Chr.8: 22690623-22690623 on Build GRCh38
rs200374053 Chr.8: 22690474-22690474 on Build GRCh38
rs755561201 Chr.8: 22690626-22690626 on Build GRCh38

View Full Product Details