Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6554564-6555076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLDC
Entrez Gene ID
2731
Gene Name
glycine decarboxylase
Gene Aliases
GCE, GCSP, HYGN1
UniGene
Hs.584238
Gene Chromosome Location
Chr.9: 6532464-6645692 on Build GRCh38
Transcripts
NM_000170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6183597 c.2261A>G p.K754R Chr.9: 6554723-6554723 on Build GRCh38
3675349 c.2259C>A p.H753Q Chr.9: 6554725-6554725 on Build GRCh38
2777838 c.2209A>G p.I737V Chr.9: 6554775-6554775 on Build GRCh38
6400880 c.2239G>T p.V747F Chr.9: 6554745-6554745 on Build GRCh38
5034729 c.2213G>A p.C738Y Chr.9: 6554771-6554771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951358115 Chr.9: 6554988-6554988 on Build GRCh38
rs914067399 Chr.9: 6554858-6554858 on Build GRCh38
rs112211409 Chr.9: 6554997-6554997 on Build GRCh38
rs762611136 Chr.9: 6554676-6554676 on Build GRCh38
rs954961288 Chr.9: 6554722-6554722 on Build GRCh38
rs565834029 Chr.9: 6554677-6554677 on Build GRCh38
rs750384225 Chr.9: 6554687-6554687 on Build GRCh38
rs767258496 Chr.9: 6554793-6554793 on Build GRCh38
rs1039468392 Chr.9: 6554658-6554658 on Build GRCh38
rs149068409 Chr.9: 6554602-6554602 on Build GRCh38
rs891931666 Chr.9: 6555030-6555030 on Build GRCh38
rs375733985 Chr.9: 6554743-6554743 on Build GRCh38
rs375850752 Chr.9: 6554772-6554772 on Build GRCh38
rs747219893 Chr.9: 6554712-6554712 on Build GRCh38
rs751798296 Chr.9: 6554765-6554765 on Build GRCh38
rs121964980 Chr.9: 6554768-6554768 on Build GRCh38
rs549929780 Chr.9: 6554599-6554599 on Build GRCh38
rs777041727 Chr.9: 6554671-6554671 on Build GRCh38
rs370855096 Chr.9: 6554799-6554799 on Build GRCh38
rs765661419 Chr.9: 6554674-6554674 on Build GRCh38
rs746687513 Chr.9: 6554626-6554627 on Build GRCh38
rs564642740 Chr.9: 6554963-6554963 on Build GRCh38
rs907708315 Chr.9: 6554940-6554940 on Build GRCh38
rs12683164 Chr.9: 6554589-6554589 on Build GRCh38
rs386833545 Chr.9: 6554783-6554783 on Build GRCh38
rs766368793 Chr.9: 6554753-6554753 on Build GRCh38
rs1005329653 Chr.9: 6554643-6554643 on Build GRCh38
rs574333403 Chr.9: 6554849-6554849 on Build GRCh38
rs772351971 Chr.9: 6554656-6554656 on Build GRCh38
rs749882600 Chr.9: 6554746-6554746 on Build GRCh38
rs560335288 Chr.9: 6554882-6554882 on Build GRCh38
rs386833553 Chr.9: 6554673-6554673 on Build GRCh38
rs759989554 Chr.9: 6554592-6554592 on Build GRCh38
rs1001872049 Chr.9: 6554891-6554891 on Build GRCh38
rs764949509 Chr.9: 6554825-6554825 on Build GRCh38
rs1020492830 Chr.9: 6554883-6554883 on Build GRCh38
rs111353471 Chr.9: 6555004-6555004 on Build GRCh38
rs760700185 Chr.9: 6554752-6554752 on Build GRCh38
rs386833551 Chr.9: 6554691-6554691 on Build GRCh38
rs386833552 Chr.9: 6554678-6554678 on Build GRCh38
rs754894930 Chr.9: 6555016-6555016 on Build GRCh38
rs770318617 Chr.9: 6554770-6554770 on Build GRCh38
rs780954019 Chr.9: 6554631-6554631 on Build GRCh38
rs547442356 Chr.9: 6554617-6554617 on Build GRCh38
rs190579680 Chr.9: 6554923-6554923 on Build GRCh38
rs770421041 Chr.9: 6554939-6554939 on Build GRCh38
rs373878910 Chr.9: 6554944-6554944 on Build GRCh38
rs1053289301 Chr.9: 6555029-6555029 on Build GRCh38
rs386833547 Chr.9: 6554726-6554726 on Build GRCh38
rs769035358 Chr.9: 6554741-6554741 on Build GRCh38
rs955760026 Chr.9: 6554651-6554651 on Build GRCh38
rs756022662 Chr.9: 6554638-6554638 on Build GRCh38
rs760077981 Chr.9: 6554672-6554672 on Build GRCh38
rs749773420 Chr.9: 6554779-6554779 on Build GRCh38
rs746383783 Chr.9: 6554720-6554720 on Build GRCh38
rs386833549 Chr.9: 6554703-6554703 on Build GRCh38
rs143119940 Chr.9: 6554781-6554781 on Build GRCh38
rs533775952 Chr.9: 6554680-6554680 on Build GRCh38
rs970388208 Chr.9: 6554707-6554707 on Build GRCh38
rs773354023 Chr.9: 6554661-6554661 on Build GRCh38
rs386833550 Chr.9: 6554700-6554700 on Build GRCh38
rs765951835 Chr.9: 6554802-6554802 on Build GRCh38
rs983288780 Chr.9: 6554924-6554924 on Build GRCh38
rs770961258 Chr.9: 6554713-6554713 on Build GRCh38
rs757437950 Chr.9: 6554767-6554767 on Build GRCh38
rs753873563 Chr.9: 6554810-6554810 on Build GRCh38
rs144317480 Chr.9: 6554755-6554755 on Build GRCh38
rs148775220 Chr.9: 6554769-6554769 on Build GRCh38
rs776772464 Chr.9: 6554715-6554715 on Build GRCh38
rs753857149 Chr.9: 6555011-6555011 on Build GRCh38
rs899542521 Chr.9: 6554847-6554847 on Build GRCh38
rs770745274 Chr.9: 6554665-6554666 on Build GRCh38
rs949858682 Chr.9: 6555036-6555036 on Build GRCh38
rs765937255 Chr.9: 6554966-6554966 on Build GRCh38
rs926215065 Chr.9: 6554981-6554981 on Build GRCh38
rs936235295 Chr.9: 6555023-6555023 on Build GRCh38
rs969726878 Chr.9: 6554890-6554890 on Build GRCh38
rs747009828 Chr.9: 6554662-6554662 on Build GRCh38
rs185654900 Chr.9: 6554605-6554605 on Build GRCh38
rs779980458 Chr.9: 6554640-6554640 on Build GRCh38
rs775769857 Chr.9: 6554748-6554748 on Build GRCh38
rs904704773 Chr.9: 6554838-6554838 on Build GRCh38
rs121964975 Chr.9: 6554716-6554718 on Build GRCh38
rs748447557 Chr.9: 6554645-6554645 on Build GRCh38
rs951947692 Chr.9: 6554922-6554922 on Build GRCh38
rs757241907 Chr.9: 6554625-6554625 on Build GRCh38
rs1031396184 Chr.9: 6554646-6554646 on Build GRCh38
rs999965197 Chr.9: 6554614-6554614 on Build GRCh38
rs370427322 Chr.9: 6554831-6554831 on Build GRCh38
rs751114163 Chr.9: 6554679-6554679 on Build GRCh38
rs1050754710 Chr.9: 6554867-6554867 on Build GRCh38
rs1031003330 Chr.9: 6554686-6554686 on Build GRCh38
rs532009973 Chr.9: 6554593-6554593 on Build GRCh38
rs1053260532 Chr.9: 6554854-6554854 on Build GRCh38
rs370061961 Chr.9: 6554754-6554754 on Build GRCh38
rs372693840 Chr.9: 6554740-6554740 on Build GRCh38
rs116133476 Chr.9: 6554968-6554968 on Build GRCh38
rs777816136 Chr.9: 6554704-6554704 on Build GRCh38
rs761374321 Chr.9: 6554791-6554791 on Build GRCh38
rs541948151 Chr.9: 6554857-6554857 on Build GRCh38
rs2274874 Chr.9: 6554844-6554844 on Build GRCh38
rs961447122 Chr.9: 6554942-6554942 on Build GRCh38
rs765624698 Chr.9: 6554731-6554731 on Build GRCh38
rs370479312 Chr.9: 6554846-6554846 on Build GRCh38
rs386833548 Chr.9: 6554715-6554717 on Build GRCh38
rs572157378 Chr.9: 6554893-6554893 on Build GRCh38
rs779998377 Chr.9: 6554694-6554694 on Build GRCh38
rs745786774 Chr.9: 6554636-6554636 on Build GRCh38
rs763601156 Chr.9: 6554624-6554624 on Build GRCh38
rs922593132 Chr.9: 6554797-6554797 on Build GRCh38
rs386833546 Chr.9: 6554770-6554771 on Build GRCh38
rs1024330693 Chr.9: 6554980-6554980 on Build GRCh38
rs901463355 Chr.9: 6554598-6554598 on Build GRCh38
rs767001572 Chr.9: 6554745-6554745 on Build GRCh38
rs771496861 Chr.9: 6554666-6554666 on Build GRCh38
rs1024394917 Chr.9: 6554880-6554880 on Build GRCh38
rs111361364 Chr.9: 6554983-6554983 on Build GRCh38
rs749032015 Chr.9: 6554695-6554695 on Build GRCh38
rs759529980 Chr.9: 6554812-6554812 on Build GRCh38
rs1027603308 Chr.9: 6554913-6554913 on Build GRCh38
rs13440229 Chr.9: 6554800-6554800 on Build GRCh38
rs771697455 Chr.9: 6554792-6554792 on Build GRCh38
rs775858027 Chr.9: 6554736-6554736 on Build GRCh38

View Full Product Details