Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179990400-179990930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP350
Entrez Gene ID
9857
Gene Name
centrosomal protein 350
Gene Aliases
CAP350, GM133
UniGene
Hs.413045
Gene Chromosome Location
Chr.1: 179954773-180114880 on Build GRCh38
Transcripts
NM_014810.4 XM_011510191.2 XM_011510198.2 XM_017002929.1 XM_017002930.1 XM_017002931.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6583623 c.121-5T>G p.? Chr.1: 179990502-179990502 on Build GRCh38
900339 c.152C>T p.A51V Chr.1: 179990538-179990538 on Build GRCh38
4671654 c.172T>G p.C58G Chr.1: 179990558-179990558 on Build GRCh38
1284094 c.187G>T p.D63Y Chr.1: 179990573-179990573 on Build GRCh38
5174331 c.231delA p.D79fs*38 Chr.1: 179990617-179990617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765087167 Chr.1: 179990571-179990571 on Build GRCh38
rs760146553 Chr.1: 179990501-179990501 on Build GRCh38
rs115186595 Chr.1: 179990589-179990589 on Build GRCh38
rs2477109 Chr.1: 179990899-179990899 on Build GRCh38
rs1033724137 Chr.1: 179990575-179990575 on Build GRCh38
rs777459010 Chr.1: 179990633-179990633 on Build GRCh38
rs550159034 Chr.1: 179990786-179990786 on Build GRCh38
rs570069992 Chr.1: 179990800-179990800 on Build GRCh38
rs750564859 Chr.1: 179990603-179990603 on Build GRCh38
rs770473993 Chr.1: 179990542-179990542 on Build GRCh38
rs758718352 Chr.1: 179990623-179990624 on Build GRCh38
rs780482124 Chr.1: 179990604-179990604 on Build GRCh38
rs759427074 Chr.1: 179990570-179990570 on Build GRCh38
rs777219780 Chr.1: 179990634-179990634 on Build GRCh38
rs889676821 Chr.1: 179990674-179990674 on Build GRCh38
rs779430253 Chr.1: 179990550-179990550 on Build GRCh38
rs749626002 Chr.1: 179990475-179990475 on Build GRCh38
rs922563124 Chr.1: 179990846-179990846 on Build GRCh38
rs775886906 Chr.1: 179990458-179990458 on Build GRCh38
rs567738179 Chr.1: 179990689-179990689 on Build GRCh38
rs772805916 Chr.1: 179990492-179990492 on Build GRCh38
rs764377285 Chr.1: 179990579-179990579 on Build GRCh38
rs558851624 Chr.1: 179990888-179990888 on Build GRCh38
rs770596942 Chr.1: 179990668-179990668 on Build GRCh38
rs751873814 Chr.1: 179990583-179990583 on Build GRCh38
rs977068517 Chr.1: 179990828-179990828 on Build GRCh38
rs536759077 Chr.1: 179990746-179990746 on Build GRCh38
rs184884786 Chr.1: 179990808-179990808 on Build GRCh38
rs781372672 Chr.1: 179990591-179990591 on Build GRCh38
rs927555328 Chr.1: 179990442-179990442 on Build GRCh38
rs961255750 Chr.1: 179990820-179990820 on Build GRCh38
rs955733200 Chr.1: 179990755-179990755 on Build GRCh38
rs369434488 Chr.1: 179990480-179990480 on Build GRCh38
rs762716692 Chr.1: 179990578-179990578 on Build GRCh38
rs940731724 Chr.1: 179990556-179990556 on Build GRCh38
rs1015859222 Chr.1: 179990721-179990721 on Build GRCh38
rs1010215096 Chr.1: 179990507-179990507 on Build GRCh38
rs140306241 Chr.1: 179990615-179990615 on Build GRCh38
rs547554278 Chr.1: 179990681-179990681 on Build GRCh38
rs907878331 Chr.1: 179990487-179990487 on Build GRCh38
rs749658362 Chr.1: 179990613-179990613 on Build GRCh38
rs771840070 Chr.1: 179990745-179990745 on Build GRCh38
rs746612764 Chr.1: 179990646-179990646 on Build GRCh38
rs1019661728 Chr.1: 179990748-179990748 on Build GRCh38
rs763224214 Chr.1: 179990837-179990837 on Build GRCh38
rs552053057 Chr.1: 179990572-179990572 on Build GRCh38

View Full Product Details