Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83298611-83299119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD3
Entrez Gene ID
257019
Gene Name
FERM domain containing 3
Gene Aliases
4.1O, EPB41L4O, EPB41LO, P410
UniGene
Hs.127535
Gene Chromosome Location
Chr.9: 83242990-83560208 on Build GRCh38
Transcripts
NM_001244959.1 NM_001244960.1 NM_001244961.1 NM_174938.5 XM_017014588.1 XM_017014589.1 XM_017014590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5930284 c.1064T>G p.V355G Chr.9: 83298754-83298754 on Build GRCh38
5930285 c.482T>G p.V161G Chr.9: 83298754-83298754 on Build GRCh38
5064118 c.750G>T p.E250D Chr.9: 83298756-83298756 on Build GRCh38
3659159 c.455C>T p.S152F Chr.9: 83298781-83298781 on Build GRCh38
2156694 c.738G>A p.R246R Chr.9: 83298768-83298768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934723344 Chr.9: 83298677-83298677 on Build GRCh38
rs555814173 Chr.9: 83298844-83298844 on Build GRCh38
rs923525596 Chr.9: 83298660-83298660 on Build GRCh38
rs1015263141 Chr.9: 83298832-83298833 on Build GRCh38
rs769310741 Chr.9: 83298857-83298857 on Build GRCh38
rs1030325692 Chr.9: 83299020-83299020 on Build GRCh38
rs759952321 Chr.9: 83299075-83299075 on Build GRCh38
rs768069610 Chr.9: 83299085-83299085 on Build GRCh38
rs780679222 Chr.9: 83298854-83298854 on Build GRCh38
rs768697927 Chr.9: 83298845-83298845 on Build GRCh38
rs773527868 Chr.9: 83298852-83298855 on Build GRCh38
rs754134406 Chr.9: 83298829-83298829 on Build GRCh38
rs749144582 Chr.9: 83298712-83298713 on Build GRCh38
rs899684653 Chr.9: 83298718-83298718 on Build GRCh38
rs768021957 Chr.9: 83298716-83298716 on Build GRCh38
rs376954266 Chr.9: 83298816-83298816 on Build GRCh38
rs772860418 Chr.9: 83298765-83298765 on Build GRCh38
rs760136820 Chr.9: 83298800-83298800 on Build GRCh38
rs552043234 Chr.9: 83298827-83298827 on Build GRCh38
rs772248048 Chr.9: 83298712-83298712 on Build GRCh38
rs944681762 Chr.9: 83299018-83299018 on Build GRCh38
rs147357950 Chr.9: 83298731-83298731 on Build GRCh38
rs375118684 Chr.9: 83299073-83299073 on Build GRCh38
rs945740720 Chr.9: 83298750-83298750 on Build GRCh38
rs767479443 Chr.9: 83298802-83298802 on Build GRCh38
rs997468646 Chr.9: 83299016-83299016 on Build GRCh38
rs200854226 Chr.9: 83298855-83298855 on Build GRCh38
rs1001479938 Chr.9: 83298870-83298870 on Build GRCh38
rs926104216 Chr.9: 83298973-83298973 on Build GRCh38
rs774834059 Chr.9: 83299074-83299074 on Build GRCh38
rs758089919 Chr.9: 83298830-83298830 on Build GRCh38
rs761884144 Chr.9: 83298804-83298804 on Build GRCh38
rs774712672 Chr.9: 83299070-83299070 on Build GRCh38
rs771078003 Chr.9: 83299074-83299074 on Build GRCh38
rs369387550 Chr.9: 83298711-83298711 on Build GRCh38
rs776168785 Chr.9: 83298719-83298719 on Build GRCh38
rs148420261 Chr.9: 83299004-83299004 on Build GRCh38
rs899791213 Chr.9: 83299064-83299064 on Build GRCh38
rs372995347 Chr.9: 83298813-83298813 on Build GRCh38
rs375646971 Chr.9: 83298699-83298699 on Build GRCh38
rs766794362 Chr.9: 83298782-83298782 on Build GRCh38
rs779939909 Chr.9: 83298831-83298831 on Build GRCh38
rs747015951 Chr.9: 83299069-83299069 on Build GRCh38
rs566961126 Chr.9: 83298663-83298663 on Build GRCh38
rs986256555 Chr.9: 83298668-83298668 on Build GRCh38
rs370135582 Chr.9: 83298836-83298836 on Build GRCh38
rs746589122 Chr.9: 83298672-83298672 on Build GRCh38
rs769506542 Chr.9: 83298764-83298764 on Build GRCh38
rs868324808 Chr.9: 83298691-83298691 on Build GRCh38
rs368454551 Chr.9: 83299087-83299087 on Build GRCh38
rs373238207 Chr.9: 83298840-83298840 on Build GRCh38
rs1055616433 Chr.9: 83298878-83298878 on Build GRCh38
rs772807069 Chr.9: 83298865-83298865 on Build GRCh38
rs1026763361 Chr.9: 83298798-83298798 on Build GRCh38
rs1015014675 Chr.9: 83298954-83298954 on Build GRCh38
rs887635977 Chr.9: 83298946-83298946 on Build GRCh38
rs762703661 Chr.9: 83299071-83299071 on Build GRCh38
rs934734586 Chr.9: 83298979-83298979 on Build GRCh38
rs11140011 Chr.9: 83298675-83298675 on Build GRCh38
rs58813904 Chr.9: 83298947-83298947 on Build GRCh38
rs751974611 Chr.9: 83298794-83298794 on Build GRCh38
rs763249143 Chr.9: 83298781-83298781 on Build GRCh38

View Full Product Details