Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83739343-83739846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GKAP1
Entrez Gene ID
80318
Gene Name
G kinase anchoring protein 1
Gene Aliases
FKSG21, GKAP42
UniGene
Hs.522255
Gene Chromosome Location
Chr.9: 83739421-83817837 on Build GRCh38
Transcripts
NM_001135953.1 NM_025211.3 XM_005252241.2 XM_006717293.1 XM_011519058.2 XM_017015171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5943329 c.1054-5delT p.? Chr.9: 83739749-83739749 on Build GRCh38
311466 c.1057G>T p.G353C Chr.9: 83739741-83739741 on Build GRCh38
1674576 c.1060A>T p.R354* Chr.9: 83739738-83739738 on Build GRCh38
3659188 c.1090C>T p.Q364* Chr.9: 83739708-83739708 on Build GRCh38
342639 c.1061G>A p.R354K Chr.9: 83739737-83739737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984833742 Chr.9: 83739675-83739675 on Build GRCh38
rs147000647 Chr.9: 83739384-83739384 on Build GRCh38
rs4750 Chr.9: 83739596-83739596 on Build GRCh38
rs767757263 Chr.9: 83739754-83739754 on Build GRCh38
rs779759891 Chr.9: 83739685-83739685 on Build GRCh38
rs762130046 Chr.9: 83739721-83739721 on Build GRCh38
rs533002247 Chr.9: 83739441-83739441 on Build GRCh38
rs889484087 Chr.9: 83739489-83739489 on Build GRCh38
rs568352866 Chr.9: 83739639-83739639 on Build GRCh38
rs766306669 Chr.9: 83739744-83739744 on Build GRCh38
rs907464022 Chr.9: 83739590-83739590 on Build GRCh38
rs745613597 Chr.9: 83739457-83739457 on Build GRCh38
rs753727864 Chr.9: 83739764-83739764 on Build GRCh38
rs779007668 Chr.9: 83739768-83739768 on Build GRCh38
rs767213117 Chr.9: 83739604-83739604 on Build GRCh38
rs972643001 Chr.9: 83739404-83739404 on Build GRCh38
rs865804547 Chr.9: 83739819-83739819 on Build GRCh38
rs745393010 Chr.9: 83739670-83739670 on Build GRCh38
rs746503920 Chr.9: 83739795-83739795 on Build GRCh38
rs183725773 Chr.9: 83739785-83739785 on Build GRCh38
rs551358005 Chr.9: 83739520-83739520 on Build GRCh38
rs977669233 Chr.9: 83739808-83739809 on Build GRCh38
rs569507314 Chr.9: 83739538-83739538 on Build GRCh38
rs746705362 Chr.9: 83739686-83739686 on Build GRCh38
rs761658703 Chr.9: 83739749-83739751 on Build GRCh38
rs899080447 Chr.9: 83739579-83739579 on Build GRCh38
rs902845200 Chr.9: 83739463-83739463 on Build GRCh38
rs1010826823 Chr.9: 83739562-83739562 on Build GRCh38
rs3217184 Chr.9: 83739418-83739419 on Build GRCh38
rs759736903 Chr.9: 83739747-83739747 on Build GRCh38
rs993393796 Chr.9: 83739587-83739590 on Build GRCh38
rs947663700 Chr.9: 83739772-83739772 on Build GRCh38
rs200846020 Chr.9: 83739671-83739671 on Build GRCh38
rs757308868 Chr.9: 83739653-83739653 on Build GRCh38
rs1024518930 Chr.9: 83739574-83739574 on Build GRCh38
rs763036939 Chr.9: 83739743-83739743 on Build GRCh38
rs750317646 Chr.9: 83739769-83739769 on Build GRCh38
rs922516232 Chr.9: 83739779-83739779 on Build GRCh38
rs535646275 Chr.9: 83739701-83739701 on Build GRCh38
rs933647748 Chr.9: 83739446-83739446 on Build GRCh38
rs769676596 Chr.9: 83739536-83739536 on Build GRCh38
rs1007817226 Chr.9: 83739541-83739541 on Build GRCh38
rs757069336 Chr.9: 83739765-83739765 on Build GRCh38
rs568665080 Chr.9: 83739817-83739817 on Build GRCh38
rs537174819 Chr.9: 83739595-83739595 on Build GRCh38
rs773663461 Chr.9: 83739742-83739742 on Build GRCh38
rs891953532 Chr.9: 83739543-83739543 on Build GRCh38
rs997307470 Chr.9: 83739517-83739517 on Build GRCh38
rs779424988 Chr.9: 83739749-83739750 on Build GRCh38
rs770312752 Chr.9: 83739738-83739738 on Build GRCh38
rs781168580 Chr.9: 83739413-83739413 on Build GRCh38
rs768690901 Chr.9: 83739748-83739749 on Build GRCh38
rs748846032 Chr.9: 83739749-83739749 on Build GRCh38
rs751656430 Chr.9: 83739745-83739745 on Build GRCh38
rs1034296543 Chr.9: 83739635-83739635 on Build GRCh38
rs992072002 Chr.9: 83739706-83739706 on Build GRCh38
rs369376225 Chr.9: 83739712-83739712 on Build GRCh38
rs758350231 Chr.9: 83739778-83739778 on Build GRCh38
rs554092166 Chr.9: 83739711-83739711 on Build GRCh38
rs377667957 Chr.9: 83739475-83739475 on Build GRCh38
rs771764362 Chr.9: 83739764-83739764 on Build GRCh38
rs909717579 Chr.9: 83739383-83739383 on Build GRCh38
rs914713882 Chr.9: 83739763-83739763 on Build GRCh38
rs778963072 Chr.9: 83739660-83739660 on Build GRCh38
rs1041413142 Chr.9: 83739390-83739390 on Build GRCh38

View Full Product Details