Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133470317-133470818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACFD1
Entrez Gene ID
11094
Gene Name
calcium channel flower domain containing 1
Gene Aliases
C9orf7, D9S2135, FLOWER
UniGene
Hs.62003
Gene Chromosome Location
Chr.9: 133459965-133470787 on Build GRCh38
Transcripts
NM_001135775.2 NM_001242369.1 NM_001242370.1 NM_017586.3 XM_011518181.2 XM_011518182.1 XM_011518184.1 XM_011518185.2 XM_011518186.1

SNP Detail

SNP ID Chromosome Location
rs1048086231 Chr.9: 133470508-133470508 on Build GRCh38
rs772296587 Chr.9: 133470763-133470763 on Build GRCh38
rs45489391 Chr.9: 133470753-133470753 on Build GRCh38
rs191685507 Chr.9: 133470596-133470596 on Build GRCh38
rs966132760 Chr.9: 133470772-133470772 on Build GRCh38
rs979839701 Chr.9: 133470477-133470477 on Build GRCh38
rs1053085634 Chr.9: 133470728-133470728 on Build GRCh38
rs930352814 Chr.9: 133470494-133470494 on Build GRCh38
rs587604258 Chr.9: 133470607-133470607 on Build GRCh38
rs951798143 Chr.9: 133470662-133470662 on Build GRCh38
rs782454764 Chr.9: 133470463-133470463 on Build GRCh38
rs993240929 Chr.9: 133470391-133470391 on Build GRCh38
rs587604153 Chr.9: 133470531-133470531 on Build GRCh38
rs146501678 Chr.9: 133470663-133470663 on Build GRCh38
rs904640220 Chr.9: 133470714-133470714 on Build GRCh38
rs756574135 Chr.9: 133470399-133470399 on Build GRCh38
rs939088737 Chr.9: 133470762-133470762 on Build GRCh38
rs1054379 Chr.9: 133470702-133470702 on Build GRCh38
rs187098485 Chr.9: 133470585-133470585 on Build GRCh38
rs369375908 Chr.9: 133470653-133470653 on Build GRCh38
rs112767008 Chr.9: 133470730-133470730 on Build GRCh38
rs1024541027 Chr.9: 133470737-133470737 on Build GRCh38
rs1022126314 Chr.9: 133470696-133470696 on Build GRCh38
rs138121308 Chr.9: 133470521-133470521 on Build GRCh38
rs920824127 Chr.9: 133470751-133470751 on Build GRCh38
rs898955968 Chr.9: 133470369-133470369 on Build GRCh38
rs944529319 Chr.9: 133470528-133470528 on Build GRCh38
rs921751887 Chr.9: 133470703-133470703 on Build GRCh38
rs954938638 Chr.9: 133470703-133470705 on Build GRCh38
rs115934943 Chr.9: 133470691-133470691 on Build GRCh38
rs920232119 Chr.9: 133470479-133470479 on Build GRCh38
rs977478556 Chr.9: 133470538-133470538 on Build GRCh38
rs1014921865 Chr.9: 133470635-133470635 on Build GRCh38
rs1014429813 Chr.9: 133470732-133470732 on Build GRCh38
rs951794170 Chr.9: 133470761-133470761 on Build GRCh38
rs917422278 Chr.9: 133470412-133470412 on Build GRCh38
rs1031947810 Chr.9: 133470380-133470380 on Build GRCh38
rs782615230 Chr.9: 133470530-133470530 on Build GRCh38
rs1028306547 Chr.9: 133470354-133470354 on Build GRCh38
rs904506682 Chr.9: 133470593-133470593 on Build GRCh38
rs902370706 Chr.9: 133470698-133470699 on Build GRCh38
rs587727854 Chr.9: 133470349-133470349 on Build GRCh38
rs999286078 Chr.9: 133470699-133470699 on Build GRCh38
rs149166186 Chr.9: 133470606-133470606 on Build GRCh38
rs749596475 Chr.9: 133470650-133470650 on Build GRCh38
rs112092403 Chr.9: 133470471-133470471 on Build GRCh38
rs993046255 Chr.9: 133470407-133470407 on Build GRCh38
rs936093125 Chr.9: 133470715-133470715 on Build GRCh38
rs921884737 Chr.9: 133470794-133470794 on Build GRCh38
rs910827849 Chr.9: 133470595-133470595 on Build GRCh38
rs782536929 Chr.9: 133470362-133470362 on Build GRCh38
rs587740698 Chr.9: 133470729-133470729 on Build GRCh38
rs1010733045 Chr.9: 133470520-133470520 on Build GRCh38
rs373192527 Chr.9: 133470742-133470742 on Build GRCh38
rs1037853894 Chr.9: 133470351-133470351 on Build GRCh38
rs1000844428 Chr.9: 133470378-133470378 on Build GRCh38
rs377314373 Chr.9: 133470748-133470748 on Build GRCh38
rs587625250 Chr.9: 133470373-133470373 on Build GRCh38
rs1049838294 Chr.9: 133470713-133470713 on Build GRCh38
rs996907945 Chr.9: 133470571-133470571 on Build GRCh38
rs900357096 Chr.9: 133470543-133470543 on Build GRCh38
rs781797738 Chr.9: 133470408-133470408 on Build GRCh38
rs71503202 Chr.9: 133470495-133470495 on Build GRCh38
rs45627341 Chr.9: 133470651-133470651 on Build GRCh38
rs956402660 Chr.9: 133470394-133470394 on Build GRCh38
rs1014565679 Chr.9: 133470766-133470766 on Build GRCh38
rs931828852 Chr.9: 133470705-133470705 on Build GRCh38
rs929420668 Chr.9: 133470641-133470641 on Build GRCh38
rs920769770 Chr.9: 133470640-133470640 on Build GRCh38
rs983626249 Chr.9: 133470765-133470765 on Build GRCh38
rs587696631 Chr.9: 133470731-133470731 on Build GRCh38
rs1024403593 Chr.9: 133470636-133470636 on Build GRCh38
rs907539098 Chr.9: 133470664-133470664 on Build GRCh38
rs781917540 Chr.9: 133470346-133470346 on Build GRCh38
rs1049871019 Chr.9: 133470583-133470583 on Build GRCh38
rs943600974 Chr.9: 133470597-133470597 on Build GRCh38
rs1055182167 Chr.9: 133470682-133470683 on Build GRCh38
rs587675227 Chr.9: 133470453-133470453 on Build GRCh38
rs587650684 Chr.9: 133470760-133470760 on Build GRCh38
rs975980955 Chr.9: 133470790-133470790 on Build GRCh38

View Full Product Details