Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136996433-136996937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC3
Entrez Gene ID
9022
Gene Name
chloride intracellular channel 3
Gene Aliases
-
UniGene
Hs.64746
Gene Chromosome Location
Chr.9: 136994608-136997929 on Build GRCh38
Transcripts
NM_004669.2 XM_017015281.1 XM_017015282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1461271 c.6G>A p.A2A Chr.9: 136996538-136996538 on Build GRCh38
7187460 c.22C>G p.L8V Chr.9: 136996522-136996522 on Build GRCh38
4518561 c.6_7GG>AA p.E3K Chr.9: 136996537-136996538 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs544292180 Chr.9: 136996780-136996780 on Build GRCh38
rs114743245 Chr.9: 136996760-136996760 on Build GRCh38
rs879685353 Chr.9: 136996882-136996882 on Build GRCh38
rs987444474 Chr.9: 136996794-136996794 on Build GRCh38
rs540154081 Chr.9: 136996804-136996804 on Build GRCh38
rs34871854 Chr.9: 136996692-136996692 on Build GRCh38
rs982822486 Chr.9: 136996612-136996612 on Build GRCh38
rs761767487 Chr.9: 136996900-136996900 on Build GRCh38
rs955680277 Chr.9: 136996783-136996783 on Build GRCh38
rs931637662 Chr.9: 136996558-136996558 on Build GRCh38
rs747533726 Chr.9: 136996665-136996665 on Build GRCh38
rs765039704 Chr.9: 136996623-136996623 on Build GRCh38
rs754185672 Chr.9: 136996495-136996495 on Build GRCh38
rs189446197 Chr.9: 136996588-136996588 on Build GRCh38
rs976666364 Chr.9: 136996815-136996815 on Build GRCh38
rs937427827 Chr.9: 136996870-136996870 on Build GRCh38
rs929973413 Chr.9: 136996827-136996827 on Build GRCh38
rs747585856 Chr.9: 136996463-136996463 on Build GRCh38
rs150822045 Chr.9: 136996538-136996538 on Build GRCh38
rs893664717 Chr.9: 136996698-136996698 on Build GRCh38
rs773299490 Chr.9: 136996568-136996568 on Build GRCh38
rs548837936 Chr.9: 136996606-136996606 on Build GRCh38
rs985289336 Chr.9: 136996571-136996571 on Build GRCh38
rs921492109 Chr.9: 136996639-136996639 on Build GRCh38
rs908834 Chr.9: 136996466-136996466 on Build GRCh38
rs914961770 Chr.9: 136996807-136996807 on Build GRCh38
rs969809330 Chr.9: 136996600-136996600 on Build GRCh38
rs2292922 Chr.9: 136996489-136996489 on Build GRCh38
rs1020455023 Chr.9: 136996717-136996717 on Build GRCh38
rs540501501 Chr.9: 136996557-136996557 on Build GRCh38
rs200825202 Chr.9: 136996490-136996490 on Build GRCh38
rs376163123 Chr.9: 136996481-136996481 on Build GRCh38
rs1010853895 Chr.9: 136996593-136996593 on Build GRCh38
rs1016944236 Chr.9: 136996757-136996757 on Build GRCh38
rs951639300 Chr.9: 136996476-136996476 on Build GRCh38
rs538007538 Chr.9: 136996714-136996714 on Build GRCh38
rs7855602 Chr.9: 136996746-136996746 on Build GRCh38
rs1003868899 Chr.9: 136996716-136996716 on Build GRCh38
rs775617666 Chr.9: 136996811-136996811 on Build GRCh38
rs766649532 Chr.9: 136996574-136996574 on Build GRCh38
rs541632884 Chr.9: 136996822-136996822 on Build GRCh38
rs182049213 Chr.9: 136996652-136996652 on Build GRCh38
rs950186920 Chr.9: 136996898-136996898 on Build GRCh38
rs749223050 Chr.9: 136996559-136996559 on Build GRCh38
rs554858688 Chr.9: 136996555-136996555 on Build GRCh38
rs962827758 Chr.9: 136996625-136996625 on Build GRCh38
rs745607787 Chr.9: 136996548-136996548 on Build GRCh38
rs568666867 Chr.9: 136996644-136996644 on Build GRCh38
rs765902448 Chr.9: 136996502-136996502 on Build GRCh38
rs909980735 Chr.9: 136996863-136996863 on Build GRCh38
rs939312970 Chr.9: 136996832-136996832 on Build GRCh38
rs1045397115 Chr.9: 136996709-136996709 on Build GRCh38
rs560754544 Chr.9: 136996795-136996795 on Build GRCh38
rs576814863 Chr.9: 136996676-136996676 on Build GRCh38
rs910076510 Chr.9: 136996614-136996614 on Build GRCh38
rs777315086 Chr.9: 136996524-136996524 on Build GRCh38
rs377313643 Chr.9: 136996467-136996467 on Build GRCh38
rs753242320 Chr.9: 136996510-136996510 on Build GRCh38
rs554464949 Chr.9: 136996677-136996677 on Build GRCh38
rs187425042 Chr.9: 136996847-136996847 on Build GRCh38
rs545936766 Chr.9: 136996728-136996728 on Build GRCh38
rs560740663 Chr.9: 136996485-136996485 on Build GRCh38
rs934962310 Chr.9: 136996796-136996796 on Build GRCh38
rs115105355 Chr.9: 136996569-136996569 on Build GRCh38
rs34406787 Chr.9: 136996718-136996719 on Build GRCh38
rs779621052 Chr.9: 136996539-136996539 on Build GRCh38
rs976082632 Chr.9: 136996637-136996637 on Build GRCh38
rs1035518138 Chr.9: 136996607-136996607 on Build GRCh38
rs772062896 Chr.9: 136996566-136996566 on Build GRCh38
rs911536846 Chr.9: 136996672-136996672 on Build GRCh38
rs1023530269 Chr.9: 136996594-136996594 on Build GRCh38
rs951505363 Chr.9: 136996611-136996611 on Build GRCh38
rs368911023 Chr.9: 136996537-136996537 on Build GRCh38
rs918586907 Chr.9: 136996634-136996634 on Build GRCh38
rs966358086 Chr.9: 136996740-136996740 on Build GRCh38
rs1044946247 Chr.9: 136996813-136996813 on Build GRCh38
rs757885849 Chr.9: 136996511-136996511 on Build GRCh38
rs776803504 Chr.9: 136996726-136996726 on Build GRCh38
rs748938459 Chr.9: 136996567-136996567 on Build GRCh38
rs752592603 Chr.9: 136996914-136996914 on Build GRCh38
rs538284493 Chr.9: 136996715-136996715 on Build GRCh38
rs199613987 Chr.9: 136996482-136996482 on Build GRCh38
rs201670284 Chr.9: 136996570-136996570 on Build GRCh38
rs531552797 Chr.9: 136996803-136996803 on Build GRCh38
rs552322271 Chr.9: 136996895-136996895 on Build GRCh38
rs890172058 Chr.9: 136996564-136996564 on Build GRCh38
rs532412254 Chr.9: 136996893-136996893 on Build GRCh38
rs560125561 Chr.9: 136996501-136996501 on Build GRCh38
rs572810523 Chr.9: 136996700-136996700 on Build GRCh38
rs746262982 Chr.9: 136996770-136996770 on Build GRCh38
rs769567122 Chr.9: 136996554-136996554 on Build GRCh38

View Full Product Details