Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 126326459-126326955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MVB12B
Entrez Gene ID
89853
Gene Name
multivesicular body subunit 12B
Gene Aliases
C9orf28, FAM125B
UniGene
Hs.162659
Gene Chromosome Location
Chr.9: 126326844-126507041 on Build GRCh38
Transcripts
NM_001011703.2 NM_033446.2 XM_017015275.1 XM_017015277.1 XM_017015278.1

SNP Detail

SNP ID Chromosome Location
rs945620772 Chr.9: 126326710-126326715 on Build GRCh38
rs1045575943 Chr.9: 126326785-126326785 on Build GRCh38
rs1015215326 Chr.9: 126326526-126326526 on Build GRCh38
rs1024532543 Chr.9: 126326498-126326498 on Build GRCh38
rs528926661 Chr.9: 126326821-126326823 on Build GRCh38
rs570708646 Chr.9: 126326589-126326589 on Build GRCh38
rs898196658 Chr.9: 126326831-126326831 on Build GRCh38
rs913853729 Chr.9: 126326879-126326879 on Build GRCh38
rs966866367 Chr.9: 126326895-126326895 on Build GRCh38
rs974970603 Chr.9: 126326745-126326766 on Build GRCh38
rs978367238 Chr.9: 126326900-126326900 on Build GRCh38
rs954380925 Chr.9: 126326670-126326670 on Build GRCh38
rs1034144030 Chr.9: 126326609-126326609 on Build GRCh38
rs1008360237 Chr.9: 126326862-126326862 on Build GRCh38
rs991987737 Chr.9: 126326820-126326820 on Build GRCh38
rs960571697 Chr.9: 126326772-126326772 on Build GRCh38
rs922079194 Chr.9: 126326767-126326790 on Build GRCh38
rs1012732630 Chr.9: 126326488-126326488 on Build GRCh38
rs1028383177 Chr.9: 126326843-126326843 on Build GRCh38
rs571725863 Chr.9: 126326824-126326824 on Build GRCh38
rs1019784076 Chr.9: 126326892-126326893 on Build GRCh38
rs912695783 Chr.9: 126326700-126326700 on Build GRCh38
rs906990632 Chr.9: 126326796-126326796 on Build GRCh38
rs953836201 Chr.9: 126326849-126326849 on Build GRCh38
rs907130297 Chr.9: 126326501-126326501 on Build GRCh38
rs148744220 Chr.9: 126326521-126326521 on Build GRCh38
rs539600336 Chr.9: 126326825-126326825 on Build GRCh38
rs936640823 Chr.9: 126326760-126326760 on Build GRCh38
rs916686634 Chr.9: 126326780-126326780 on Build GRCh38
rs949469926 Chr.9: 126326784-126326784 on Build GRCh38
rs978301998 Chr.9: 126326733-126326733 on Build GRCh38
rs906475018 Chr.9: 126326484-126326484 on Build GRCh38
rs987005035 Chr.9: 126326689-126326689 on Build GRCh38
rs551555296 Chr.9: 126326620-126326620 on Build GRCh38
rs995116253 Chr.9: 126326833-126326833 on Build GRCh38
rs191364691 Chr.9: 126326885-126326885 on Build GRCh38
rs1028060378 Chr.9: 126326650-126326650 on Build GRCh38
rs925505621 Chr.9: 126326925-126326925 on Build GRCh38
rs1055205886 Chr.9: 126326480-126326480 on Build GRCh38

View Full Product Details