Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127823978-127824502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENG
Entrez Gene ID
2022
Gene Name
endoglin
Gene Aliases
END, HHT1, ORW1
UniGene
Hs.76753
Gene Chromosome Location
Chr.9: 127815012-127854773 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1460146 c.1133C>T p.A378V Chr.9: 127824305-127824305 on Build GRCh38
5757313 c.1130T>C p.V377A Chr.9: 127824308-127824308 on Build GRCh38
5757314 c.1130T>C p.V377A Chr.9: 127824308-127824308 on Build GRCh38
6472936 c.1038C>A p.P346P Chr.9: 127824400-127824400 on Build GRCh38
4152075 c.1077C>G p.I359M Chr.9: 127824361-127824361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770665738 Chr.9: 127824294-127824294 on Build GRCh38
rs774429348 Chr.9: 127824414-127824414 on Build GRCh38
rs147017175 Chr.9: 127824344-127824344 on Build GRCh38
rs966677126 Chr.9: 127824140-127824140 on Build GRCh38
rs1033314945 Chr.9: 127824410-127824410 on Build GRCh38
rs890987118 Chr.9: 127824068-127824068 on Build GRCh38
rs766610172 Chr.9: 127824018-127824018 on Build GRCh38
rs543428700 Chr.9: 127824250-127824250 on Build GRCh38
rs757885743 Chr.9: 127824276-127824276 on Build GRCh38
rs959155421 Chr.9: 127824166-127824166 on Build GRCh38
rs190011129 Chr.9: 127824197-127824197 on Build GRCh38
rs761939575 Chr.9: 127824416-127824416 on Build GRCh38
rs765446986 Chr.9: 127824476-127824477 on Build GRCh38
rs997297796 Chr.9: 127824042-127824042 on Build GRCh38
rs778191568 Chr.9: 127824198-127824198 on Build GRCh38
rs780484184 Chr.9: 127824262-127824262 on Build GRCh38
rs986696330 Chr.9: 127824108-127824108 on Build GRCh38
rs367734769 Chr.9: 127824397-127824397 on Build GRCh38
rs992285787 Chr.9: 127824200-127824200 on Build GRCh38
rs949270423 Chr.9: 127824216-127824216 on Build GRCh38
rs751594882 Chr.9: 127824279-127824279 on Build GRCh38
rs574405265 Chr.9: 127824244-127824244 on Build GRCh38
rs769530262 Chr.9: 127824382-127824382 on Build GRCh38
rs777177134 Chr.9: 127824349-127824349 on Build GRCh38
rs561372233 Chr.9: 127824480-127824490 on Build GRCh38
rs780814647 Chr.9: 127824469-127824469 on Build GRCh38
rs761463410 Chr.9: 127824423-127824423 on Build GRCh38
rs1000620593 Chr.9: 127824231-127824231 on Build GRCh38
rs932659087 Chr.9: 127824341-127824341 on Build GRCh38
rs375427398 Chr.9: 127824346-127824346 on Build GRCh38
rs1050953073 Chr.9: 127824060-127824060 on Build GRCh38
rs909048513 Chr.9: 127824211-127824211 on Build GRCh38
rs917866100 Chr.9: 127824215-127824215 on Build GRCh38
rs775352287 Chr.9: 127824332-127824332 on Build GRCh38
rs560794600 Chr.9: 127824201-127824201 on Build GRCh38
rs746679633 Chr.9: 127824287-127824287 on Build GRCh38
rs868730895 Chr.9: 127824380-127824380 on Build GRCh38
rs112809381 Chr.9: 127824303-127824303 on Build GRCh38
rs766675335 Chr.9: 127824430-127824430 on Build GRCh38
rs949300932 Chr.9: 127824453-127824453 on Build GRCh38
rs971994795 Chr.9: 127824208-127824209 on Build GRCh38
rs1053567941 Chr.9: 127824258-127824258 on Build GRCh38
rs749797568 Chr.9: 127824269-127824269 on Build GRCh38
rs756890201 Chr.9: 127824452-127824452 on Build GRCh38
rs544624587 Chr.9: 127824288-127824288 on Build GRCh38
rs773463077 Chr.9: 127824420-127824420 on Build GRCh38
rs374734948 Chr.9: 127824473-127824473 on Build GRCh38
rs138190783 Chr.9: 127824371-127824371 on Build GRCh38
rs765715581 Chr.9: 127824451-127824451 on Build GRCh38
rs746450444 Chr.9: 127824355-127824355 on Build GRCh38
rs759964114 Chr.9: 127824442-127824442 on Build GRCh38
rs1800956 Chr.9: 127824342-127824342 on Build GRCh38
rs902157748 Chr.9: 127824227-127824227 on Build GRCh38
rs143054595 Chr.9: 127824305-127824305 on Build GRCh38
rs541137910 Chr.9: 127824174-127824174 on Build GRCh38
rs572458481 Chr.9: 127824107-127824107 on Build GRCh38
rs186991519 Chr.9: 127824103-127824103 on Build GRCh38
rs199531650 Chr.9: 127824338-127824338 on Build GRCh38
rs984798866 Chr.9: 127824209-127824209 on Build GRCh38
rs937895537 Chr.9: 127824213-127824213 on Build GRCh38
rs568777436 Chr.9: 127824077-127824077 on Build GRCh38
rs1038350300 Chr.9: 127824029-127824029 on Build GRCh38
rs3739817 Chr.9: 127824409-127824409 on Build GRCh38
rs863223540 Chr.9: 127824355-127824358 on Build GRCh38
rs150829904 Chr.9: 127824398-127824398 on Build GRCh38
rs919229180 Chr.9: 127824183-127824183 on Build GRCh38
rs747388144 Chr.9: 127824222-127824222 on Build GRCh38
rs142803546 Chr.9: 127824343-127824343 on Build GRCh38
rs769615362 Chr.9: 127824330-127824330 on Build GRCh38
rs200780733 Chr.9: 127824295-127824295 on Build GRCh38
rs138628226 Chr.9: 127824039-127824039 on Build GRCh38
rs941370375 Chr.9: 127824013-127824013 on Build GRCh38
rs1055364540 Chr.9: 127824214-127824214 on Build GRCh38
rs982934594 Chr.9: 127824267-127824267 on Build GRCh38
rs941569630 Chr.9: 127824275-127824275 on Build GRCh38
rs1033198063 Chr.9: 127824102-127824102 on Build GRCh38
rs777314748 Chr.9: 127824277-127824277 on Build GRCh38
rs548690138 Chr.9: 127824345-127824345 on Build GRCh38
rs976081447 Chr.9: 127824155-127824155 on Build GRCh38
rs201497772 Chr.9: 127824340-127824340 on Build GRCh38
rs772135786 Chr.9: 127824419-127824419 on Build GRCh38
rs371174838 Chr.9: 127824439-127824439 on Build GRCh38
rs773748110 Chr.9: 127824418-127824418 on Build GRCh38
rs372371435 Chr.9: 127824339-127824339 on Build GRCh38
rs909339898 Chr.9: 127824415-127824415 on Build GRCh38
rs1000589360 Chr.9: 127824079-127824079 on Build GRCh38
rs1017893891 Chr.9: 127824139-127824139 on Build GRCh38
rs36092484 Chr.9: 127824378-127824378 on Build GRCh38

View Full Product Details