Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127885239-127885746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK1
Entrez Gene ID
203
Gene Name
adenylate kinase 1
Gene Aliases
HTL-S-58j
UniGene
Hs.175473
Gene Chromosome Location
Chr.9: 127866480-127886749 on Build GRCh38
Transcripts
XM_017014427.1 XM_017014428.1 NM_001286999.1 NM_001287000.1 NM_001287001.1 NM_001287002.1 NM_001287003.1 NM_013443.4 NR_104629.1 XM_011518610.2 XM_011518611.2 XM_017014658.1 XM_017014659.1 XM_017014660.1 XM_017014661.1 XM_017014662.1 XM_017014663.1 XM_017014664.1 XM_017014665.1 XM_017014666.1 XM_017014667.1

Gene Symbol
ST6GALNAC6
Entrez Gene ID
30815
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
Gene Aliases
SIAT7-F, SIAT7F, ST6GALNACVI
UniGene
Hs.109672
Gene Chromosome Location
Chr.9: 127885321-127906725 on Build GRCh38
Transcripts
XM_017014427.1 XM_017014428.1 NM_001286999.1 NM_001287000.1 NM_001287001.1 NM_001287002.1 NM_001287003.1 NM_013443.4 NR_104629.1 XM_011518610.2 XM_011518611.2 XM_017014658.1 XM_017014659.1 XM_017014660.1 XM_017014661.1 XM_017014662.1 XM_017014663.1 XM_017014664.1 XM_017014665.1 XM_017014666.1 XM_017014667.1

SNP Detail

SNP ID Chromosome Location
rs1005566211 Chr.9: 127885498-127885498 on Build GRCh38
rs941454526 Chr.9: 127885718-127885718 on Build GRCh38
rs967478234 Chr.9: 127885578-127885578 on Build GRCh38
rs1024393405 Chr.9: 127885268-127885268 on Build GRCh38
rs377183917 Chr.9: 127885638-127885638 on Build GRCh38
rs3178243 Chr.9: 127885321-127885321 on Build GRCh38
rs1055803394 Chr.9: 127885719-127885719 on Build GRCh38
rs990117447 Chr.9: 127885505-127885505 on Build GRCh38
rs775600121 Chr.9: 127885391-127885391 on Build GRCh38
rs567158974 Chr.9: 127885610-127885610 on Build GRCh38
rs936845384 Chr.9: 127885305-127885305 on Build GRCh38
rs1019155407 Chr.9: 127885433-127885433 on Build GRCh38
rs977609665 Chr.9: 127885296-127885296 on Build GRCh38
rs149245204 Chr.9: 127885599-127885599 on Build GRCh38
rs898421976 Chr.9: 127885630-127885630 on Build GRCh38
rs2031696 Chr.9: 127885304-127885304 on Build GRCh38
rs1014166067 Chr.9: 127885552-127885552 on Build GRCh38
rs745782924 Chr.9: 127885261-127885261 on Build GRCh38
rs985843398 Chr.9: 127885614-127885614 on Build GRCh38
rs978098706 Chr.9: 127885457-127885457 on Build GRCh38
rs1049464053 Chr.9: 127885636-127885636 on Build GRCh38
rs989665252 Chr.9: 127885310-127885310 on Build GRCh38
rs768633027 Chr.9: 127885546-127885546 on Build GRCh38
rs531222525 Chr.9: 127885273-127885273 on Build GRCh38
rs907028554 Chr.9: 127885266-127885266 on Build GRCh38
rs1036933612 Chr.9: 127885295-127885295 on Build GRCh38
rs764218249 Chr.9: 127885259-127885259 on Build GRCh38
rs902235639 Chr.9: 127885714-127885714 on Build GRCh38
rs922865503 Chr.9: 127885641-127885641 on Build GRCh38
rs1053520 Chr.9: 127885435-127885435 on Build GRCh38
rs939888632 Chr.9: 127885601-127885601 on Build GRCh38
rs774617297 Chr.9: 127885564-127885564 on Build GRCh38
rs909854879 Chr.9: 127885709-127885709 on Build GRCh38
rs981478693 Chr.9: 127885571-127885571 on Build GRCh38
rs564641365 Chr.9: 127885368-127885368 on Build GRCh38
rs138414464 Chr.9: 127885442-127885442 on Build GRCh38
rs1027961305 Chr.9: 127885323-127885323 on Build GRCh38
rs888559552 Chr.9: 127885494-127885494 on Build GRCh38
rs1047071737 Chr.9: 127885692-127885692 on Build GRCh38
rs957122083 Chr.9: 127885608-127885608 on Build GRCh38
rs540087777 Chr.9: 127885265-127885265 on Build GRCh38
rs964202303 Chr.9: 127885631-127885631 on Build GRCh38
rs967068182 Chr.9: 127885274-127885274 on Build GRCh38
rs998513265 Chr.9: 127885595-127885595 on Build GRCh38
rs533225502 Chr.9: 127885613-127885613 on Build GRCh38
rs911364526 Chr.9: 127885315-127885315 on Build GRCh38
rs763025020 Chr.9: 127885411-127885411 on Build GRCh38
rs924906220 Chr.9: 127885474-127885474 on Build GRCh38
rs915873223 Chr.9: 127885510-127885510 on Build GRCh38
rs942935801 Chr.9: 127885322-127885322 on Build GRCh38
rs1055563956 Chr.9: 127885509-127885509 on Build GRCh38
rs1041257792 Chr.9: 127885327-127885327 on Build GRCh38
rs1019339883 Chr.9: 127885654-127885654 on Build GRCh38
rs911393828 Chr.9: 127885623-127885623 on Build GRCh38
rs111806373 Chr.9: 127885320-127885320 on Build GRCh38
rs930198489 Chr.9: 127885646-127885646 on Build GRCh38

View Full Product Details