Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50294776-50295288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB3
Entrez Gene ID
85417
Gene Name
cyclin B3
Gene Aliases
-
UniGene
Hs.130310
Gene Chromosome Location
Chr.X: 50202713-50351914 on Build GRCh38
Transcripts
NM_033031.2 NM_033670.2 XM_017029913.1 XM_017029914.1 XM_017029915.1 XM_017029916.1 XM_017029917.1 XM_017029918.1

SNP Detail

SNP ID Chromosome Location
rs781905774 Chr.X: 50294981-50294981 on Build GRCh38
rs190650938 Chr.X: 50295004-50295004 on Build GRCh38
rs782775027 Chr.X: 50294980-50294980 on Build GRCh38
rs782768945 Chr.X: 50295018-50295018 on Build GRCh38
rs1028411624 Chr.X: 50294969-50294969 on Build GRCh38
rs781817931 Chr.X: 50294879-50294879 on Build GRCh38
rs913965099 Chr.X: 50295100-50295100 on Build GRCh38
rs782147068 Chr.X: 50294887-50294887 on Build GRCh38
rs141684318 Chr.X: 50294971-50294971 on Build GRCh38
rs782428089 Chr.X: 50294949-50294949 on Build GRCh38
rs782787092 Chr.X: 50294836-50294836 on Build GRCh38
rs782612336 Chr.X: 50295174-50295174 on Build GRCh38
rs782683699 Chr.X: 50294834-50294834 on Build GRCh38
rs782440652 Chr.X: 50295166-50295166 on Build GRCh38
rs782614811 Chr.X: 50294871-50294871 on Build GRCh38
rs200248441 Chr.X: 50295121-50295121 on Build GRCh38
rs782576907 Chr.X: 50294899-50294899 on Build GRCh38
rs782035784 Chr.X: 50294849-50294850 on Build GRCh38
rs781809453 Chr.X: 50294837-50294837 on Build GRCh38
rs781843350 Chr.X: 50295054-50295054 on Build GRCh38
rs925769011 Chr.X: 50295072-50295072 on Build GRCh38
rs980018055 Chr.X: 50295060-50295060 on Build GRCh38
rs140921996 Chr.X: 50295086-50295086 on Build GRCh38
rs782766037 Chr.X: 50294812-50294812 on Build GRCh38
rs200435913 Chr.X: 50294977-50294977 on Build GRCh38
rs782493836 Chr.X: 50295014-50295014 on Build GRCh38
rs782008142 Chr.X: 50294951-50294951 on Build GRCh38
rs782145889 Chr.X: 50294835-50294836 on Build GRCh38
rs151272775 Chr.X: 50294948-50294948 on Build GRCh38
rs782607844 Chr.X: 50295017-50295017 on Build GRCh38
rs782413297 Chr.X: 50294850-50294850 on Build GRCh38
rs935848925 Chr.X: 50295078-50295078 on Build GRCh38
rs782327723 Chr.X: 50294898-50294898 on Build GRCh38
rs782276010 Chr.X: 50294896-50294896 on Build GRCh38
rs782728145 Chr.X: 50295147-50295148 on Build GRCh38
rs142293112 Chr.X: 50294943-50294943 on Build GRCh38
rs782478097 Chr.X: 50295059-50295059 on Build GRCh38
rs782059556 Chr.X: 50294967-50294967 on Build GRCh38
rs901173922 Chr.X: 50295183-50295183 on Build GRCh38
rs961314720 Chr.X: 50295005-50295005 on Build GRCh38
rs782677573 Chr.X: 50294888-50294888 on Build GRCh38
rs145017627 Chr.X: 50295261-50295261 on Build GRCh38
rs782237362 Chr.X: 50294905-50294905 on Build GRCh38
rs782438945 Chr.X: 50294858-50294858 on Build GRCh38
rs782593661 Chr.X: 50295009-50295009 on Build GRCh38
rs901399081 Chr.X: 50294934-50294934 on Build GRCh38
rs201311546 Chr.X: 50294826-50294826 on Build GRCh38
rs181833827 Chr.X: 50295211-50295211 on Build GRCh38
rs782139898 Chr.X: 50295000-50295001 on Build GRCh38
rs185845142 Chr.X: 50294913-50294913 on Build GRCh38
rs782802301 Chr.X: 50295123-50295123 on Build GRCh38
rs1024403213 Chr.X: 50295036-50295036 on Build GRCh38

View Full Product Details