Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231353111-231353606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRTN
Entrez Gene ID
83932
Gene Name
SprT-like N-terminal domain
Gene Aliases
C1orf124, DVC1, PRO4323, spartan
UniGene
Hs.554892
Gene Chromosome Location
Chr.1: 231337936-231355029 on Build GRCh38
Transcripts
NM_001010984.3 NM_001261462.2 NM_032018.6 XM_006711818.2 XM_011544289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
905492 c.1223G>A p.R408Q Chr.1: 231353114-231353114 on Build GRCh38
1246642 c.1300C>T p.P434S Chr.1: 231353191-231353191 on Build GRCh38
905493 c.1262T>G p.F421C Chr.1: 231353153-231353153 on Build GRCh38
464140 c.1446A>G p.K482K Chr.1: 231353337-231353337 on Build GRCh38
1998455 c.1256_1257insT p.I422fs*11 Chr.1: 231353147-231353148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965510807 Chr.1: 231353573-231353573 on Build GRCh38
rs755606181 Chr.1: 231353381-231353381 on Build GRCh38
rs764191857 Chr.1: 231353342-231353342 on Build GRCh38
rs774016148 Chr.1: 231353509-231353509 on Build GRCh38
rs772211381 Chr.1: 231353315-231353315 on Build GRCh38
rs778677371 Chr.1: 231353409-231353409 on Build GRCh38
rs200841470 Chr.1: 231353192-231353192 on Build GRCh38
rs776875259 Chr.1: 231353346-231353346 on Build GRCh38
rs540856655 Chr.1: 231353225-231353225 on Build GRCh38
rs373368337 Chr.1: 231353320-231353320 on Build GRCh38
rs373387621 Chr.1: 231353387-231353387 on Build GRCh38
rs767531110 Chr.1: 231353208-231353208 on Build GRCh38
rs754129906 Chr.1: 231353223-231353223 on Build GRCh38
rs769305407 Chr.1: 231353254-231353254 on Build GRCh38
rs143333462 Chr.1: 231353511-231353511 on Build GRCh38
rs779664072 Chr.1: 231353160-231353160 on Build GRCh38
rs1018464567 Chr.1: 231353569-231353569 on Build GRCh38
rs764998663 Chr.1: 231353136-231353137 on Build GRCh38
rs994230295 Chr.1: 231353544-231353545 on Build GRCh38
rs149065971 Chr.1: 231353351-231353351 on Build GRCh38
rs762108108 Chr.1: 231353347-231353347 on Build GRCh38
rs368491865 Chr.1: 231353227-231353227 on Build GRCh38
rs192060983 Chr.1: 231353335-231353335 on Build GRCh38
rs774288244 Chr.1: 231353307-231353307 on Build GRCh38
rs752219141 Chr.1: 231353371-231353371 on Build GRCh38
rs764577876 Chr.1: 231353215-231353215 on Build GRCh38
rs377221472 Chr.1: 231353309-231353309 on Build GRCh38
rs752925605 Chr.1: 231353209-231353209 on Build GRCh38
rs756487057 Chr.1: 231353147-231353147 on Build GRCh38
rs1042317699 Chr.1: 231353435-231353435 on Build GRCh38
rs888452527 Chr.1: 231353576-231353576 on Build GRCh38
rs960999314 Chr.1: 231353186-231353186 on Build GRCh38
rs746084136 Chr.1: 231353452-231353452 on Build GRCh38
rs377714192 Chr.1: 231353506-231353506 on Build GRCh38
rs745867595 Chr.1: 231353412-231353412 on Build GRCh38
rs754537278 Chr.1: 231353235-231353235 on Build GRCh38
rs139764980 Chr.1: 231353176-231353176 on Build GRCh38
rs1007086052 Chr.1: 231353565-231353565 on Build GRCh38
rs765569992 Chr.1: 231353350-231353350 on Build GRCh38
rs550457013 Chr.1: 231353425-231353425 on Build GRCh38
rs757009337 Chr.1: 231353395-231353395 on Build GRCh38
rs111353550 Chr.1: 231353375-231353375 on Build GRCh38
rs62617126 Chr.1: 231353206-231353206 on Build GRCh38
rs567833527 Chr.1: 231353365-231353365 on Build GRCh38
rs115782522 Chr.1: 231353530-231353530 on Build GRCh38
rs906647927 Chr.1: 231353188-231353188 on Build GRCh38
rs747733787 Chr.1: 231353167-231353167 on Build GRCh38
rs113842682 Chr.1: 231353559-231353559 on Build GRCh38
rs375357639 Chr.1: 231353193-231353193 on Build GRCh38
rs772869072 Chr.1: 231353189-231353189 on Build GRCh38
rs760886636 Chr.1: 231353211-231353211 on Build GRCh38
rs928105691 Chr.1: 231353523-231353523 on Build GRCh38
rs914192016 Chr.1: 231353507-231353507 on Build GRCh38
rs372548439 Chr.1: 231353301-231353301 on Build GRCh38
rs777937373 Chr.1: 231353149-231353149 on Build GRCh38
rs761254645 Chr.1: 231353517-231353517 on Build GRCh38
rs748910029 Chr.1: 231353393-231353393 on Build GRCh38
rs754439752 Chr.1: 231353155-231353155 on Build GRCh38
rs567805129 Chr.1: 231353147-231353148 on Build GRCh38
rs1054018568 Chr.1: 231353141-231353141 on Build GRCh38
rs985872267 Chr.1: 231353410-231353410 on Build GRCh38
rs200987948 Chr.1: 231353163-231353163 on Build GRCh38
rs142369937 Chr.1: 231353207-231353207 on Build GRCh38
rs775537322 Chr.1: 231353322-231353322 on Build GRCh38
rs936558300 Chr.1: 231353481-231353481 on Build GRCh38
rs560897223 Chr.1: 231353140-231353140 on Build GRCh38
rs957691146 Chr.1: 231353462-231353462 on Build GRCh38
rs188253824 Chr.1: 231353284-231353284 on Build GRCh38
rs536855918 Chr.1: 231353366-231353366 on Build GRCh38
rs200999774 Chr.1: 231353162-231353162 on Build GRCh38
rs113002283 Chr.1: 231353303-231353303 on Build GRCh38
rs531550374 Chr.1: 231353226-231353226 on Build GRCh38
rs914076588 Chr.1: 231353146-231353146 on Build GRCh38
rs780845237 Chr.1: 231353244-231353244 on Build GRCh38
rs369698419 Chr.1: 231353250-231353250 on Build GRCh38
rs757693366 Chr.1: 231353159-231353159 on Build GRCh38
rs954645715 Chr.1: 231353358-231353358 on Build GRCh38
rs1055174440 Chr.1: 231353552-231353552 on Build GRCh38

View Full Product Details