Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66021438-66021956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER1
Entrez Gene ID
117144
Gene Name
cation channel sperm associated 1
Gene Aliases
CATSPER, SPGF7
UniGene
Hs.189105
Gene Chromosome Location
Chr.11: 66016752-66026518 on Build GRCh38
Transcripts
NM_053054.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
337288 c.1627C>A p.R543R Chr.11: 66021560-66021560 on Build GRCh38
1356127 c.1657G>T p.A553S Chr.11: 66021530-66021530 on Build GRCh38
930610 c.1486C>T p.L496L Chr.11: 66021823-66021823 on Build GRCh38
4756652 c.1679T>C p.L560P Chr.11: 66021508-66021508 on Build GRCh38
3416135 c.1602C>T p.F534F Chr.11: 66021585-66021585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758208893 Chr.11: 66021676-66021677 on Build GRCh38
rs759188008 Chr.11: 66021901-66021901 on Build GRCh38
rs930999199 Chr.11: 66021928-66021928 on Build GRCh38
rs60581004 Chr.11: 66021605-66021605 on Build GRCh38
rs763259517 Chr.11: 66021483-66021483 on Build GRCh38
rs767160886 Chr.11: 66021579-66021579 on Build GRCh38
rs777461773 Chr.11: 66021735-66021736 on Build GRCh38
rs764462036 Chr.11: 66021491-66021491 on Build GRCh38
rs770529266 Chr.11: 66021789-66021789 on Build GRCh38
rs764930175 Chr.11: 66021904-66021904 on Build GRCh38
rs776993682 Chr.11: 66021764-66021764 on Build GRCh38
rs770141425 Chr.11: 66021462-66021462 on Build GRCh38
rs894052330 Chr.11: 66021918-66021918 on Build GRCh38
rs767571845 Chr.11: 66021592-66021592 on Build GRCh38
rs931091793 Chr.11: 66021674-66021674 on Build GRCh38
rs550366747 Chr.11: 66021477-66021477 on Build GRCh38
rs1783562 Chr.11: 66021536-66021536 on Build GRCh38
rs745555831 Chr.11: 66021663-66021663 on Build GRCh38
rs200868978 Chr.11: 66021753-66021753 on Build GRCh38
rs928590431 Chr.11: 66021620-66021620 on Build GRCh38
rs769374210 Chr.11: 66021543-66021543 on Build GRCh38
rs769189808 Chr.11: 66021561-66021561 on Build GRCh38
rs766140407 Chr.11: 66021896-66021896 on Build GRCh38
rs745360172 Chr.11: 66021892-66021893 on Build GRCh38
rs777682212 Chr.11: 66021518-66021518 on Build GRCh38
rs747388246 Chr.11: 66021757-66021757 on Build GRCh38
rs757076081 Chr.11: 66021655-66021655 on Build GRCh38
rs774843895 Chr.11: 66021675-66021675 on Build GRCh38
rs368032844 Chr.11: 66021479-66021479 on Build GRCh38
rs770016571 Chr.11: 66021790-66021790 on Build GRCh38
rs774913580 Chr.11: 66021928-66021929 on Build GRCh38
rs758402955 Chr.11: 66021651-66021651 on Build GRCh38
rs765046118 Chr.11: 66021514-66021514 on Build GRCh38
rs764182570 Chr.11: 66021653-66021653 on Build GRCh38
rs373237398 Chr.11: 66021914-66021914 on Build GRCh38
rs760622742 Chr.11: 66021630-66021630 on Build GRCh38
rs997724274 Chr.11: 66021670-66021670 on Build GRCh38
rs773502579 Chr.11: 66021683-66021683 on Build GRCh38
rs746865417 Chr.11: 66021532-66021532 on Build GRCh38
rs776835270 Chr.11: 66021691-66021691 on Build GRCh38
rs60415681 Chr.11: 66021727-66021727 on Build GRCh38
rs770179439 Chr.11: 66021872-66021872 on Build GRCh38
rs780701695 Chr.11: 66021836-66021836 on Build GRCh38
rs567657527 Chr.11: 66021654-66021654 on Build GRCh38
rs74484098 Chr.11: 66021795-66021795 on Build GRCh38
rs371777676 Chr.11: 66021824-66021824 on Build GRCh38
rs752479166 Chr.11: 66021636-66021638 on Build GRCh38
rs901134861 Chr.11: 66021712-66021712 on Build GRCh38
rs375690376 Chr.11: 66021513-66021513 on Build GRCh38
rs535618233 Chr.11: 66021804-66021804 on Build GRCh38
rs760923006 Chr.11: 66021688-66021688 on Build GRCh38
rs939887832 Chr.11: 66021704-66021704 on Build GRCh38
rs757135673 Chr.11: 66021576-66021576 on Build GRCh38
rs748670632 Chr.11: 66021752-66021752 on Build GRCh38
rs778692528 Chr.11: 66021876-66021876 on Build GRCh38
rs748472911 Chr.11: 66021677-66021677 on Build GRCh38
rs200193011 Chr.11: 66021754-66021754 on Build GRCh38
rs750177081 Chr.11: 66021809-66021809 on Build GRCh38
rs754515348 Chr.11: 66021745-66021745 on Build GRCh38
rs200047406 Chr.11: 66021672-66021672 on Build GRCh38
rs886048520 Chr.11: 66021767-66021767 on Build GRCh38
rs75366838 Chr.11: 66021842-66021842 on Build GRCh38
rs779562251 Chr.11: 66021741-66021741 on Build GRCh38
rs779506893 Chr.11: 66021671-66021671 on Build GRCh38
rs1032573943 Chr.11: 66021710-66021710 on Build GRCh38
rs540311484 Chr.11: 66021690-66021690 on Build GRCh38
rs778890873 Chr.11: 66021592-66021592 on Build GRCh38
rs762182903 Chr.11: 66021584-66021584 on Build GRCh38
rs756226578 Chr.11: 66021506-66021506 on Build GRCh38
rs34114713 Chr.11: 66021666-66021666 on Build GRCh38
rs1055684410 Chr.11: 66021632-66021632 on Build GRCh38
rs184633777 Chr.11: 66021621-66021621 on Build GRCh38
rs201190307 Chr.11: 66021923-66021923 on Build GRCh38
rs886845418 Chr.11: 66021697-66021697 on Build GRCh38
rs754679017 Chr.11: 66021836-66021836 on Build GRCh38
rs202004405 Chr.11: 66021864-66021864 on Build GRCh38
rs775633705 Chr.11: 66021879-66021879 on Build GRCh38
rs746678520 Chr.11: 66021768-66021788 on Build GRCh38
rs751966872 Chr.11: 66021496-66021496 on Build GRCh38
rs773515324 Chr.11: 66021616-66021616 on Build GRCh38
rs751728743 Chr.11: 66021505-66021505 on Build GRCh38
rs371625829 Chr.11: 66021717-66021717 on Build GRCh38
rs759644610 Chr.11: 66021769-66021769 on Build GRCh38
rs771981376 Chr.11: 66021887-66021887 on Build GRCh38
rs143644621 Chr.11: 66021808-66021808 on Build GRCh38
rs988204643 Chr.11: 66021469-66021469 on Build GRCh38
rs188321248 Chr.11: 66021758-66021758 on Build GRCh38
rs774688128 Chr.11: 66021580-66021580 on Build GRCh38
rs769082865 Chr.11: 66021893-66021893 on Build GRCh38
rs760393532 Chr.11: 66021818-66021818 on Build GRCh38
rs747328548 Chr.11: 66021862-66021862 on Build GRCh38
rs775808039 Chr.11: 66021559-66021559 on Build GRCh38
rs201337851 Chr.11: 66021716-66021716 on Build GRCh38
rs147398525 Chr.11: 66021649-66021649 on Build GRCh38
rs749769569 Chr.11: 66021560-66021560 on Build GRCh38
rs200714710 Chr.11: 66021515-66021515 on Build GRCh38
rs375866031 Chr.11: 66021740-66021740 on Build GRCh38
rs756955557 Chr.11: 66021533-66021533 on Build GRCh38
rs142758999 Chr.11: 66021852-66021852 on Build GRCh38
rs772512635 Chr.11: 66021682-66021682 on Build GRCh38
rs760470378 Chr.11: 66021894-66021894 on Build GRCh38
rs376008588 Chr.11: 66021828-66021828 on Build GRCh38
rs992124316 Chr.11: 66021859-66021859 on Build GRCh38
rs766579277 Chr.11: 66021689-66021689 on Build GRCh38
rs369774821 Chr.11: 66021910-66021910 on Build GRCh38
rs745309713 Chr.11: 66021537-66021537 on Build GRCh38
rs941710321 Chr.11: 66021799-66021799 on Build GRCh38
rs762021038 Chr.11: 66021498-66021498 on Build GRCh38
rs866031033 Chr.11: 66021711-66021711 on Build GRCh38
rs762999530 Chr.11: 66021884-66021884 on Build GRCh38
rs988070450 Chr.11: 66021713-66021713 on Build GRCh38
rs34270689 Chr.11: 66021474-66021474 on Build GRCh38
rs886048521 Chr.11: 66021865-66021865 on Build GRCh38
rs766461275 Chr.11: 66021642-66021642 on Build GRCh38
rs746065311 Chr.11: 66021868-66021868 on Build GRCh38
rs778154291 Chr.11: 66021854-66021854 on Build GRCh38

View Full Product Details