Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137437804-137438305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD8
Entrez Gene ID
377841
Gene Name
ectonucleoside triphosphate diphosphohydrolase 8
Gene Aliases
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492
UniGene
Hs.512562
Gene Chromosome Location
Chr.9: 137434364-137441710 on Build GRCh38
Transcripts
NM_001033113.1 NM_198585.2 XM_011518665.2 XM_011518666.2 XM_011518668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1675003 c.62G>A p.G21D Chr.9: 137438224-137438224 on Build GRCh38
4762047 c.14G>A p.R5Q Chr.9: 137438272-137438272 on Build GRCh38
6473048 c.127-1G>A p.? Chr.9: 137438085-137438085 on Build GRCh38
4762046 c.14G>A p.R5Q Chr.9: 137438272-137438272 on Build GRCh38
6497755 c.215T>C p.V72A Chr.9: 137437996-137437996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551375296 Chr.9: 137438045-137438045 on Build GRCh38
rs368175211 Chr.9: 137438133-137438133 on Build GRCh38
rs757130003 Chr.9: 137438231-137438239 on Build GRCh38
rs769601323 Chr.9: 137438281-137438281 on Build GRCh38
rs776380909 Chr.9: 137438198-137438198 on Build GRCh38
rs142343085 Chr.9: 137438059-137438059 on Build GRCh38
rs546107466 Chr.9: 137438093-137438093 on Build GRCh38
rs764348127 Chr.9: 137438128-137438128 on Build GRCh38
rs761687099 Chr.9: 137438171-137438171 on Build GRCh38
rs144868718 Chr.9: 137438146-137438146 on Build GRCh38
rs758015896 Chr.9: 137437952-137437952 on Build GRCh38
rs754400533 Chr.9: 137438222-137438222 on Build GRCh38
rs773812361 Chr.9: 137437994-137437994 on Build GRCh38
rs565797700 Chr.9: 137438284-137438284 on Build GRCh38
rs777105208 Chr.9: 137438156-137438156 on Build GRCh38
rs745854376 Chr.9: 137438274-137438274 on Build GRCh38
rs995736773 Chr.9: 137437842-137437842 on Build GRCh38
rs1018298872 Chr.9: 137437845-137437845 on Build GRCh38
rs1053974096 Chr.9: 137438002-137438002 on Build GRCh38
rs201964108 Chr.9: 137438010-137438010 on Build GRCh38
rs200317835 Chr.9: 137437937-137437937 on Build GRCh38
rs753022448 Chr.9: 137437933-137437933 on Build GRCh38
rs6606582 Chr.9: 137438026-137438026 on Build GRCh38
rs374384192 Chr.9: 137438149-137438149 on Build GRCh38
rs751603146 Chr.9: 137438025-137438025 on Build GRCh38
rs970335998 Chr.9: 137438203-137438203 on Build GRCh38
rs551148760 Chr.9: 137438119-137438119 on Build GRCh38
rs1042293608 Chr.9: 137438154-137438154 on Build GRCh38
rs746536485 Chr.9: 137438174-137438174 on Build GRCh38
rs183434736 Chr.9: 137437964-137437964 on Build GRCh38
rs61730140 Chr.9: 137438050-137438050 on Build GRCh38
rs777491769 Chr.9: 137438252-137438252 on Build GRCh38
rs989106801 Chr.9: 137437915-137437915 on Build GRCh38
rs767814224 Chr.9: 137438207-137438207 on Build GRCh38
rs202087483 Chr.9: 137438158-137438158 on Build GRCh38
rs200546494 Chr.9: 137437943-137437943 on Build GRCh38
rs748989905 Chr.9: 137437981-137437981 on Build GRCh38
rs750911448 Chr.9: 137438134-137438134 on Build GRCh38
rs761981355 Chr.9: 137438206-137438206 on Build GRCh38
rs755313497 Chr.9: 137438226-137438226 on Build GRCh38
rs375173244 Chr.9: 137438112-137438112 on Build GRCh38
rs1006902581 Chr.9: 137437850-137437850 on Build GRCh38
rs768124732 Chr.9: 137437989-137437989 on Build GRCh38
rs935456022 Chr.9: 137437990-137437990 on Build GRCh38
rs141081294 Chr.9: 137437823-137437823 on Build GRCh38
rs771021299 Chr.9: 137438102-137438102 on Build GRCh38
rs145017191 Chr.9: 137438022-137438022 on Build GRCh38
rs563384871 Chr.9: 137438004-137438004 on Build GRCh38
rs371503672 Chr.9: 137437925-137437925 on Build GRCh38
rs368312647 Chr.9: 137437955-137437955 on Build GRCh38
rs747800597 Chr.9: 137438152-137438152 on Build GRCh38
rs111636573 Chr.9: 137438272-137438272 on Build GRCh38
rs757936735 Chr.9: 137438240-137438240 on Build GRCh38
rs969382995 Chr.9: 137438136-137438136 on Build GRCh38
rs765149820 Chr.9: 137438234-137438234 on Build GRCh38
rs777083097 Chr.9: 137438145-137438146 on Build GRCh38
rs531135716 Chr.9: 137438064-137438064 on Build GRCh38
rs746980982 Chr.9: 137438183-137438185 on Build GRCh38
rs760501158 Chr.9: 137438006-137438006 on Build GRCh38
rs574630684 Chr.9: 137438075-137438075 on Build GRCh38
rs754566549 Chr.9: 137438057-137438057 on Build GRCh38
rs750951232 Chr.9: 137438040-137438040 on Build GRCh38
rs760747546 Chr.9: 137438218-137438218 on Build GRCh38
rs746181110 Chr.9: 137437952-137437952 on Build GRCh38
rs773118015 Chr.9: 137438217-137438217 on Build GRCh38
rs758525518 Chr.9: 137437934-137437934 on Build GRCh38
rs963278489 Chr.9: 137437923-137437923 on Build GRCh38
rs950642154 Chr.9: 137437857-137437857 on Build GRCh38
rs142877518 Chr.9: 137438027-137438027 on Build GRCh38
rs751049999 Chr.9: 137438266-137438266 on Build GRCh38
rs927411941 Chr.9: 137437950-137437950 on Build GRCh38
rs374891120 Chr.9: 137437896-137437896 on Build GRCh38
rs774507616 Chr.9: 137438129-137438129 on Build GRCh38
rs113099702 Chr.9: 137438076-137438076 on Build GRCh38
rs1027108067 Chr.9: 137437847-137437847 on Build GRCh38
rs368524965 Chr.9: 137438155-137438155 on Build GRCh38
rs188781909 Chr.9: 137437831-137437831 on Build GRCh38
rs191126792 Chr.9: 137437895-137437895 on Build GRCh38
rs909099240 Chr.9: 137437904-137437904 on Build GRCh38
rs766895529 Chr.9: 137438144-137438144 on Build GRCh38
rs937756061 Chr.9: 137437974-137437974 on Build GRCh38
rs539854992 Chr.9: 137438242-137438242 on Build GRCh38
rs559799144 Chr.9: 137438023-137438023 on Build GRCh38
rs868248458 Chr.9: 137438065-137438065 on Build GRCh38
rs897013265 Chr.9: 137437834-137437834 on Build GRCh38
rs765558030 Chr.9: 137438235-137438235 on Build GRCh38
rs543970929 Chr.9: 137437853-137437853 on Build GRCh38
rs1013521618 Chr.9: 137437827-137437827 on Build GRCh38
rs200099005 Chr.9: 137437944-137437944 on Build GRCh38
rs781722487 Chr.9: 137438029-137438029 on Build GRCh38
rs865955515 Chr.9: 137438017-137438023 on Build GRCh38
rs199873802 Chr.9: 137438049-137438049 on Build GRCh38
rs771539442 Chr.9: 137438068-137438068 on Build GRCh38
rs755399830 Chr.9: 137438147-137438147 on Build GRCh38
rs376869050 Chr.9: 137437924-137437924 on Build GRCh38
rs769538473 Chr.9: 137438193-137438193 on Build GRCh38
rs781266790 Chr.9: 137438273-137438273 on Build GRCh38
rs756575058 Chr.9: 137438138-137438138 on Build GRCh38
rs564570975 Chr.9: 137437890-137437901 on Build GRCh38
rs371065621 Chr.9: 137438005-137438005 on Build GRCh38
rs770937585 Chr.9: 137438183-137438183 on Build GRCh38
rs894063085 Chr.9: 137437832-137437832 on Build GRCh38
rs1019645545 Chr.9: 137438090-137438090 on Build GRCh38
rs753133596 Chr.9: 137438236-137438236 on Build GRCh38
rs954138556 Chr.9: 137437849-137437849 on Build GRCh38
rs770743998 Chr.9: 137438184-137438184 on Build GRCh38
rs778036942 Chr.9: 137437935-137437935 on Build GRCh38
rs370063158 Chr.9: 137438121-137438121 on Build GRCh38
rs779668900 Chr.9: 137437978-137437978 on Build GRCh38

View Full Product Details