Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130622662-130623195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUBP3
Entrez Gene ID
8939
Gene Name
far upstream element binding protein 3
Gene Aliases
FBP3
UniGene
Hs.673029
Gene Chromosome Location
Chr.9: 130579567-130638352 on Build GRCh38
Transcripts
NM_003934.1 XM_005272232.1 XM_006717313.2 XM_011519172.2 XM_011519173.2 XM_011519174.2 XM_017015269.1 XM_017015270.1 XM_017015271.1 XM_017015272.1 XM_017015273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6472979 c.610G>T p.A204S Chr.9: 130622726-130622726 on Build GRCh38
3904787 c.871C>T p.P291S Chr.9: 130622807-130622807 on Build GRCh38
6472978 c.790G>T p.A264S Chr.9: 130622726-130622726 on Build GRCh38
7083965 c.596C>A p.S199Y Chr.9: 130622712-130622712 on Build GRCh38
6845814 c.874+1G>A p.? Chr.9: 130622811-130622811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988732233 Chr.9: 130622958-130622958 on Build GRCh38
rs551909349 Chr.9: 130622917-130622917 on Build GRCh38
rs779526523 Chr.9: 130622843-130622843 on Build GRCh38
rs578074951 Chr.9: 130622967-130622967 on Build GRCh38
rs936733285 Chr.9: 130623152-130623152 on Build GRCh38
rs753657849 Chr.9: 130622865-130622865 on Build GRCh38
rs775452429 Chr.9: 130622734-130622734 on Build GRCh38
rs952560860 Chr.9: 130623003-130623003 on Build GRCh38
rs1013279783 Chr.9: 130623134-130623134 on Build GRCh38
rs374074032 Chr.9: 130623040-130623040 on Build GRCh38
rs765214155 Chr.9: 130622944-130622944 on Build GRCh38
rs1049433099 Chr.9: 130622719-130622719 on Build GRCh38
rs777188942 Chr.9: 130623067-130623067 on Build GRCh38
rs764895842 Chr.9: 130622843-130622845 on Build GRCh38
rs759136227 Chr.9: 130622816-130622816 on Build GRCh38
rs757847767 Chr.9: 130622861-130622861 on Build GRCh38
rs78737313 Chr.9: 130622856-130622856 on Build GRCh38
rs1013341545 Chr.9: 130622891-130622891 on Build GRCh38
rs1057092270 Chr.9: 130623020-130623020 on Build GRCh38
rs75117961 Chr.9: 130623014-130623014 on Build GRCh38
rs534508624 Chr.9: 130622843-130622844 on Build GRCh38
rs1019796014 Chr.9: 130622909-130622909 on Build GRCh38
rs556701035 Chr.9: 130623061-130623061 on Build GRCh38
rs979829625 Chr.9: 130623006-130623006 on Build GRCh38
rs201114029 Chr.9: 130622726-130622726 on Build GRCh38
rs576484851 Chr.9: 130623138-130623138 on Build GRCh38
rs746045481 Chr.9: 130622716-130622716 on Build GRCh38
rs776398890 Chr.9: 130622814-130622814 on Build GRCh38
rs78083080 Chr.9: 130622855-130622857 on Build GRCh38
rs7860771 Chr.9: 130622896-130622896 on Build GRCh38
rs944742711 Chr.9: 130623002-130623002 on Build GRCh38
rs549556975 Chr.9: 130622766-130622766 on Build GRCh38
rs913116280 Chr.9: 130623050-130623050 on Build GRCh38
rs1008421105 Chr.9: 130622901-130622901 on Build GRCh38
rs191070044 Chr.9: 130623057-130623057 on Build GRCh38
rs764907010 Chr.9: 130622817-130622817 on Build GRCh38
rs781459739 Chr.9: 130622710-130622710 on Build GRCh38
rs542007455 Chr.9: 130622687-130622687 on Build GRCh38
rs1036322154 Chr.9: 130623091-130623091 on Build GRCh38
rs1023363831 Chr.9: 130623136-130623136 on Build GRCh38
rs757034072 Chr.9: 130622708-130622708 on Build GRCh38
rs762322895 Chr.9: 130622786-130622786 on Build GRCh38
rs757898620 Chr.9: 130623004-130623004 on Build GRCh38
rs766123843 Chr.9: 130622752-130622752 on Build GRCh38
rs527582757 Chr.9: 130623059-130623059 on Build GRCh38
rs773544852 Chr.9: 130622785-130622785 on Build GRCh38
rs752156038 Chr.9: 130622827-130622827 on Build GRCh38
rs796785870 Chr.9: 130622843-130622844 on Build GRCh38
rs973271985 Chr.9: 130622952-130622952 on Build GRCh38
rs548062651 Chr.9: 130623012-130623012 on Build GRCh38
rs76796364 Chr.9: 130623000-130623000 on Build GRCh38
rs896148474 Chr.9: 130623130-130623130 on Build GRCh38
rs769287763 Chr.9: 130622759-130622759 on Build GRCh38
rs957651784 Chr.9: 130622945-130622945 on Build GRCh38
rs903977129 Chr.9: 130623111-130623111 on Build GRCh38
rs903896445 Chr.9: 130622700-130622700 on Build GRCh38
rs770521538 Chr.9: 130622842-130622843 on Build GRCh38
rs773490513 Chr.9: 130622838-130622839 on Build GRCh38
rs557951079 Chr.9: 130622922-130622922 on Build GRCh38
rs575563176 Chr.9: 130622793-130622793 on Build GRCh38

View Full Product Details