Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154408848-154409342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1L1
Entrez Gene ID
1774
Gene Name
deoxyribonuclease I-like 1
Gene Aliases
DNAS1L1, DNASEX, DNL1L, G4.8, XIB
UniGene
Hs.401929
Gene Chromosome Location
Chr.X: 154401236-154412101 on Build GRCh38
Transcripts
NM_001009932.2 NM_001009933.2 NM_001009934.2 NM_001303620.1 NM_006730.3 XM_005277829.4 XM_011531122.2 XM_017029332.1

SNP Detail

SNP ID Chromosome Location
rs1002044696 Chr.X: 154408986-154408986 on Build GRCh38
rs965556650 Chr.X: 154409229-154409229 on Build GRCh38
rs782273504 Chr.X: 154409135-154409135 on Build GRCh38
rs944488476 Chr.X: 154408896-154408896 on Build GRCh38
rs782737706 Chr.X: 154409160-154409160 on Build GRCh38
rs4898486 Chr.X: 154409250-154409250 on Build GRCh38
rs995435252 Chr.X: 154409090-154409090 on Build GRCh38
rs896309415 Chr.X: 154409008-154409008 on Build GRCh38
rs34103533 Chr.X: 154409250-154409251 on Build GRCh38
rs782056443 Chr.X: 154409256-154409256 on Build GRCh38
rs368213748 Chr.X: 154409313-154409313 on Build GRCh38
rs1027197563 Chr.X: 154409107-154409107 on Build GRCh38
rs961374309 Chr.X: 154409089-154409089 on Build GRCh38
rs913593457 Chr.X: 154408940-154408940 on Build GRCh38
rs782082666 Chr.X: 154409112-154409112 on Build GRCh38
rs4898485 Chr.X: 154409242-154409242 on Build GRCh38
rs10127226 Chr.X: 154408926-154408926 on Build GRCh38
rs945031623 Chr.X: 154409315-154409315 on Build GRCh38
rs1015611078 Chr.X: 154409032-154409032 on Build GRCh38
rs913624809 Chr.X: 154409303-154409303 on Build GRCh38
rs1055406145 Chr.X: 154408997-154408997 on Build GRCh38
rs782803484 Chr.X: 154409314-154409314 on Build GRCh38
rs183937354 Chr.X: 154408985-154408985 on Build GRCh38
rs112063428 Chr.X: 154409235-154409235 on Build GRCh38
rs782001928 Chr.X: 154409016-154409016 on Build GRCh38
rs1014114527 Chr.X: 154409016-154409016 on Build GRCh38
rs781910991 Chr.X: 154409207-154409207 on Build GRCh38
rs985724269 Chr.X: 154409159-154409159 on Build GRCh38
rs2070807 Chr.X: 154408969-154408969 on Build GRCh38
rs4898487 Chr.X: 154409302-154409302 on Build GRCh38
rs1043675373 Chr.X: 154408974-154408974 on Build GRCh38

View Full Product Details