Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 51697189-51697721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF2T
Entrez Gene ID
23283
Gene Name
cleavage stimulation factor subunit 2, tau variant
Gene Aliases
CstF-64T
UniGene
Hs.591358
Gene Chromosome Location
Chr.10: 51695486-51699595 on Build GRCh38
Transcripts
NM_015235.2 NM_001098512.2 NM_006258.3 XM_011539952.2 XM_017016412.1 XM_017016413.1

Gene Symbol
PRKG1
Entrez Gene ID
5592
Gene Name
protein kinase, cGMP-dependent, type I
Gene Aliases
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha
UniGene
Hs.407535
Gene Chromosome Location
Chr.10: 50991151-52298350 on Build GRCh38
Transcripts
NM_015235.2 NM_001098512.2 NM_006258.3 XM_011539952.2 XM_017016412.1 XM_017016413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637047 c.1844C>T p.A615V Chr.10: 51697706-51697706 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056235824 Chr.10: 51697250-51697250 on Build GRCh38
rs759639363 Chr.10: 51697679-51697679 on Build GRCh38
rs745331818 Chr.10: 51697303-51697304 on Build GRCh38
rs938696673 Chr.10: 51697216-51697216 on Build GRCh38
rs537541770 Chr.10: 51697527-51697527 on Build GRCh38
rs751757473 Chr.10: 51697363-51697363 on Build GRCh38
rs753632908 Chr.10: 51697684-51697684 on Build GRCh38
rs973771094 Chr.10: 51697254-51697254 on Build GRCh38
rs763061471 Chr.10: 51697661-51697661 on Build GRCh38
rs961162425 Chr.10: 51697518-51697518 on Build GRCh38
rs566264949 Chr.10: 51697366-51697366 on Build GRCh38
rs139403403 Chr.10: 51697529-51697529 on Build GRCh38
rs901461692 Chr.10: 51697416-51697416 on Build GRCh38
rs1051322464 Chr.10: 51697337-51697337 on Build GRCh38
rs894544948 Chr.10: 51697299-51697299 on Build GRCh38
rs747587915 Chr.10: 51697529-51697529 on Build GRCh38
rs962445091 Chr.10: 51697213-51697213 on Build GRCh38
rs751830434 Chr.10: 51697675-51697675 on Build GRCh38
rs973803541 Chr.10: 51697545-51697545 on Build GRCh38
rs375932725 Chr.10: 51697654-51697654 on Build GRCh38
rs942834824 Chr.10: 51697350-51697350 on Build GRCh38
rs563529892 Chr.10: 51697340-51697340 on Build GRCh38
rs991485630 Chr.10: 51697551-51697551 on Build GRCh38
rs375755078 Chr.10: 51697471-51697475 on Build GRCh38
rs1043087392 Chr.10: 51697607-51697607 on Build GRCh38
rs763289523 Chr.10: 51697697-51697698 on Build GRCh38
rs367823598 Chr.10: 51697516-51697516 on Build GRCh38
rs757108476 Chr.10: 51697688-51697688 on Build GRCh38
rs962475101 Chr.10: 51697534-51697534 on Build GRCh38
rs1000903918 Chr.10: 51697454-51697454 on Build GRCh38
rs7908187 Chr.10: 51697374-51697374 on Build GRCh38
rs748874228 Chr.10: 51697472-51697472 on Build GRCh38
rs937956590 Chr.10: 51697530-51697530 on Build GRCh38
rs766347859 Chr.10: 51697670-51697670 on Build GRCh38
rs996517803 Chr.10: 51697433-51697433 on Build GRCh38
rs755829142 Chr.10: 51697410-51697410 on Build GRCh38
rs572448215 Chr.10: 51697562-51697562 on Build GRCh38
rs568925720 Chr.10: 51697500-51697500 on Build GRCh38
rs947480294 Chr.10: 51697321-51697321 on Build GRCh38
rs918258323 Chr.10: 51697546-51697549 on Build GRCh38
rs924701077 Chr.10: 51697625-51697625 on Build GRCh38
rs763957208 Chr.10: 51697682-51697682 on Build GRCh38
rs73343318 Chr.10: 51697331-51697331 on Build GRCh38
rs529479702 Chr.10: 51697345-51697345 on Build GRCh38
rs548585998 Chr.10: 51697505-51697506 on Build GRCh38
rs952430988 Chr.10: 51697482-51697482 on Build GRCh38
rs140943180 Chr.10: 51697636-51697636 on Build GRCh38
rs773843170 Chr.10: 51697501-51697501 on Build GRCh38
rs11601 Chr.10: 51697364-51697364 on Build GRCh38
rs182181937 Chr.10: 51697462-51697462 on Build GRCh38
rs915872436 Chr.10: 51697593-51697593 on Build GRCh38
rs796317370 Chr.10: 51697538-51697572 on Build GRCh38
rs557534493 Chr.10: 51697595-51697595 on Build GRCh38
rs947422212 Chr.10: 51697600-51697600 on Build GRCh38
rs770329927 Chr.10: 51697570-51697570 on Build GRCh38

View Full Product Details