Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14754921-14755143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE2
Entrez Gene ID
30817
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
CD312, EMR2, VBU
UniGene
Hs.531619
Gene Chromosome Location
Chr.19: 14732697-14778541 on Build GRCh38
Transcripts
NM_001271052.1 NM_013447.3 XM_011527948.2 XM_011527949.2 XM_011527951.2 XM_011527952.2 XM_011527953.2 XM_011527954.2 XM_011527955.2 XM_017026726.1 XM_017026727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1205290 c.1437G>T p.K479N Chr.19: 14755107-14755107 on Build GRCh38
6653913 c.1573G>A p.A525T Chr.19: 14754971-14754971 on Build GRCh38
330184 c.1564G>A p.V522I Chr.19: 14754980-14754980 on Build GRCh38
4930108 c.1565T>C p.V522A Chr.19: 14754979-14754979 on Build GRCh38
1636707 c.1552A>G p.S518G Chr.19: 14754992-14754992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764868326 Chr.19: 14755088-14755088 on Build GRCh38
rs762453696 Chr.19: 14754946-14754946 on Build GRCh38
rs768970857 Chr.19: 14755036-14755036 on Build GRCh38
rs199718602 Chr.19: 14755069-14755069 on Build GRCh38
rs746782632 Chr.19: 14754974-14754974 on Build GRCh38
rs768118665 Chr.19: 14754955-14754955 on Build GRCh38
rs937498481 Chr.19: 14755074-14755074 on Build GRCh38
rs563911043 Chr.19: 14754988-14754988 on Build GRCh38
rs150972875 Chr.19: 14755045-14755045 on Build GRCh38
rs545929404 Chr.19: 14755030-14755030 on Build GRCh38
rs750718286 Chr.19: 14755076-14755076 on Build GRCh38
rs771415140 Chr.19: 14755052-14755052 on Build GRCh38
rs752115546 Chr.19: 14755098-14755098 on Build GRCh38
rs112007450 Chr.19: 14755109-14755109 on Build GRCh38
rs144104308 Chr.19: 14755006-14755006 on Build GRCh38
rs745722678 Chr.19: 14754997-14754997 on Build GRCh38
rs150442380 Chr.19: 14755059-14755059 on Build GRCh38
rs138210363 Chr.19: 14755116-14755116 on Build GRCh38
rs758041727 Chr.19: 14754968-14754968 on Build GRCh38
rs143149076 Chr.19: 14754964-14754964 on Build GRCh38
rs1054685455 Chr.19: 14755003-14755003 on Build GRCh38
rs115480163 Chr.19: 14754990-14754990 on Build GRCh38
rs542436363 Chr.19: 14754980-14754980 on Build GRCh38
rs758093170 Chr.19: 14755106-14755106 on Build GRCh38
rs774906601 Chr.19: 14755044-14755044 on Build GRCh38
rs972666517 Chr.19: 14754981-14754981 on Build GRCh38
rs373173364 Chr.19: 14755117-14755117 on Build GRCh38
rs754549569 Chr.19: 14754963-14754963 on Build GRCh38
rs146763976 Chr.19: 14754982-14754982 on Build GRCh38
rs553690768 Chr.19: 14754959-14754959 on Build GRCh38
rs775421833 Chr.19: 14755013-14755013 on Build GRCh38
rs749594154 Chr.19: 14755017-14755017 on Build GRCh38
rs773773357 Chr.19: 14755068-14755068 on Build GRCh38
rs140501087 Chr.19: 14755047-14755047 on Build GRCh38
rs777582808 Chr.19: 14754973-14754973 on Build GRCh38
rs954008643 Chr.19: 14754962-14754962 on Build GRCh38
rs992977589 Chr.19: 14755112-14755112 on Build GRCh38
rs933287399 Chr.19: 14754954-14754954 on Build GRCh38

View Full Product Details